Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 63954525-63955032 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNE2
Entrez Gene ID
23224
Gene Name
spectrin repeat containing nuclear envelope protein 2
Gene Aliases
EDMD5, NUA, NUANCE, Nesp2, Nesprin-2, SYNE-2, TROPH
UniGene
Hs.745014
Gene Chromosome Location
Chr.14: 63761904-64226449 on Build GRCh38
Transcripts
NM_015180.4 NM_182914.2 XM_005267454.1 XM_005267456.1 XM_005267457.1 XM_005267458.1 XM_005267459.1 XM_011536574.1 XM_011536575.2 XM_011536576.2 XM_011536577.2 XM_011536578.1 XM_011536579.1 XM_011536580.1 XM_011536581.1 XM_011536582.1 XM_011536584.2 XM_017021101.1 XM_017021102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5543492 c.765C>T p.I255I Chr.14: 63954893-63954893 on Build GRCh38
698160 c.767C>A p.P256H Chr.14: 63954895-63954895 on Build GRCh38
6794931 c.689T>C p.M230T Chr.14: 63954817-63954817 on Build GRCh38
164744 c.670C>G p.R224G Chr.14: 63954798-63954798 on Build GRCh38
7236025 c.724C>A p.L242M Chr.14: 63954852-63954852 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758188083 Chr.14: 63954839-63954839 on Build GRCh38
rs765253920 Chr.14: 63954964-63954964 on Build GRCh38
rs1054289298 Chr.14: 63954982-63954982 on Build GRCh38
rs1023973644 Chr.14: 63954647-63954647 on Build GRCh38
rs1028288020 Chr.14: 63954694-63954694 on Build GRCh38
rs538031952 Chr.14: 63954603-63954603 on Build GRCh38
rs749724415 Chr.14: 63954863-63954863 on Build GRCh38
rs757292154 Chr.14: 63954809-63954809 on Build GRCh38
rs915784815 Chr.14: 63954989-63954989 on Build GRCh38
rs746518493 Chr.14: 63954733-63954733 on Build GRCh38
rs772723936 Chr.14: 63954883-63954883 on Build GRCh38
rs1027371955 Chr.14: 63954918-63954918 on Build GRCh38
rs765789486 Chr.14: 63954673-63954673 on Build GRCh38
rs755219747 Chr.14: 63954692-63954692 on Build GRCh38
rs557811623 Chr.14: 63954604-63954604 on Build GRCh38
rs771438833 Chr.14: 63954872-63954872 on Build GRCh38
rs759889097 Chr.14: 63954906-63954906 on Build GRCh38
rs761443945 Chr.14: 63954707-63954707 on Build GRCh38
rs944787012 Chr.14: 63954821-63954821 on Build GRCh38
rs756185673 Chr.14: 63954793-63954793 on Build GRCh38
rs750258352 Chr.14: 63954817-63954817 on Build GRCh38
rs768193199 Chr.14: 63954856-63954856 on Build GRCh38
rs775097875 Chr.14: 63954668-63954668 on Build GRCh38
rs370175849 Chr.14: 63954780-63954780 on Build GRCh38
rs376949083 Chr.14: 63954841-63954841 on Build GRCh38
rs577820160 Chr.14: 63954605-63954605 on Build GRCh38
rs370682546 Chr.14: 63954696-63954696 on Build GRCh38
rs752928692 Chr.14: 63954790-63954790 on Build GRCh38
rs368187566 Chr.14: 63954958-63954958 on Build GRCh38
rs777759540 Chr.14: 63954704-63954704 on Build GRCh38
rs756204464 Chr.14: 63954697-63954697 on Build GRCh38
rs539457886 Chr.14: 63954624-63954624 on Build GRCh38
rs118004985 Chr.14: 63954815-63954815 on Build GRCh38
rs893281284 Chr.14: 63954596-63954596 on Build GRCh38
rs766686907 Chr.14: 63954760-63954760 on Build GRCh38
rs763503726 Chr.14: 63954755-63954755 on Build GRCh38
rs778874832 Chr.14: 63954719-63954719 on Build GRCh38
rs772361135 Chr.14: 63954926-63954926 on Build GRCh38
rs762228781 Chr.14: 63954816-63954816 on Build GRCh38
rs766773069 Chr.14: 63954681-63954681 on Build GRCh38
rs757095973 Chr.14: 63954712-63954712 on Build GRCh38
rs764193579 Chr.14: 63954951-63954951 on Build GRCh38
rs971711814 Chr.14: 63954990-63954990 on Build GRCh38
rs764325987 Chr.14: 63954651-63954651 on Build GRCh38
rs767426656 Chr.14: 63954693-63954693 on Build GRCh38
rs187906577 Chr.14: 63954631-63954631 on Build GRCh38
rs765142996 Chr.14: 63954772-63954773 on Build GRCh38
rs753781068 Chr.14: 63954800-63954800 on Build GRCh38
rs974800972 Chr.14: 63954857-63954857 on Build GRCh38
rs527481262 Chr.14: 63954992-63954992 on Build GRCh38
rs373504858 Chr.14: 63954708-63954708 on Build GRCh38
rs900308203 Chr.14: 63954577-63954577 on Build GRCh38
rs751727620 Chr.14: 63954685-63954685 on Build GRCh38
rs1054741680 Chr.14: 63954616-63954616 on Build GRCh38
rs377436719 Chr.14: 63954721-63954721 on Build GRCh38
rs771762803 Chr.14: 63954720-63954720 on Build GRCh38
rs780707928 Chr.14: 63954858-63954858 on Build GRCh38
rs371326813 Chr.14: 63954881-63954881 on Build GRCh38
rs764128650 Chr.14: 63954799-63954799 on Build GRCh38
rs776719014 Chr.14: 63954671-63954672 on Build GRCh38
rs575795574 Chr.14: 63954866-63954866 on Build GRCh38
rs1033820272 Chr.14: 63954667-63954667 on Build GRCh38
rs773742506 Chr.14: 63954741-63954741 on Build GRCh38
rs374295389 Chr.14: 63954930-63954930 on Build GRCh38
rs763307743 Chr.14: 63954677-63954677 on Build GRCh38
rs762242596 Chr.14: 63954641-63954641 on Build GRCh38
rs746684931 Chr.14: 63954845-63954845 on Build GRCh38
rs991263385 Chr.14: 63954929-63954931 on Build GRCh38
rs375642734 Chr.14: 63954788-63954788 on Build GRCh38
rs35773099 Chr.14: 63954668-63954669 on Build GRCh38
rs1255874 Chr.14: 63954674-63954674 on Build GRCh38
rs1000133225 Chr.14: 63954609-63954609 on Build GRCh38
rs1054563751 Chr.14: 63954575-63954575 on Build GRCh38
rs1001224475 Chr.14: 63954654-63954654 on Build GRCh38
rs768120109 Chr.14: 63954735-63954735 on Build GRCh38
rs775758502 Chr.14: 63954927-63954927 on Build GRCh38
rs1010352429 Chr.14: 63954640-63954640 on Build GRCh38
rs759694907 Chr.14: 63954782-63954782 on Build GRCh38
rs935787094 Chr.14: 63954900-63954900 on Build GRCh38

View Full Product Details