Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 65093939-65094444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100506321
Entrez Gene ID
100506321
Gene Name
uncharacterized LOC100506321
Gene Aliases
-
UniGene
Hs.711267
Gene Chromosome Location
Chr.14: 65089918-65094311 on Build GRCh38
Transcripts
NR_045122.1 NM_001271068.1 NM_001271069.1 NM_001320415.1 NM_002382.4 NM_145112.2 NM_145113.2 NM_145114.2 NM_197957.3 NR_073137.1 NR_073138.1 XM_011536773.2 XM_017021312.1 XM_017021313.1

Gene Symbol
MAX
Entrez Gene ID
4149
Gene Name
MYC associated factor X
Gene Aliases
bHLHd4
UniGene
Hs.285354
Gene Chromosome Location
Chr.14: 65006101-65102695 on Build GRCh38
Transcripts
NR_045122.1 NM_001271068.1 NM_001271069.1 NM_001320415.1 NM_002382.4 NM_145112.2 NM_145113.2 NM_145114.2 NM_197957.3 NR_073137.1 NR_073138.1 XM_011536773.2 XM_017021312.1 XM_017021313.1

SNP Detail

SNP ID Chromosome Location
rs1006160960 Chr.14: 65094382-65094382 on Build GRCh38
rs546258912 Chr.14: 65094187-65094187 on Build GRCh38
rs73272603 Chr.14: 65094396-65094396 on Build GRCh38
rs567328022 Chr.14: 65094014-65094014 on Build GRCh38
rs72728280 Chr.14: 65094266-65094266 on Build GRCh38
rs558419863 Chr.14: 65094121-65094121 on Build GRCh38
rs947675684 Chr.14: 65094189-65094189 on Build GRCh38
rs981047939 Chr.14: 65094010-65094010 on Build GRCh38
rs184803066 Chr.14: 65093984-65093984 on Build GRCh38
rs954749274 Chr.14: 65094164-65094164 on Build GRCh38
rs139829219 Chr.14: 65094089-65094089 on Build GRCh38
rs775135120 Chr.14: 65094015-65094015 on Build GRCh38
rs897163823 Chr.14: 65094169-65094169 on Build GRCh38
rs971192504 Chr.14: 65093995-65093995 on Build GRCh38
rs917676738 Chr.14: 65094313-65094313 on Build GRCh38
rs1026198535 Chr.14: 65094026-65094026 on Build GRCh38
rs762873859 Chr.14: 65094109-65094109 on Build GRCh38
rs571502690 Chr.14: 65094374-65094374 on Build GRCh38
rs913451755 Chr.14: 65094186-65094186 on Build GRCh38
rs912740750 Chr.14: 65094412-65094412 on Build GRCh38
rs571497372 Chr.14: 65093962-65093962 on Build GRCh38
rs551325249 Chr.14: 65094351-65094351 on Build GRCh38
rs553520868 Chr.14: 65094008-65094008 on Build GRCh38
rs914864085 Chr.14: 65094116-65094116 on Build GRCh38
rs1013499977 Chr.14: 65094023-65094023 on Build GRCh38
rs1037002679 Chr.14: 65094134-65094134 on Build GRCh38
rs995548852 Chr.14: 65094174-65094174 on Build GRCh38
rs1023122428 Chr.14: 65093993-65093993 on Build GRCh38
rs181597943 Chr.14: 65094352-65094352 on Build GRCh38
rs531531723 Chr.14: 65094321-65094321 on Build GRCh38
rs527612290 Chr.14: 65094380-65094380 on Build GRCh38
rs1049002376 Chr.14: 65094206-65094206 on Build GRCh38
rs578214032 Chr.14: 65094154-65094154 on Build GRCh38
rs145064218 Chr.14: 65094298-65094298 on Build GRCh38
rs1013143066 Chr.14: 65094212-65094212 on Build GRCh38
rs944150249 Chr.14: 65094129-65094129 on Build GRCh38
rs1049950703 Chr.14: 65094122-65094122 on Build GRCh38
rs1030281859 Chr.14: 65094124-65094124 on Build GRCh38
rs1054774792 Chr.14: 65093991-65093991 on Build GRCh38
rs761867397 Chr.14: 65094318-65094318 on Build GRCh38
rs539680908 Chr.14: 65094309-65094309 on Build GRCh38
rs914791217 Chr.14: 65094361-65094361 on Build GRCh38
rs573991215 Chr.14: 65094009-65094009 on Build GRCh38
rs768344807 Chr.14: 65094258-65094258 on Build GRCh38
rs576948338 Chr.14: 65094066-65094066 on Build GRCh38
rs189218275 Chr.14: 65094211-65094211 on Build GRCh38
rs529368957 Chr.14: 65094282-65094282 on Build GRCh38
rs1006440918 Chr.14: 65094056-65094056 on Build GRCh38
rs147456119 Chr.14: 65094040-65094040 on Build GRCh38
rs988774628 Chr.14: 65094184-65094184 on Build GRCh38
rs979313649 Chr.14: 65094233-65094233 on Build GRCh38
rs555500024 Chr.14: 65094055-65094055 on Build GRCh38
rs986004176 Chr.14: 65094397-65094397 on Build GRCh38

View Full Product Details