Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 74863378-74863886 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PROX2
Entrez Gene ID
283571
Gene Name
prospero homeobox 2
Gene Aliases
PROX-2
UniGene
Hs.675293
Gene Chromosome Location
Chr.14: 74853033-74876663 on Build GRCh38
Transcripts
NM_001080408.2 NM_001243007.1 XM_005267543.3 XM_017021222.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5900793 c.370G>A p.G124S Chr.14: 74863465-74863465 on Build GRCh38
957869 c.299G>A p.R100Q Chr.14: 74863536-74863536 on Build GRCh38
3936621 c.281G>A p.R94H Chr.14: 74863554-74863554 on Build GRCh38
4776371 c.205G>C p.E69Q Chr.14: 74863630-74863630 on Build GRCh38
7300923 c.75C>T p.G25G Chr.14: 74863760-74863760 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774572009 Chr.14: 74863482-74863482 on Build GRCh38
rs564000021 Chr.14: 74863443-74863443 on Build GRCh38
rs772746725 Chr.14: 74863690-74863690 on Build GRCh38
rs751325893 Chr.14: 74863510-74863510 on Build GRCh38
rs749601374 Chr.14: 74863775-74863775 on Build GRCh38
rs765862135 Chr.14: 74863698-74863698 on Build GRCh38
rs749072521 Chr.14: 74863465-74863465 on Build GRCh38
rs376935828 Chr.14: 74863660-74863660 on Build GRCh38
rs372540137 Chr.14: 74863444-74863444 on Build GRCh38
rs756816458 Chr.14: 74863514-74863514 on Build GRCh38
rs570245599 Chr.14: 74863555-74863555 on Build GRCh38
rs753787443 Chr.14: 74863608-74863608 on Build GRCh38
rs751793935 Chr.14: 74863745-74863745 on Build GRCh38
rs774127039 Chr.14: 74863578-74863578 on Build GRCh38
rs374677617 Chr.14: 74863730-74863730 on Build GRCh38
rs754775136 Chr.14: 74863551-74863551 on Build GRCh38
rs936587323 Chr.14: 74863800-74863800 on Build GRCh38
rs773315825 Chr.14: 74863558-74863558 on Build GRCh38
rs777017206 Chr.14: 74863575-74863575 on Build GRCh38
rs767956938 Chr.14: 74863405-74863405 on Build GRCh38
rs117853159 Chr.14: 74863603-74863603 on Build GRCh38
rs199570102 Chr.14: 74863611-74863611 on Build GRCh38
rs200363606 Chr.14: 74863599-74863599 on Build GRCh38
rs201978225 Chr.14: 74863759-74863759 on Build GRCh38
rs368123183 Chr.14: 74863767-74863767 on Build GRCh38
rs773692074 Chr.14: 74863493-74863493 on Build GRCh38
rs541255850 Chr.14: 74863657-74863657 on Build GRCh38
rs1015947940 Chr.14: 74863615-74863615 on Build GRCh38
rs529608422 Chr.14: 74863449-74863449 on Build GRCh38
rs184323713 Chr.14: 74863553-74863553 on Build GRCh38
rs770375655 Chr.14: 74863769-74863769 on Build GRCh38
rs61747287 Chr.14: 74863502-74863502 on Build GRCh38
rs760230963 Chr.14: 74863695-74863695 on Build GRCh38
rs988495942 Chr.14: 74863860-74863860 on Build GRCh38
rs770674279 Chr.14: 74863747-74863747 on Build GRCh38
rs749057074 Chr.14: 74863537-74863537 on Build GRCh38
rs955078773 Chr.14: 74863668-74863668 on Build GRCh38
rs777879727 Chr.14: 74863626-74863626 on Build GRCh38
rs763779614 Chr.14: 74863506-74863506 on Build GRCh38
rs750931588 Chr.14: 74863432-74863432 on Build GRCh38
rs760291293 Chr.14: 74863796-74863796 on Build GRCh38
rs975869773 Chr.14: 74863751-74863751 on Build GRCh38
rs751064033 Chr.14: 74863446-74863446 on Build GRCh38
rs943339551 Chr.14: 74863501-74863501 on Build GRCh38
rs770440534 Chr.14: 74863826-74863826 on Build GRCh38
rs780724661 Chr.14: 74863774-74863774 on Build GRCh38
rs201405244 Chr.14: 74863617-74863617 on Build GRCh38
rs745876721 Chr.14: 74863451-74863451 on Build GRCh38
rs777983317 Chr.14: 74863736-74863736 on Build GRCh38
rs375166231 Chr.14: 74863766-74863766 on Build GRCh38
rs775957531 Chr.14: 74863675-74863675 on Build GRCh38
rs199595015 Chr.14: 74863811-74863811 on Build GRCh38
rs146941408 Chr.14: 74863780-74863780 on Build GRCh38
rs749903509 Chr.14: 74863598-74863598 on Build GRCh38
rs759506818 Chr.14: 74863718-74863718 on Build GRCh38
rs115990005 Chr.14: 74863455-74863455 on Build GRCh38
rs767186286 Chr.14: 74863518-74863518 on Build GRCh38
rs747052181 Chr.14: 74863635-74863635 on Build GRCh38
rs756042170 Chr.14: 74863532-74863532 on Build GRCh38
rs373721677 Chr.14: 74863637-74863637 on Build GRCh38
rs770252889 Chr.14: 74863661-74863661 on Build GRCh38
rs189269863 Chr.14: 74863556-74863556 on Build GRCh38
rs768877818 Chr.14: 74863469-74863469 on Build GRCh38
rs753788918 Chr.14: 74863701-74863701 on Build GRCh38
rs61740783 Chr.14: 74863454-74863454 on Build GRCh38
rs919913714 Chr.14: 74863522-74863522 on Build GRCh38
rs766821053 Chr.14: 74863440-74863440 on Build GRCh38
rs370728105 Chr.14: 74863623-74863623 on Build GRCh38
rs376608651 Chr.14: 74863795-74863795 on Build GRCh38
rs772203484 Chr.14: 74863492-74863492 on Build GRCh38
rs978709446 Chr.14: 74863782-74863782 on Build GRCh38
rs771419514 Chr.14: 74863574-74863574 on Build GRCh38
rs750298029 Chr.14: 74863529-74863529 on Build GRCh38
rs371637936 Chr.14: 74863719-74863719 on Build GRCh38
rs372079820 Chr.14: 74863794-74863794 on Build GRCh38
rs748335827 Chr.14: 74863622-74863622 on Build GRCh38
rs779882928 Chr.14: 74863536-74863536 on Build GRCh38
rs748423943 Chr.14: 74863554-74863554 on Build GRCh38
rs555931182 Chr.14: 74863585-74863585 on Build GRCh38
rs180960227 Chr.14: 74863732-74863732 on Build GRCh38
rs994178240 Chr.14: 74863618-74863618 on Build GRCh38
rs776074689 Chr.14: 74863863-74863863 on Build GRCh38
rs773080155 Chr.14: 74863644-74863645 on Build GRCh38
rs866413865 Chr.14: 74863550-74863550 on Build GRCh38
rs767310766 Chr.14: 74863588-74863588 on Build GRCh38
rs761816812 Chr.14: 74863488-74863488 on Build GRCh38
rs759839583 Chr.14: 74863504-74863504 on Build GRCh38
rs952688966 Chr.14: 74863783-74863783 on Build GRCh38
rs771027207 Chr.14: 74863577-74863577 on Build GRCh38
rs371967421 Chr.14: 74863793-74863793 on Build GRCh38
rs200595982 Chr.14: 74863680-74863680 on Build GRCh38
rs777014202 Chr.14: 74863503-74863503 on Build GRCh38
rs374756212 Chr.14: 74863559-74863559 on Build GRCh38
rs762885845 Chr.14: 74863657-74863658 on Build GRCh38

View Full Product Details