Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104770658-104771161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKT1
Entrez Gene ID
207
Gene Name
AKT serine/threonine kinase 1
Gene Aliases
AKT, CWS6, PKB, PKB-ALPHA, PRKBA, RAC, RAC-ALPHA
UniGene
Hs.525622
Gene Chromosome Location
Chr.14: 104769349-104795743 on Build GRCh38
Transcripts
NM_001014431.1 NM_001014432.1 NM_005163.2 XM_005267401.1 XM_017021075.1 XM_017021076.1 XM_017021077.1 XM_017021078.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7343476 c.1338G>T p.M446I Chr.14: 104770770-104770770 on Build GRCh38
6521850 c.1290G>T p.T430T Chr.14: 104770818-104770818 on Build GRCh38
4990661 c.1304C>T p.T435I Chr.14: 104770804-104770804 on Build GRCh38
6966503 c.1339A>T p.I447F Chr.14: 104770769-104770769 on Build GRCh38
5760502 c.1328C>T p.T443M Chr.14: 104770780-104770780 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs939532465 Chr.14: 104770935-104770935 on Build GRCh38
rs112458264 Chr.14: 104770744-104770744 on Build GRCh38
rs887211887 Chr.14: 104771136-104771136 on Build GRCh38
rs775499085 Chr.14: 104770877-104770877 on Build GRCh38
rs1005542207 Chr.14: 104770683-104770683 on Build GRCh38
rs766768210 Chr.14: 104770725-104770725 on Build GRCh38
rs764632008 Chr.14: 104770882-104770882 on Build GRCh38
rs150070451 Chr.14: 104770690-104770693 on Build GRCh38
rs546083181 Chr.14: 104771081-104771081 on Build GRCh38
rs778840250 Chr.14: 104770862-104770862 on Build GRCh38
rs1021884343 Chr.14: 104771127-104771127 on Build GRCh38
rs936240850 Chr.14: 104770922-104770922 on Build GRCh38
rs1004194081 Chr.14: 104770974-104770974 on Build GRCh38
rs1016839015 Chr.14: 104770694-104770694 on Build GRCh38
rs751976958 Chr.14: 104770770-104770770 on Build GRCh38
rs887183976 Chr.14: 104770969-104770969 on Build GRCh38
rs771248358 Chr.14: 104770867-104770867 on Build GRCh38
rs913653954 Chr.14: 104770846-104770846 on Build GRCh38
rs750104452 Chr.14: 104770815-104770815 on Build GRCh38
rs998679491 Chr.14: 104770956-104770956 on Build GRCh38
rs762121960 Chr.14: 104770884-104770884 on Build GRCh38
rs980971868 Chr.14: 104770928-104770928 on Build GRCh38
rs574734523 Chr.14: 104770997-104770997 on Build GRCh38
rs763502372 Chr.14: 104770878-104770878 on Build GRCh38
rs926451222 Chr.14: 104770736-104770736 on Build GRCh38
rs780664980 Chr.14: 104770695-104770695 on Build GRCh38
rs868494963 Chr.14: 104770949-104770949 on Build GRCh38
rs201227909 Chr.14: 104770839-104770839 on Build GRCh38
rs753190812 Chr.14: 104770753-104770753 on Build GRCh38
rs576436971 Chr.14: 104771039-104771039 on Build GRCh38
rs752371856 Chr.14: 104770995-104770995 on Build GRCh38
rs989109891 Chr.14: 104771100-104771100 on Build GRCh38
rs531562828 Chr.14: 104771077-104771077 on Build GRCh38
rs1039917192 Chr.14: 104771083-104771083 on Build GRCh38
rs776954420 Chr.14: 104770875-104770875 on Build GRCh38
rs545796299 Chr.14: 104771067-104771067 on Build GRCh38
rs765264778 Chr.14: 104770749-104770749 on Build GRCh38
rs749201160 Chr.14: 104770717-104770717 on Build GRCh38
rs780860941 Chr.14: 104770803-104770803 on Build GRCh38
rs962641587 Chr.14: 104770697-104770697 on Build GRCh38
rs1051161505 Chr.14: 104770960-104770960 on Build GRCh38
rs745616456 Chr.14: 104771108-104771108 on Build GRCh38
rs749196395 Chr.14: 104771058-104771058 on Build GRCh38
rs764514218 Chr.14: 104770769-104770769 on Build GRCh38
rs1029741935 Chr.14: 104771055-104771055 on Build GRCh38
rs768546900 Chr.14: 104770856-104770856 on Build GRCh38
rs760977150 Chr.14: 104770722-104770722 on Build GRCh38
rs765680490 Chr.14: 104770858-104770859 on Build GRCh38
rs755597789 Chr.14: 104770816-104770816 on Build GRCh38
rs941359046 Chr.14: 104771028-104771028 on Build GRCh38
rs554693082 Chr.14: 104770937-104770937 on Build GRCh38
rs376893212 Chr.14: 104770779-104770779 on Build GRCh38
rs769584040 Chr.14: 104770711-104770711 on Build GRCh38
rs759702315 Chr.14: 104770747-104770747 on Build GRCh38
rs538965403 Chr.14: 104770715-104770715 on Build GRCh38
rs758833789 Chr.14: 104770764-104770764 on Build GRCh38
rs368913986 Chr.14: 104770692-104770695 on Build GRCh38
rs749186394 Chr.14: 104770819-104770819 on Build GRCh38
rs540900119 Chr.14: 104770917-104770917 on Build GRCh38
rs915011347 Chr.14: 104770900-104770900 on Build GRCh38
rs376284145 Chr.14: 104770865-104770865 on Build GRCh38
rs368238100 Chr.14: 104770876-104770876 on Build GRCh38
rs537928294 Chr.14: 104770914-104770914 on Build GRCh38
rs772039336 Chr.14: 104771129-104771129 on Build GRCh38
rs768444367 Chr.14: 104770719-104770719 on Build GRCh38
rs750738778 Chr.14: 104770869-104770870 on Build GRCh38
rs913683627 Chr.14: 104771051-104771051 on Build GRCh38
rs745428582 Chr.14: 104770707-104770707 on Build GRCh38
rs956600865 Chr.14: 104771098-104771098 on Build GRCh38
rs1040287701 Chr.14: 104770934-104770934 on Build GRCh38
rs757685843 Chr.14: 104770708-104770709 on Build GRCh38
rs997371288 Chr.14: 104771045-104771045 on Build GRCh38
rs7140735 Chr.14: 104771064-104771064 on Build GRCh38
rs1002093714 Chr.14: 104771002-104771002 on Build GRCh38
rs1016867432 Chr.14: 104770996-104770996 on Build GRCh38
rs397514645 Chr.14: 104770805-104770805 on Build GRCh38
rs1051191020 Chr.14: 104771135-104771135 on Build GRCh38
rs562656821 Chr.14: 104771068-104771068 on Build GRCh38
rs61761248 Chr.14: 104770733-104770733 on Build GRCh38
rs934193828 Chr.14: 104771111-104771111 on Build GRCh38
rs774295573 Chr.14: 104770721-104770721 on Build GRCh38
rs540582046 Chr.14: 104771031-104771031 on Build GRCh38
rs751887179 Chr.14: 104770883-104770883 on Build GRCh38
rs778972564 Chr.14: 104771088-104771088 on Build GRCh38
rs1033935937 Chr.14: 104771024-104771024 on Build GRCh38
rs779447501 Chr.14: 104770818-104770818 on Build GRCh38
rs118098425 Chr.14: 104771043-104771043 on Build GRCh38
rs989078773 Chr.14: 104770891-104770891 on Build GRCh38

View Full Product Details