Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104772778-104773281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKT1
Entrez Gene ID
207
Gene Name
AKT serine/threonine kinase 1
Gene Aliases
AKT, CWS6, PKB, PKB-ALPHA, PRKBA, RAC, RAC-ALPHA
UniGene
Hs.525622
Gene Chromosome Location
Chr.14: 104769349-104795743 on Build GRCh38
Transcripts
NM_001014431.1 NM_001014432.1 NM_005163.2 XM_005267401.1 XM_017021075.1 XM_017021076.1 XM_017021077.1 XM_017021078.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5044339 c.1069C>T p.L357F Chr.14: 104772981-104772981 on Build GRCh38
6981608 c.1061A>T p.H354L Chr.14: 104772989-104772989 on Build GRCh38
6929635 c.958G>T p.V320L Chr.14: 104773092-104773092 on Build GRCh38
6979438 c.1106C>T p.P369L Chr.14: 104772944-104772944 on Build GRCh38
5044337 c.1042C>T p.P348S Chr.14: 104773008-104773008 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751709437 Chr.14: 104773063-104773063 on Build GRCh38
rs372502847 Chr.14: 104772963-104772963 on Build GRCh38
rs976500042 Chr.14: 104772999-104772999 on Build GRCh38
rs946932540 Chr.14: 104773190-104773190 on Build GRCh38
rs377758228 Chr.14: 104773224-104773224 on Build GRCh38
rs924241695 Chr.14: 104773155-104773155 on Build GRCh38
rs770846644 Chr.14: 104773167-104773167 on Build GRCh38
rs781749630 Chr.14: 104773077-104773077 on Build GRCh38
rs778954967 Chr.14: 104772844-104772844 on Build GRCh38
rs748278837 Chr.14: 104772845-104772845 on Build GRCh38
rs758624702 Chr.14: 104772892-104772894 on Build GRCh38
rs2494732 Chr.14: 104772855-104772855 on Build GRCh38
rs766097509 Chr.14: 104773138-104773138 on Build GRCh38
rs771516815 Chr.14: 104773120-104773120 on Build GRCh38
rs369520527 Chr.14: 104772950-104772950 on Build GRCh38
rs767544462 Chr.14: 104772991-104772991 on Build GRCh38
rs750891852 Chr.14: 104773223-104773223 on Build GRCh38
rs760620929 Chr.14: 104773137-104773137 on Build GRCh38
rs746272761 Chr.14: 104772941-104772941 on Build GRCh38
rs915468534 Chr.14: 104773182-104773182 on Build GRCh38
rs747575253 Chr.14: 104772854-104772854 on Build GRCh38
rs952698197 Chr.14: 104772872-104772872 on Build GRCh38
rs755174756 Chr.14: 104772904-104772904 on Build GRCh38
rs747781896 Chr.14: 104773237-104773237 on Build GRCh38
rs201826439 Chr.14: 104773195-104773195 on Build GRCh38
rs972532841 Chr.14: 104772847-104772848 on Build GRCh38
rs757310859 Chr.14: 104773216-104773216 on Build GRCh38
rs374271808 Chr.14: 104772814-104772814 on Build GRCh38
rs964585008 Chr.14: 104772817-104772817 on Build GRCh38
rs1043166019 Chr.14: 104773039-104773039 on Build GRCh38
rs761646283 Chr.14: 104772988-104772988 on Build GRCh38
rs773553859 Chr.14: 104772847-104772847 on Build GRCh38
rs893937676 Chr.14: 104772861-104772861 on Build GRCh38
rs529348439 Chr.14: 104772869-104772869 on Build GRCh38
rs549370342 Chr.14: 104772942-104772942 on Build GRCh38
rs370843446 Chr.14: 104773225-104773225 on Build GRCh38
rs369048965 Chr.14: 104773099-104773099 on Build GRCh38
rs573290939 Chr.14: 104773220-104773220 on Build GRCh38
rs769619173 Chr.14: 104772864-104772864 on Build GRCh38
rs763680129 Chr.14: 104772871-104772871 on Build GRCh38
rs12881616 Chr.14: 104773252-104773252 on Build GRCh38
rs772539695 Chr.14: 104772846-104772846 on Build GRCh38
rs144088506 Chr.14: 104773030-104773030 on Build GRCh38
rs749553301 Chr.14: 104773163-104773163 on Build GRCh38
rs759249014 Chr.14: 104772862-104772862 on Build GRCh38
rs780293460 Chr.14: 104773259-104773259 on Build GRCh38
rs55874341 Chr.14: 104773072-104773072 on Build GRCh38
rs751320716 Chr.14: 104773260-104773260 on Build GRCh38
rs3803304 Chr.14: 104772809-104772809 on Build GRCh38
rs562284476 Chr.14: 104772801-104772801 on Build GRCh38
rs754868168 Chr.14: 104773134-104773134 on Build GRCh38
rs746397537 Chr.14: 104773079-104773079 on Build GRCh38
rs763062980 Chr.14: 104773205-104773205 on Build GRCh38
rs369527283 Chr.14: 104772916-104772916 on Build GRCh38
rs779060318 Chr.14: 104773215-104773215 on Build GRCh38
rs35314414 Chr.14: 104773233-104773233 on Build GRCh38
rs536727818 Chr.14: 104773108-104773108 on Build GRCh38
rs748884296 Chr.14: 104773100-104773100 on Build GRCh38
rs935586799 Chr.14: 104773162-104773162 on Build GRCh38
rs374865919 Chr.14: 104772928-104772928 on Build GRCh38
rs749400277 Chr.14: 104772829-104772829 on Build GRCh38
rs200669248 Chr.14: 104773194-104773194 on Build GRCh38
rs199770031 Chr.14: 104773098-104773098 on Build GRCh38
rs373253729 Chr.14: 104772927-104772927 on Build GRCh38
rs768409564 Chr.14: 104773105-104773105 on Build GRCh38
rs981118779 Chr.14: 104773154-104773154 on Build GRCh38
rs781388586 Chr.14: 104772938-104772938 on Build GRCh38
rs773975370 Chr.14: 104772958-104772958 on Build GRCh38
rs749713865 Chr.14: 104773230-104773230 on Build GRCh38
rs772715345 Chr.14: 104773247-104773247 on Build GRCh38
rs777711698 Chr.14: 104773060-104773060 on Build GRCh38
rs777807799 Chr.14: 104773148-104773148 on Build GRCh38
rs370287868 Chr.14: 104773133-104773133 on Build GRCh38
rs767207905 Chr.14: 104772901-104772901 on Build GRCh38
rs374446810 Chr.14: 104773208-104773208 on Build GRCh38
rs895830349 Chr.14: 104773172-104773172 on Build GRCh38
rs757112659 Chr.14: 104773121-104773121 on Build GRCh38
rs56289559 Chr.14: 104773018-104773018 on Build GRCh38
rs200702576 Chr.14: 104772868-104772868 on Build GRCh38
rs1045407348 Chr.14: 104772820-104772820 on Build GRCh38
rs776477466 Chr.14: 104773143-104773143 on Build GRCh38
rs889182971 Chr.14: 104772932-104772932 on Build GRCh38
rs761810870 Chr.14: 104772892-104772892 on Build GRCh38
rs1056050445 Chr.14: 104773149-104773149 on Build GRCh38
rs771514399 Chr.14: 104773244-104773244 on Build GRCh38
rs777059370 Chr.14: 104772860-104772860 on Build GRCh38
rs11555432 Chr.14: 104772980-104772980 on Build GRCh38
rs944763702 Chr.14: 104773218-104773218 on Build GRCh38
rs150399268 Chr.14: 104772943-104772943 on Build GRCh38
rs751392710 Chr.14: 104772873-104772873 on Build GRCh38
rs376942361 Chr.14: 104772997-104772997 on Build GRCh38
rs35622471 Chr.14: 104772807-104772807 on Build GRCh38
rs755264592 Chr.14: 104772840-104772840 on Build GRCh38
rs935543623 Chr.14: 104772850-104772850 on Build GRCh38
rs773088242 Chr.14: 104773129-104773129 on Build GRCh38
rs764416293 Chr.14: 104773207-104773207 on Build GRCh38
rs191423993 Chr.14: 104773234-104773234 on Build GRCh38
rs1040361009 Chr.14: 104773221-104773221 on Build GRCh38
rs1050565251 Chr.14: 104772924-104772924 on Build GRCh38
rs758651475 Chr.14: 104773045-104773045 on Build GRCh38
rs746944273 Chr.14: 104773254-104773254 on Build GRCh38
rs760425021 Chr.14: 104773000-104773000 on Build GRCh38
rs533427933 Chr.14: 104772823-104772823 on Build GRCh38
rs747408700 Chr.14: 104772937-104772937 on Build GRCh38
rs11555431 Chr.14: 104772888-104772888 on Build GRCh38
rs747738213 Chr.14: 104773111-104773111 on Build GRCh38
rs17846831 Chr.14: 104773257-104773257 on Build GRCh38

View Full Product Details