Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104865002-104865498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP170B
Entrez Gene ID
283638
Gene Name
centrosomal protein 170B
Gene Aliases
CEP170R, FAM68C, KIAA0284
UniGene
Hs.533721
Gene Chromosome Location
Chr.14: 104865313-104896770 on Build GRCh38
Transcripts
NM_001112726.2 NM_015005.2 XM_005267550.3 XM_011536665.2

SNP Detail

SNP ID Chromosome Location
rs1006665640 Chr.14: 104865068-104865068 on Build GRCh38
rs1024272937 Chr.14: 104865392-104865395 on Build GRCh38
rs1033667791 Chr.14: 104865340-104865340 on Build GRCh38
rs2841221 Chr.14: 104865129-104865129 on Build GRCh38
rs971505917 Chr.14: 104865433-104865433 on Build GRCh38
rs1033051936 Chr.14: 104865124-104865124 on Build GRCh38
rs979414000 Chr.14: 104865118-104865118 on Build GRCh38
rs867545016 Chr.14: 104865135-104865135 on Build GRCh38
rs903349425 Chr.14: 104865336-104865336 on Build GRCh38
rs1039319616 Chr.14: 104865280-104865280 on Build GRCh38
rs1041904606 Chr.14: 104865324-104865324 on Build GRCh38
rs576225070 Chr.14: 104865110-104865110 on Build GRCh38
rs1047945175 Chr.14: 104865445-104865468 on Build GRCh38
rs1020373144 Chr.14: 104865102-104865102 on Build GRCh38
rs185665842 Chr.14: 104865160-104865160 on Build GRCh38
rs540664791 Chr.14: 104865151-104865151 on Build GRCh38
rs987834706 Chr.14: 104865234-104865234 on Build GRCh38
rs898080524 Chr.14: 104865071-104865071 on Build GRCh38
rs888108382 Chr.14: 104865026-104865026 on Build GRCh38
rs1012941554 Chr.14: 104865375-104865375 on Build GRCh38
rs997854496 Chr.14: 104865289-104865289 on Build GRCh38
rs1000477212 Chr.14: 104865338-104865338 on Build GRCh38
rs571958317 Chr.14: 104865150-104865150 on Build GRCh38
rs956601447 Chr.14: 104865217-104865217 on Build GRCh38
rs1032297371 Chr.14: 104865213-104865213 on Build GRCh38
rs929948561 Chr.14: 104865261-104865261 on Build GRCh38
rs1017968173 Chr.14: 104865069-104865069 on Build GRCh38
rs1037878176 Chr.14: 104865066-104865066 on Build GRCh38
rs900931060 Chr.14: 104865284-104865284 on Build GRCh38
rs1048770733 Chr.14: 104865265-104865265 on Build GRCh38
rs8008126 Chr.14: 104865120-104865120 on Build GRCh38
rs865907219 Chr.14: 104865257-104865257 on Build GRCh38
rs909577147 Chr.14: 104865274-104865274 on Build GRCh38
rs868414451 Chr.14: 104865192-104865192 on Build GRCh38
rs1000783984 Chr.14: 104865208-104865208 on Build GRCh38
rs942372160 Chr.14: 104865280-104865280 on Build GRCh38
rs929345643 Chr.14: 104865076-104865076 on Build GRCh38
rs998329747 Chr.14: 104865101-104865101 on Build GRCh38
rs867122709 Chr.14: 104865086-104865086 on Build GRCh38
rs1057049617 Chr.14: 104865087-104865087 on Build GRCh38
rs866817225 Chr.14: 104865352-104865352 on Build GRCh38
rs2841222 Chr.14: 104865146-104865146 on Build GRCh38
rs548529289 Chr.14: 104865391-104865392 on Build GRCh38
rs958958855 Chr.14: 104865347-104865347 on Build GRCh38
rs981944076 Chr.14: 104865254-104865254 on Build GRCh38
rs868850040 Chr.14: 104865449-104865449 on Build GRCh38

View Full Product Details