Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34527261-34527758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA8B
Entrez Gene ID
440270
Gene Name
golgin A8 family member B
Gene Aliases
GOLGA5
UniGene
Hs.720151
Gene Chromosome Location
Chr.15: 34525276-34583651 on Build GRCh38
Transcripts
NM_001023567.4 NR_027410.1 XM_011521596.1 XM_011521597.1 XM_017022210.1 XM_017022211.1 XM_017022212.1 XM_017022213.1 XM_017022214.1 XM_017022215.1 XM_017022216.1 XM_017022217.1 XM_017022218.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
383151 c.1738G>T p.E580* Chr.15: 34527706-34527706 on Build GRCh38
1152819 c.1796C>T p.P599L Chr.15: 34527648-34527648 on Build GRCh38
3678147 c.1702G>A p.V568I Chr.15: 34527742-34527742 on Build GRCh38
3744565 c.1705C>G p.L569V Chr.15: 34527739-34527739 on Build GRCh38
3744567 c.1795C>G p.L599V Chr.15: 34527739-34527739 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565070904 Chr.15: 34527446-34527446 on Build GRCh38
rs752300855 Chr.15: 34527699-34527699 on Build GRCh38
rs534702670 Chr.15: 34527615-34527616 on Build GRCh38
rs963969281 Chr.15: 34527293-34527293 on Build GRCh38
rs567501959 Chr.15: 34527396-34527396 on Build GRCh38
rs950140716 Chr.15: 34527527-34527527 on Build GRCh38
rs577333662 Chr.15: 34527647-34527647 on Build GRCh38
rs777265006 Chr.15: 34527603-34527603 on Build GRCh38
rs548197020 Chr.15: 34527567-34527567 on Build GRCh38
rs955121234 Chr.15: 34527358-34527358 on Build GRCh38
rs903150613 Chr.15: 34527529-34527529 on Build GRCh38
rs2615360 Chr.15: 34527739-34527739 on Build GRCh38
rs775582230 Chr.15: 34527638-34527638 on Build GRCh38
rs190112464 Chr.15: 34527456-34527456 on Build GRCh38
rs973801631 Chr.15: 34527442-34527442 on Build GRCh38
rs920847097 Chr.15: 34527450-34527450 on Build GRCh38
rs574451752 Chr.15: 34527353-34527353 on Build GRCh38
rs538158452 Chr.15: 34527689-34527689 on Build GRCh38
rs753433882 Chr.15: 34527727-34527727 on Build GRCh38
rs941029056 Chr.15: 34527386-34527386 on Build GRCh38
rs773659907 Chr.15: 34527659-34527659 on Build GRCh38
rs541924444 Chr.15: 34527357-34527357 on Build GRCh38
rs745822399 Chr.15: 34527627-34527627 on Build GRCh38
rs546513830 Chr.15: 34527421-34527421 on Build GRCh38
rs1055961587 Chr.15: 34527492-34527492 on Build GRCh38
rs763585298 Chr.15: 34527684-34527684 on Build GRCh38
rs747064134 Chr.15: 34527642-34527642 on Build GRCh38
rs544860558 Chr.15: 34527304-34527304 on Build GRCh38
rs778700464 Chr.15: 34527618-34527618 on Build GRCh38
rs982955584 Chr.15: 34527370-34527370 on Build GRCh38
rs768973584 Chr.15: 34527644-34527644 on Build GRCh38
rs765525736 Chr.15: 34527657-34527657 on Build GRCh38
rs533862125 Chr.15: 34527605-34527605 on Build GRCh38
rs4144689 Chr.15: 34527318-34527318 on Build GRCh38
rs200453978 Chr.15: 34527451-34527451 on Build GRCh38
rs568882474 Chr.15: 34527543-34527543 on Build GRCh38
rs558906988 Chr.15: 34527629-34527629 on Build GRCh38
rs762307348 Chr.15: 34527649-34527649 on Build GRCh38
rs773937208 Chr.15: 34527486-34527491 on Build GRCh38
rs756995127 Chr.15: 34527608-34527608 on Build GRCh38
rs566477685 Chr.15: 34527568-34527568 on Build GRCh38
rs899746375 Chr.15: 34527576-34527576 on Build GRCh38
rs550862118 Chr.15: 34527458-34527458 on Build GRCh38
rs201197912 Chr.15: 34527726-34527726 on Build GRCh38
rs896001238 Chr.15: 34527504-34527504 on Build GRCh38
rs755562044 Chr.15: 34527718-34527718 on Build GRCh38
rs1000601849 Chr.15: 34527549-34527549 on Build GRCh38
rs755871019 Chr.15: 34527588-34527588 on Build GRCh38
rs756879438 Chr.15: 34527730-34527730 on Build GRCh38
rs1042196439 Chr.15: 34527528-34527528 on Build GRCh38
rs752214878 Chr.15: 34527582-34527582 on Build GRCh38
rs536297061 Chr.15: 34527544-34527544 on Build GRCh38
rs778597895 Chr.15: 34527737-34527737 on Build GRCh38
rs560260566 Chr.15: 34527371-34527371 on Build GRCh38
rs185685651 Chr.15: 34527326-34527326 on Build GRCh38

View Full Product Details