Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43775227-43775736 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELL3
Entrez Gene ID
80237
Gene Name
elongation factor for RNA polymerase II 3
Gene Aliases
-
UniGene
Hs.706346
Gene Chromosome Location
Chr.15: 43772600-43777304 on Build GRCh38
Transcripts
NM_025165.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
962063 c.589A>G p.N197D Chr.15: 43775362-43775362 on Build GRCh38
3361411 c.558G>A p.Q186Q Chr.15: 43775536-43775536 on Build GRCh38
3794189 c.570-1G>A p.? Chr.15: 43775382-43775382 on Build GRCh38
1608402 c.489A>T p.S163S Chr.15: 43775605-43775605 on Build GRCh38
1323187 c.583G>A p.V195I Chr.15: 43775368-43775368 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369172951 Chr.15: 43775480-43775480 on Build GRCh38
rs755067689 Chr.15: 43775410-43775410 on Build GRCh38
rs753580518 Chr.15: 43775292-43775300 on Build GRCh38
rs144962107 Chr.15: 43775323-43775323 on Build GRCh38
rs778743007 Chr.15: 43775658-43775658 on Build GRCh38
rs982638159 Chr.15: 43775545-43775545 on Build GRCh38
rs770900024 Chr.15: 43775419-43775419 on Build GRCh38
rs765451789 Chr.15: 43775385-43775385 on Build GRCh38
rs751833506 Chr.15: 43775631-43775631 on Build GRCh38
rs1055674381 Chr.15: 43775672-43775672 on Build GRCh38
rs963543797 Chr.15: 43775443-43775443 on Build GRCh38
rs778880317 Chr.15: 43775585-43775585 on Build GRCh38
rs933696109 Chr.15: 43775647-43775648 on Build GRCh38
rs368527844 Chr.15: 43775375-43775375 on Build GRCh38
rs376311400 Chr.15: 43775316-43775316 on Build GRCh38
rs764566562 Chr.15: 43775267-43775267 on Build GRCh38
rs372579644 Chr.15: 43775491-43775491 on Build GRCh38
rs758748971 Chr.15: 43775389-43775389 on Build GRCh38
rs758507980 Chr.15: 43775317-43775317 on Build GRCh38
rs774064368 Chr.15: 43775344-43775344 on Build GRCh38
rs918311101 Chr.15: 43775476-43775476 on Build GRCh38
rs779219082 Chr.15: 43775301-43775301 on Build GRCh38
rs146412353 Chr.15: 43775473-43775473 on Build GRCh38
rs749693139 Chr.15: 43775278-43775278 on Build GRCh38
rs777397774 Chr.15: 43775284-43775285 on Build GRCh38
rs754270202 Chr.15: 43775269-43775269 on Build GRCh38
rs771473047 Chr.15: 43775343-43775343 on Build GRCh38
rs909782821 Chr.15: 43775563-43775563 on Build GRCh38
rs780397585 Chr.15: 43775396-43775396 on Build GRCh38
rs764621382 Chr.15: 43775373-43775373 on Build GRCh38
rs767759116 Chr.15: 43775643-43775643 on Build GRCh38
rs779697747 Chr.15: 43775607-43775607 on Build GRCh38
rs750799344 Chr.15: 43775496-43775496 on Build GRCh38
rs754632661 Chr.15: 43775691-43775691 on Build GRCh38
rs752761104 Chr.15: 43775644-43775644 on Build GRCh38
rs909458385 Chr.15: 43775435-43775435 on Build GRCh38
rs369931248 Chr.15: 43775652-43775652 on Build GRCh38
rs766790843 Chr.15: 43775626-43775626 on Build GRCh38
rs531883967 Chr.15: 43775683-43775683 on Build GRCh38
rs768290291 Chr.15: 43775610-43775610 on Build GRCh38
rs755120703 Chr.15: 43775543-43775543 on Build GRCh38
rs763374753 Chr.15: 43775498-43775498 on Build GRCh38
rs745655619 Chr.15: 43775422-43775422 on Build GRCh38
rs766739456 Chr.15: 43775537-43775537 on Build GRCh38
rs377049838 Chr.15: 43775411-43775411 on Build GRCh38
rs768099954 Chr.15: 43775488-43775488 on Build GRCh38
rs201197535 Chr.15: 43775551-43775551 on Build GRCh38
rs761359622 Chr.15: 43775617-43775617 on Build GRCh38
rs775088692 Chr.15: 43775433-43775433 on Build GRCh38
rs745697910 Chr.15: 43775586-43775586 on Build GRCh38
rs747639403 Chr.15: 43775713-43775713 on Build GRCh38
rs757523719 Chr.15: 43775290-43775290 on Build GRCh38
rs752202244 Chr.15: 43775255-43775255 on Build GRCh38
rs750792902 Chr.15: 43775387-43775387 on Build GRCh38
rs375557328 Chr.15: 43775565-43775565 on Build GRCh38
rs778770300 Chr.15: 43775509-43775510 on Build GRCh38
rs372229990 Chr.15: 43775265-43775265 on Build GRCh38
rs776059471 Chr.15: 43775614-43775614 on Build GRCh38
rs140782164 Chr.15: 43775687-43775687 on Build GRCh38
rs775033845 Chr.15: 43775363-43775363 on Build GRCh38
rs756941564 Chr.15: 43775359-43775362 on Build GRCh38
rs752179804 Chr.15: 43775276-43775276 on Build GRCh38
rs755660687 Chr.15: 43775275-43775276 on Build GRCh38
rs750306906 Chr.15: 43775679-43775679 on Build GRCh38
rs747071839 Chr.15: 43775322-43775322 on Build GRCh38
rs746790926 Chr.15: 43775688-43775688 on Build GRCh38
rs773728364 Chr.15: 43775618-43775618 on Build GRCh38
rs369771946 Chr.15: 43775625-43775625 on Build GRCh38
rs780346680 Chr.15: 43775321-43775321 on Build GRCh38
rs771751830 Chr.15: 43775427-43775427 on Build GRCh38
rs757657139 Chr.15: 43775383-43775383 on Build GRCh38
rs771808909 Chr.15: 43775604-43775604 on Build GRCh38
rs373378172 Chr.15: 43775697-43775697 on Build GRCh38
rs758935342 Chr.15: 43775272-43775272 on Build GRCh38
rs749135819 Chr.15: 43775328-43775328 on Build GRCh38
rs113243709 Chr.15: 43775409-43775409 on Build GRCh38
rs151327600 Chr.15: 43775369-43775369 on Build GRCh38
rs779408639 Chr.15: 43775380-43775380 on Build GRCh38
rs757191401 Chr.15: 43775580-43775580 on Build GRCh38
rs745751156 Chr.15: 43775588-43775589 on Build GRCh38
rs375110222 Chr.15: 43775420-43775420 on Build GRCh38
rs765758493 Chr.15: 43775492-43775492 on Build GRCh38
rs992677007 Chr.15: 43775434-43775434 on Build GRCh38
rs373227136 Chr.15: 43775639-43775639 on Build GRCh38
rs767146967 Chr.15: 43775353-43775353 on Build GRCh38
rs552668420 Chr.15: 43775656-43775657 on Build GRCh38
rs972217989 Chr.15: 43775459-43775459 on Build GRCh38
rs751782159 Chr.15: 43775538-43775538 on Build GRCh38
rs749120425 Chr.15: 43775412-43775412 on Build GRCh38
rs759180965 Chr.15: 43775351-43775351 on Build GRCh38
rs746600602 Chr.15: 43775608-43775608 on Build GRCh38

View Full Product Details