Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 44413629-44414166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASC4
Entrez Gene ID
113201
Gene Name
cancer susceptibility candidate 4
Gene Aliases
H63
UniGene
Hs.512867
Gene Chromosome Location
Chr.15: 44288616-44415761 on Build GRCh38
Transcripts
NM_138423.3 NM_177974.2

SNP Detail

SNP ID Chromosome Location
rs960609239 Chr.15: 44413855-44413855 on Build GRCh38
rs191290402 Chr.15: 44413892-44413892 on Build GRCh38
rs560254993 Chr.15: 44413899-44413899 on Build GRCh38
rs1002119335 Chr.15: 44413832-44413833 on Build GRCh38
rs983175238 Chr.15: 44413956-44413956 on Build GRCh38
rs80320444 Chr.15: 44413828-44413828 on Build GRCh38
rs28587354 Chr.15: 44414000-44414000 on Build GRCh38
rs561295684 Chr.15: 44413767-44413767 on Build GRCh38
rs772829218 Chr.15: 44413945-44413945 on Build GRCh38
rs1010299437 Chr.15: 44413711-44413711 on Build GRCh38
rs1013246000 Chr.15: 44413868-44413868 on Build GRCh38
rs768310622 Chr.15: 44413685-44413685 on Build GRCh38
rs975020961 Chr.15: 44413996-44413996 on Build GRCh38
rs1062726 Chr.15: 44413782-44413782 on Build GRCh38
rs11541697 Chr.15: 44414104-44414104 on Build GRCh38
rs904669022 Chr.15: 44413750-44413750 on Build GRCh38
rs1004134641 Chr.15: 44413810-44413810 on Build GRCh38
rs1025224240 Chr.15: 44413847-44413847 on Build GRCh38
rs921862452 Chr.15: 44414029-44414029 on Build GRCh38
rs749727730 Chr.15: 44414051-44414051 on Build GRCh38
rs540130412 Chr.15: 44413663-44413663 on Build GRCh38
rs771597633 Chr.15: 44414109-44414109 on Build GRCh38
rs774812273 Chr.15: 44414121-44414121 on Build GRCh38
rs552002751 Chr.15: 44414091-44414091 on Build GRCh38
rs773930911 Chr.15: 44414127-44414127 on Build GRCh38
rs1051696015 Chr.15: 44414066-44414066 on Build GRCh38
rs908975997 Chr.15: 44413962-44413962 on Build GRCh38
rs933333346 Chr.15: 44413680-44413680 on Build GRCh38
rs558128522 Chr.15: 44413684-44413684 on Build GRCh38
rs878960875 Chr.15: 44413811-44413811 on Build GRCh38
rs963203518 Chr.15: 44413973-44413973 on Build GRCh38
rs113021740 Chr.15: 44413837-44413837 on Build GRCh38
rs950430904 Chr.15: 44413901-44413901 on Build GRCh38
rs181351288 Chr.15: 44413984-44413984 on Build GRCh38
rs568804578 Chr.15: 44414099-44414099 on Build GRCh38
rs891867020 Chr.15: 44413703-44413703 on Build GRCh38
rs1043139640 Chr.15: 44413747-44413747 on Build GRCh38
rs141209769 Chr.15: 44413908-44413908 on Build GRCh38

View Full Product Details