Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 44667015-44667512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PATL2
Entrez Gene ID
197135
Gene Name
PAT1 homolog 2
Gene Aliases
Pat1a, hPat1a
UniGene
Hs.11594
Gene Chromosome Location
Chr.15: 44665732-44711389 on Build GRCh38
Transcripts
NM_001145112.1 XM_011521336.2 XM_011521337.2 XM_011521338.2 XM_011521339.2 XM_011521340.2 XM_011521341.1 XM_011521342.2 XM_011521343.2 XM_011521344.2 XM_011521345.2 XM_011521346.2 XM_011521347.1 XM_011521348.2 XM_017022000.1 XM_017022001.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3981522 c.1433T>C p.L478P Chr.15: 44667136-44667136 on Build GRCh38
6463911 c.1383C>G p.L461L Chr.15: 44667186-44667186 on Build GRCh38
6858776 c.1385A>G p.Y462C Chr.15: 44667184-44667184 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1029338538 Chr.15: 44667057-44667057 on Build GRCh38
rs866327063 Chr.15: 44667421-44667421 on Build GRCh38
rs117655669 Chr.15: 44667275-44667275 on Build GRCh38
rs766906968 Chr.15: 44667461-44667461 on Build GRCh38
rs201569149 Chr.15: 44667389-44667389 on Build GRCh38
rs897688808 Chr.15: 44667383-44667383 on Build GRCh38
rs772022619 Chr.15: 44667041-44667041 on Build GRCh38
rs759138042 Chr.15: 44667097-44667098 on Build GRCh38
rs775346086 Chr.15: 44667176-44667176 on Build GRCh38
rs907532823 Chr.15: 44667423-44667424 on Build GRCh38
rs888436170 Chr.15: 44667315-44667315 on Build GRCh38
rs898992304 Chr.15: 44667060-44667060 on Build GRCh38
rs748121988 Chr.15: 44667238-44667238 on Build GRCh38
rs576033770 Chr.15: 44667322-44667322 on Build GRCh38
rs980436962 Chr.15: 44667187-44667187 on Build GRCh38
rs991933825 Chr.15: 44667144-44667144 on Build GRCh38
rs770926694 Chr.15: 44667249-44667249 on Build GRCh38
rs1028874875 Chr.15: 44667096-44667096 on Build GRCh38
rs376819537 Chr.15: 44667262-44667262 on Build GRCh38
rs140029574 Chr.15: 44667214-44667215 on Build GRCh38
rs981803499 Chr.15: 44667385-44667385 on Build GRCh38
rs1014800340 Chr.15: 44667084-44667084 on Build GRCh38
rs148158070 Chr.15: 44667136-44667136 on Build GRCh38
rs536740965 Chr.15: 44667382-44667382 on Build GRCh38
rs750849097 Chr.15: 44667109-44667109 on Build GRCh38
rs375242446 Chr.15: 44667258-44667258 on Build GRCh38
rs547168925 Chr.15: 44667064-44667064 on Build GRCh38
rs565511577 Chr.15: 44667342-44667343 on Build GRCh38
rs954599033 Chr.15: 44667124-44667124 on Build GRCh38
rs567533050 Chr.15: 44667349-44667349 on Build GRCh38
rs747314039 Chr.15: 44667218-44667218 on Build GRCh38
rs1048356140 Chr.15: 44667284-44667284 on Build GRCh38
rs374192103 Chr.15: 44667077-44667077 on Build GRCh38
rs1032042597 Chr.15: 44667475-44667475 on Build GRCh38
rs996845007 Chr.15: 44667044-44667044 on Build GRCh38
rs1002287674 Chr.15: 44667474-44667474 on Build GRCh38
rs557583117 Chr.15: 44667078-44667078 on Build GRCh38
rs1013393805 Chr.15: 44667424-44667424 on Build GRCh38
rs925527614 Chr.15: 44667197-44667197 on Build GRCh38
rs3215056 Chr.15: 44667346-44667347 on Build GRCh38
rs1006475424 Chr.15: 44667068-44667068 on Build GRCh38
rs760457430 Chr.15: 44667195-44667195 on Build GRCh38
rs768757841 Chr.15: 44667219-44667219 on Build GRCh38
rs762432878 Chr.15: 44667386-44667386 on Build GRCh38
rs899900091 Chr.15: 44667272-44667272 on Build GRCh38
rs1024719184 Chr.15: 44667150-44667150 on Build GRCh38
rs917525716 Chr.15: 44667374-44667374 on Build GRCh38
rs146665035 Chr.15: 44667250-44667250 on Build GRCh38
rs755337370 Chr.15: 44667464-44667464 on Build GRCh38
rs914109493 Chr.15: 44667240-44667240 on Build GRCh38
rs141056347 Chr.15: 44667233-44667233 on Build GRCh38

View Full Product Details