Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 60352295-60352805 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA2
Entrez Gene ID
302
Gene Name
annexin A2
Gene Aliases
ANX2, ANX2L4, CAL1H, HEL-S-270, LIP2, LPC2, LPC2D, P36, PAP-IV
UniGene
Hs.511605
Gene Chromosome Location
Chr.15: 60347151-60398025 on Build GRCh38
Transcripts
NM_001002857.1 NM_001002858.2 NM_001136015.2 NM_004039.2 XM_011521477.2 XM_017022090.1 XM_017022091.1 XM_017022092.1 XM_017022093.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6368482 c.652G>T p.D218Y Chr.15: 60352467-60352467 on Build GRCh38
6860761 c.650A>G p.Y217C Chr.15: 60352469-60352469 on Build GRCh38
1373839 c.718G>A p.V240M Chr.15: 60352401-60352401 on Build GRCh38
5054389 c.673G>A p.G225R Chr.15: 60352446-60352446 on Build GRCh38
4578386 c.654C>T p.D218D Chr.15: 60352465-60352465 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs554967872 Chr.15: 60352563-60352563 on Build GRCh38
rs41306349 Chr.15: 60352579-60352579 on Build GRCh38
rs1031744330 Chr.15: 60352719-60352719 on Build GRCh38
rs189333401 Chr.15: 60352441-60352441 on Build GRCh38
rs113695974 Chr.15: 60352478-60352478 on Build GRCh38
rs1013951179 Chr.15: 60352706-60352706 on Build GRCh38
rs922546761 Chr.15: 60352646-60352646 on Build GRCh38
rs887174872 Chr.15: 60352368-60352368 on Build GRCh38
rs763372364 Chr.15: 60352346-60352346 on Build GRCh38
rs149164729 Chr.15: 60352700-60352700 on Build GRCh38
rs993980959 Chr.15: 60352488-60352488 on Build GRCh38
rs374077954 Chr.15: 60352401-60352401 on Build GRCh38
rs768952097 Chr.15: 60352468-60352469 on Build GRCh38
rs116367881 Chr.15: 60352481-60352481 on Build GRCh38
rs955038442 Chr.15: 60352469-60352469 on Build GRCh38
rs770772924 Chr.15: 60352407-60352407 on Build GRCh38
rs745354147 Chr.15: 60352410-60352410 on Build GRCh38
rs759572444 Chr.15: 60352456-60352456 on Build GRCh38
rs114189022 Chr.15: 60352333-60352333 on Build GRCh38
rs755098991 Chr.15: 60352390-60352390 on Build GRCh38
rs753565260 Chr.15: 60352489-60352489 on Build GRCh38
rs541311582 Chr.15: 60352724-60352724 on Build GRCh38
rs1023962563 Chr.15: 60352726-60352726 on Build GRCh38
rs1005347464 Chr.15: 60352692-60352692 on Build GRCh38
rs267604276 Chr.15: 60352434-60352434 on Build GRCh38
rs766974356 Chr.15: 60352352-60352352 on Build GRCh38
rs763826595 Chr.15: 60352701-60352701 on Build GRCh38
rs762206405 Chr.15: 60352527-60352528 on Build GRCh38
rs141453912 Chr.15: 60352551-60352551 on Build GRCh38
rs145194373 Chr.15: 60352640-60352640 on Build GRCh38
rs1044293966 Chr.15: 60352567-60352567 on Build GRCh38
rs144491837 Chr.15: 60352525-60352525 on Build GRCh38
rs891352983 Chr.15: 60352699-60352699 on Build GRCh38
rs191173681 Chr.15: 60352578-60352578 on Build GRCh38
rs766981748 Chr.15: 60352522-60352522 on Build GRCh38
rs752148384 Chr.15: 60352451-60352451 on Build GRCh38
rs185878054 Chr.15: 60352519-60352519 on Build GRCh38
rs182754172 Chr.15: 60352750-60352750 on Build GRCh38
rs764998469 Chr.15: 60352718-60352718 on Build GRCh38
rs746583193 Chr.15: 60352516-60352516 on Build GRCh38
rs1004340561 Chr.15: 60352374-60352374 on Build GRCh38
rs569776717 Chr.15: 60352480-60352480 on Build GRCh38
rs924095596 Chr.15: 60352600-60352600 on Build GRCh38
rs938637934 Chr.15: 60352582-60352582 on Build GRCh38
rs145003552 Chr.15: 60352672-60352672 on Build GRCh38
rs989974242 Chr.15: 60352620-60352620 on Build GRCh38
rs191212436 Chr.15: 60352680-60352680 on Build GRCh38
rs572461962 Chr.15: 60352395-60352395 on Build GRCh38
rs186759774 Chr.15: 60352623-60352623 on Build GRCh38
rs745819689 Chr.15: 60352504-60352504 on Build GRCh38
rs955711943 Chr.15: 60352744-60352744 on Build GRCh38
rs777026082 Chr.15: 60352528-60352528 on Build GRCh38
rs757129245 Chr.15: 60352492-60352492 on Build GRCh38
rs997030715 Chr.15: 60352665-60352665 on Build GRCh38
rs887141973 Chr.15: 60352616-60352616 on Build GRCh38
rs999775696 Chr.15: 60352757-60352757 on Build GRCh38
rs182967406 Chr.15: 60352583-60352583 on Build GRCh38
rs566273872 Chr.15: 60352421-60352421 on Build GRCh38
rs970948908 Chr.15: 60352587-60352587 on Build GRCh38
rs749184513 Chr.15: 60352406-60352406 on Build GRCh38
rs958187179 Chr.15: 60352611-60352611 on Build GRCh38
rs138364945 Chr.15: 60352465-60352465 on Build GRCh38
rs377502196 Chr.15: 60352524-60352524 on Build GRCh38
rs775182884 Chr.15: 60352536-60352536 on Build GRCh38
rs1013897886 Chr.15: 60352543-60352544 on Build GRCh38
rs111353136 Chr.15: 60352428-60352428 on Build GRCh38
rs777444250 Chr.15: 60352405-60352405 on Build GRCh38
rs371196182 Chr.15: 60352411-60352411 on Build GRCh38
rs755855483 Chr.15: 60352403-60352403 on Build GRCh38
rs778970589 Chr.15: 60352409-60352409 on Build GRCh38
rs765717031 Chr.15: 60352332-60352332 on Build GRCh38
rs766773335 Chr.15: 60352444-60352444 on Build GRCh38
rs989816625 Chr.15: 60352745-60352745 on Build GRCh38
rs940053775 Chr.15: 60352622-60352622 on Build GRCh38
rs969812119 Chr.15: 60352752-60352752 on Build GRCh38
rs929882600 Chr.15: 60352654-60352654 on Build GRCh38
rs761819974 Chr.15: 60352393-60352393 on Build GRCh38
rs878944999 Chr.15: 60352335-60352335 on Build GRCh38
rs759916614 Chr.15: 60352595-60352595 on Build GRCh38
rs779712643 Chr.15: 60352514-60352514 on Build GRCh38
rs529527729 Chr.15: 60352402-60352402 on Build GRCh38
rs771646992 Chr.15: 60352512-60352512 on Build GRCh38
rs767606956 Chr.15: 60352464-60352464 on Build GRCh38
rs775231291 Chr.15: 60352412-60352412 on Build GRCh38
rs751577460 Chr.15: 60352362-60352362 on Build GRCh38
rs548055644 Chr.15: 60352420-60352420 on Build GRCh38
rs778671622 Chr.15: 60352503-60352503 on Build GRCh38
rs147842368 Chr.15: 60352535-60352535 on Build GRCh38

View Full Product Details