Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 63536898-63537409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP3
Entrez Gene ID
9960
Gene Name
ubiquitin specific peptidase 3
Gene Aliases
SIH003, UBP
UniGene
Hs.458499
Gene Chromosome Location
Chr.15: 63504511-63594640 on Build GRCh38
Transcripts
NM_001256702.1 NM_006537.3 NR_046341.1 NR_046342.1 XM_017022763.1 XM_017022764.1 XM_017022765.1 XM_017022766.1 XM_017022767.1 XM_017022768.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1270073 c.175A>C p.K59Q Chr.15: 63537047-63537047 on Build GRCh38
1608561 c.192A>G p.A64A Chr.15: 63537064-63537064 on Build GRCh38
4765259 c.171_172insA p.H60fs*3 Chr.15: 63537043-63537044 on Build GRCh38
6077549 c.247A>C p.T83P Chr.15: 63537119-63537119 on Build GRCh38
4610985 c.171delA p.K59fs*10 Chr.15: 63537043-63537043 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760413539 Chr.15: 63537207-63537207 on Build GRCh38
rs376834221 Chr.15: 63536978-63536978 on Build GRCh38
rs567231691 Chr.15: 63537115-63537115 on Build GRCh38
rs1037708032 Chr.15: 63537062-63537062 on Build GRCh38
rs763440842 Chr.15: 63537179-63537180 on Build GRCh38
rs904842992 Chr.15: 63537306-63537306 on Build GRCh38
rs908488537 Chr.15: 63536948-63536948 on Build GRCh38
rs1009097282 Chr.15: 63537218-63537218 on Build GRCh38
rs371302474 Chr.15: 63536925-63536928 on Build GRCh38
rs745788699 Chr.15: 63537281-63537296 on Build GRCh38
rs549098787 Chr.15: 63537067-63537067 on Build GRCh38
rs748980494 Chr.15: 63537020-63537020 on Build GRCh38
rs916738749 Chr.15: 63537372-63537372 on Build GRCh38
rs939409205 Chr.15: 63537353-63537353 on Build GRCh38
rs201119432 Chr.15: 63537150-63537150 on Build GRCh38
rs754156214 Chr.15: 63537014-63537014 on Build GRCh38
rs1034804010 Chr.15: 63537336-63537336 on Build GRCh38
rs769522438 Chr.15: 63537379-63537379 on Build GRCh38
rs1011076733 Chr.15: 63537244-63537244 on Build GRCh38
rs758938252 Chr.15: 63537044-63537044 on Build GRCh38
rs890519503 Chr.15: 63537192-63537192 on Build GRCh38
rs879460905 Chr.15: 63536973-63536973 on Build GRCh38
rs780307574 Chr.15: 63537049-63537049 on Build GRCh38
rs773863569 Chr.15: 63537151-63537151 on Build GRCh38
rs11853890 Chr.15: 63537339-63537339 on Build GRCh38
rs551049622 Chr.15: 63537243-63537243 on Build GRCh38
rs368799536 Chr.15: 63537164-63537164 on Build GRCh38
rs569444348 Chr.15: 63537311-63537311 on Build GRCh38
rs757704333 Chr.15: 63537037-63537037 on Build GRCh38
rs779130724 Chr.15: 63537040-63537040 on Build GRCh38
rs769680894 Chr.15: 63537107-63537107 on Build GRCh38
rs982675399 Chr.15: 63537346-63537346 on Build GRCh38
rs1021927229 Chr.15: 63537284-63537284 on Build GRCh38
rs892761004 Chr.15: 63537230-63537230 on Build GRCh38
rs74361579 Chr.15: 63537360-63537360 on Build GRCh38
rs748306078 Chr.15: 63537105-63537105 on Build GRCh38
rs771514191 Chr.15: 63537187-63537215 on Build GRCh38
rs781546757 Chr.15: 63537104-63537104 on Build GRCh38
rs768767849 Chr.15: 63537090-63537090 on Build GRCh38
rs746421914 Chr.15: 63537043-63537043 on Build GRCh38
rs1026054166 Chr.15: 63537261-63537261 on Build GRCh38
rs962627796 Chr.15: 63537358-63537358 on Build GRCh38
rs1008732729 Chr.15: 63537287-63537287 on Build GRCh38
rs941272635 Chr.15: 63536990-63536990 on Build GRCh38
rs200387796 Chr.15: 63537154-63537154 on Build GRCh38
rs371961517 Chr.15: 63536994-63536994 on Build GRCh38
rs574140240 Chr.15: 63537361-63537361 on Build GRCh38
rs1015803667 Chr.15: 63537350-63537350 on Build GRCh38
rs760219609 Chr.15: 63537180-63537180 on Build GRCh38
rs146458254 Chr.15: 63537326-63537326 on Build GRCh38
rs995036827 Chr.15: 63537214-63537214 on Build GRCh38
rs1053027438 Chr.15: 63537219-63537219 on Build GRCh38
rs532810476 Chr.15: 63537172-63537172 on Build GRCh38
rs765733462 Chr.15: 63537189-63537189 on Build GRCh38
rs759017095 Chr.15: 63537158-63537158 on Build GRCh38
rs762908480 Chr.15: 63537138-63537138 on Build GRCh38
rs1001806081 Chr.15: 63537314-63537314 on Build GRCh38

View Full Product Details