Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 71114988-71115486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CT62
Entrez Gene ID
196993
Gene Name
cancer/testis antigen 62
Gene Aliases
-
UniGene
Hs.646533
Gene Chromosome Location
Chr.15: 71110244-71115551 on Build GRCh38
Transcripts
NM_001102658.1 XM_006720429.3

SNP Detail

SNP ID Chromosome Location
rs527252134 Chr.15: 71115459-71115459 on Build GRCh38
rs867175007 Chr.15: 71115156-71115156 on Build GRCh38
rs975064789 Chr.15: 71115342-71115342 on Build GRCh38
rs919562439 Chr.15: 71115395-71115395 on Build GRCh38
rs929038684 Chr.15: 71115357-71115357 on Build GRCh38
rs145330951 Chr.15: 71115064-71115065 on Build GRCh38
rs1020119119 Chr.15: 71115215-71115215 on Build GRCh38
rs999351889 Chr.15: 71115160-71115160 on Build GRCh38
rs923188636 Chr.15: 71115420-71115420 on Build GRCh38
rs909027876 Chr.15: 71115394-71115394 on Build GRCh38
rs777254203 Chr.15: 71115199-71115199 on Build GRCh38
rs967654926 Chr.15: 71115217-71115217 on Build GRCh38
rs531539389 Chr.15: 71115418-71115418 on Build GRCh38
rs574449919 Chr.15: 71115017-71115017 on Build GRCh38
rs903227462 Chr.15: 71115148-71115148 on Build GRCh38
rs887621486 Chr.15: 71115084-71115084 on Build GRCh38
rs907280916 Chr.15: 71115083-71115083 on Build GRCh38
rs893078346 Chr.15: 71115145-71115145 on Build GRCh38
rs540246842 Chr.15: 71115035-71115035 on Build GRCh38
rs559985716 Chr.15: 71115107-71115107 on Build GRCh38
rs1033151143 Chr.15: 71115198-71115198 on Build GRCh38
rs907999972 Chr.15: 71115034-71115034 on Build GRCh38
rs867624695 Chr.15: 71115239-71115239 on Build GRCh38
rs548268322 Chr.15: 71115425-71115425 on Build GRCh38
rs1019350380 Chr.15: 71115305-71115305 on Build GRCh38
rs998925622 Chr.15: 71115443-71115443 on Build GRCh38
rs903195369 Chr.15: 71115437-71115437 on Build GRCh38
rs1054523981 Chr.15: 71115447-71115447 on Build GRCh38
rs781324864 Chr.15: 71115280-71115280 on Build GRCh38
rs1011880157 Chr.15: 71115166-71115166 on Build GRCh38
rs1013079682 Chr.15: 71115457-71115457 on Build GRCh38
rs971676191 Chr.15: 71115339-71115339 on Build GRCh38
rs1030115529 Chr.15: 71115143-71115143 on Build GRCh38
rs1038946473 Chr.15: 71115113-71115113 on Build GRCh38
rs901791257 Chr.15: 71115121-71115121 on Build GRCh38
rs943261406 Chr.15: 71115095-71115095 on Build GRCh38
rs561782843 Chr.15: 71115450-71115450 on Build GRCh38
rs984961592 Chr.15: 71115393-71115393 on Build GRCh38
rs546110210 Chr.15: 71115284-71115284 on Build GRCh38
rs915555407 Chr.15: 71115026-71115026 on Build GRCh38
rs577039337 Chr.15: 71115141-71115141 on Build GRCh38
rs562541537 Chr.15: 71115296-71115296 on Build GRCh38
rs919007939 Chr.15: 71115037-71115037 on Build GRCh38
rs940791550 Chr.15: 71115039-71115039 on Build GRCh38
rs748534245 Chr.15: 71115313-71115313 on Build GRCh38
rs1046276327 Chr.15: 71115060-71115060 on Build GRCh38
rs572924263 Chr.15: 71115241-71115241 on Build GRCh38
rs918922153 Chr.15: 71115354-71115354 on Build GRCh38
rs981130780 Chr.15: 71115349-71115349 on Build GRCh38
rs957541994 Chr.15: 71115214-71115214 on Build GRCh38
rs1051070952 Chr.15: 71115428-71115428 on Build GRCh38

View Full Product Details