Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90241245-90241751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIB1
Entrez Gene ID
10519
Gene Name
calcium and integrin binding 1
Gene Aliases
CIB, CIBP, KIP1, PRKDCIP, SIP2-28
UniGene
Hs.715556
Gene Chromosome Location
Chr.15: 90230245-90265759 on Build GRCh38
Transcripts
NR_102428.1 NM_001013657.2 NM_001322811.1

Gene Symbol
GDPGP1
Entrez Gene ID
390637
Gene Name
GDP-D-glucose phosphorylase 1
Gene Aliases
C15orf58, VTC2
UniGene
Hs.304673
Gene Chromosome Location
Chr.15: 90234255-90242080 on Build GRCh38
Transcripts
NR_102428.1 NM_001013657.2 NM_001322811.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4805415 c.556C>G p.L186V Chr.15: 90241464-90241464 on Build GRCh38
2151223 c.687C>T p.P229P Chr.15: 90241595-90241595 on Build GRCh38
234465 c.360C>T p.D120D Chr.15: 90241268-90241268 on Build GRCh38
5054593 c.390G>A p.R130R Chr.15: 90241298-90241298 on Build GRCh38
2139817 c.807G>C p.L269F Chr.15: 90241715-90241715 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs965490300 Chr.15: 90241498-90241498 on Build GRCh38
rs370091855 Chr.15: 90241546-90241546 on Build GRCh38
rs767518998 Chr.15: 90241592-90241592 on Build GRCh38
rs779239841 Chr.15: 90241617-90241617 on Build GRCh38
rs562900337 Chr.15: 90241351-90241351 on Build GRCh38
rs751054576 Chr.15: 90241327-90241327 on Build GRCh38
rs755991077 Chr.15: 90241446-90241446 on Build GRCh38
rs779887210 Chr.15: 90241511-90241511 on Build GRCh38
rs761821660 Chr.15: 90241590-90241590 on Build GRCh38
rs767981256 Chr.15: 90241343-90241343 on Build GRCh38
rs748996399 Chr.15: 90241388-90241388 on Build GRCh38
rs148271561 Chr.15: 90241721-90241721 on Build GRCh38
rs754157909 Chr.15: 90241355-90241355 on Build GRCh38
rs938049729 Chr.15: 90241403-90241403 on Build GRCh38
rs147678028 Chr.15: 90241611-90241611 on Build GRCh38
rs755241008 Chr.15: 90241359-90241359 on Build GRCh38
rs746618743 Chr.15: 90241614-90241614 on Build GRCh38
rs753839752 Chr.15: 90241593-90241593 on Build GRCh38
rs200655681 Chr.15: 90241547-90241547 on Build GRCh38
rs113159812 Chr.15: 90241324-90241324 on Build GRCh38
rs182830938 Chr.15: 90241287-90241287 on Build GRCh38
rs781206862 Chr.15: 90241373-90241373 on Build GRCh38
rs768273968 Chr.15: 90241389-90241389 on Build GRCh38
rs371699554 Chr.15: 90241431-90241431 on Build GRCh38
rs10152994 Chr.15: 90241698-90241698 on Build GRCh38
rs774441022 Chr.15: 90241656-90241656 on Build GRCh38
rs770347958 Chr.15: 90241617-90241617 on Build GRCh38
rs754133795 Chr.15: 90241420-90241420 on Build GRCh38
rs1035537924 Chr.15: 90241607-90241607 on Build GRCh38
rs746438201 Chr.15: 90241623-90241623 on Build GRCh38
rs866474098 Chr.15: 90241651-90241651 on Build GRCh38
rs754897853 Chr.15: 90241596-90241596 on Build GRCh38
rs757640966 Chr.15: 90241568-90241582 on Build GRCh38
rs149982920 Chr.15: 90241557-90241557 on Build GRCh38
rs553590266 Chr.15: 90241676-90241676 on Build GRCh38
rs142411677 Chr.15: 90241456-90241456 on Build GRCh38
rs537420206 Chr.15: 90241507-90241507 on Build GRCh38
rs749051346 Chr.15: 90241451-90241451 on Build GRCh38
rs756959758 Chr.15: 90241724-90241724 on Build GRCh38
rs147221532 Chr.15: 90241326-90241326 on Build GRCh38
rs760006537 Chr.15: 90241486-90241486 on Build GRCh38
rs772585229 Chr.15: 90241469-90241469 on Build GRCh38
rs763282865 Chr.15: 90241577-90241577 on Build GRCh38
rs779185782 Chr.15: 90241361-90241361 on Build GRCh38
rs753945188 Chr.15: 90241498-90241499 on Build GRCh38
rs777385902 Chr.15: 90241520-90241520 on Build GRCh38
rs768400375 Chr.15: 90241452-90241452 on Build GRCh38
rs763887610 Chr.15: 90241713-90241713 on Build GRCh38
rs766767553 Chr.15: 90241478-90241478 on Build GRCh38
rs930593703 Chr.15: 90241619-90241619 on Build GRCh38
rs770974219 Chr.15: 90241317-90241317 on Build GRCh38
rs758875240 Chr.15: 90241686-90241686 on Build GRCh38
rs747857376 Chr.15: 90241517-90241517 on Build GRCh38
rs1044774542 Chr.15: 90241602-90241602 on Build GRCh38
rs775141206 Chr.15: 90241383-90241383 on Build GRCh38
rs923755131 Chr.15: 90241503-90241503 on Build GRCh38
rs146611941 Chr.15: 90241626-90241626 on Build GRCh38
rs200135769 Chr.15: 90241513-90241513 on Build GRCh38
rs199729244 Chr.15: 90241613-90241613 on Build GRCh38
rs201540680 Chr.15: 90241492-90241492 on Build GRCh38
rs775982050 Chr.15: 90241569-90241569 on Build GRCh38
rs761334602 Chr.15: 90241406-90241406 on Build GRCh38
rs766827240 Chr.15: 90241415-90241415 on Build GRCh38
rs780900707 Chr.15: 90241727-90241727 on Build GRCh38
rs140114924 Chr.15: 90241453-90241453 on Build GRCh38
rs200657228 Chr.15: 90241382-90241382 on Build GRCh38
rs764189936 Chr.15: 90241588-90241588 on Build GRCh38
rs543564025 Chr.15: 90241432-90241432 on Build GRCh38
rs762355066 Chr.15: 90241333-90241333 on Build GRCh38
rs199500826 Chr.15: 90241512-90241512 on Build GRCh38
rs773251991 Chr.15: 90241686-90241686 on Build GRCh38
rs750981767 Chr.15: 90241683-90241683 on Build GRCh38
rs751764803 Chr.15: 90241589-90241589 on Build GRCh38
rs745731193 Chr.15: 90241374-90241374 on Build GRCh38
rs762090432 Chr.15: 90241666-90241666 on Build GRCh38
rs746535811 Chr.15: 90241539-90241539 on Build GRCh38
rs761296044 Chr.15: 90241472-90241472 on Build GRCh38
rs139186837 Chr.15: 90241407-90241407 on Build GRCh38
rs763245804 Chr.15: 90241646-90241646 on Build GRCh38
rs575977797 Chr.15: 90241567-90241567 on Build GRCh38
rs778730567 Chr.15: 90241597-90241597 on Build GRCh38
rs573404545 Chr.15: 90241297-90241297 on Build GRCh38
rs368724206 Chr.15: 90241296-90241296 on Build GRCh38
rs201738314 Chr.15: 90241606-90241606 on Build GRCh38
rs994397262 Chr.15: 90241429-90241429 on Build GRCh38
rs140605827 Chr.15: 90241502-90241502 on Build GRCh38
rs769016716 Chr.15: 90241647-90241647 on Build GRCh38
rs781594391 Chr.15: 90241270-90241270 on Build GRCh38
rs377175098 Chr.15: 90241518-90241518 on Build GRCh38
rs112078428 Chr.15: 90241620-90241620 on Build GRCh38
rs376585765 Chr.15: 90241683-90241683 on Build GRCh38
rs75296384 Chr.15: 90241630-90241630 on Build GRCh38
rs112697825 Chr.15: 90241349-90241349 on Build GRCh38
rs200606347 Chr.15: 90241302-90241302 on Build GRCh38
rs777240949 Chr.15: 90241483-90241483 on Build GRCh38
rs766587375 Chr.15: 90241353-90241353 on Build GRCh38
rs770404210 Chr.15: 90241625-90241625 on Build GRCh38
rs565450447 Chr.15: 90241322-90241322 on Build GRCh38
rs754200604 Chr.15: 90241459-90241459 on Build GRCh38
rs760049795 Chr.15: 90241556-90241556 on Build GRCh38
rs765186934 Chr.15: 90241696-90241696 on Build GRCh38
rs775218294 Chr.15: 90241325-90241325 on Build GRCh38
rs144466387 Chr.15: 90241400-90241400 on Build GRCh38
rs780347406 Chr.15: 90241292-90241292 on Build GRCh38
rs758517091 Chr.15: 90241368-90241368 on Build GRCh38
rs557009666 Chr.15: 90241447-90241447 on Build GRCh38
rs764395845 Chr.15: 90241335-90241335 on Build GRCh38
rs996266683 Chr.15: 90241471-90241471 on Build GRCh38
rs748212146 Chr.15: 90241362-90241362 on Build GRCh38
rs373465372 Chr.15: 90241457-90241457 on Build GRCh38
rs151315312 Chr.15: 90241496-90241496 on Build GRCh38
rs947450292 Chr.15: 90241578-90241578 on Build GRCh38
rs144061266 Chr.15: 90241413-90241413 on Build GRCh38
rs1048672722 Chr.15: 90241649-90241649 on Build GRCh38
rs750778443 Chr.15: 90241274-90241274 on Build GRCh38
rs200990512 Chr.15: 90241504-90241504 on Build GRCh38
rs752969170 Chr.15: 90241501-90241501 on Build GRCh38
rs756394524 Chr.15: 90241291-90241291 on Build GRCh38
rs143773020 Chr.15: 90241318-90241318 on Build GRCh38
rs142135462 Chr.15: 90241687-90241687 on Build GRCh38
rs758137130 Chr.15: 90241707-90241707 on Build GRCh38
rs751568296 Chr.15: 90241505-90241505 on Build GRCh38
rs755294533 Chr.15: 90241427-90241427 on Build GRCh38
rs536275149 Chr.15: 90241313-90241313 on Build GRCh38
rs138559985 Chr.15: 90241439-90241439 on Build GRCh38

View Full Product Details