Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90931601-90932110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDDC3
Entrez Gene ID
374659
Gene Name
HD domain containing 3
Gene Aliases
(ppGpp)ase, MESH1
UniGene
Hs.349979
Gene Chromosome Location
Chr.15: 90930918-90932569 on Build GRCh38
Transcripts
NM_001286451.1 NM_198527.3 NR_104447.1 NM_001039675.1 XM_011521779.2

Gene Symbol
UNC45A
Entrez Gene ID
55898
Gene Name
unc-45 myosin chaperone A
Gene Aliases
GC-UNC45, GCUNC-45, GCUNC45, IRO039700, SMAP-1, SMAP1, UNC-45A
UniGene
Hs.389461
Gene Chromosome Location
Chr.15: 90929980-90954093 on Build GRCh38
Transcripts
NM_001286451.1 NM_198527.3 NR_104447.1 NM_001039675.1 XM_011521779.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3387152 c.274G>A p.D92N Chr.15: 90931839-90931839 on Build GRCh38
161474 c.363A>C p.A121A Chr.15: 90931750-90931750 on Build GRCh38
4057965 c.316G>A p.E106K Chr.15: 90931797-90931797 on Build GRCh38
3741737 c.301A>G p.K101E Chr.15: 90931812-90931812 on Build GRCh38
3817076 c.217G>A p.E73K Chr.15: 90931896-90931896 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs981666865 Chr.15: 90931793-90931793 on Build GRCh38
rs371163282 Chr.15: 90931990-90931990 on Build GRCh38
rs778839266 Chr.15: 90931917-90931917 on Build GRCh38
rs957292376 Chr.15: 90931693-90931693 on Build GRCh38
rs1024119139 Chr.15: 90931734-90931734 on Build GRCh38
rs189226087 Chr.15: 90931962-90931962 on Build GRCh38
rs749359481 Chr.15: 90931681-90931681 on Build GRCh38
rs762658478 Chr.15: 90931744-90931744 on Build GRCh38
rs554875348 Chr.15: 90931899-90931899 on Build GRCh38
rs572934314 Chr.15: 90931902-90931902 on Build GRCh38
rs780552478 Chr.15: 90932083-90932083 on Build GRCh38
rs149008342 Chr.15: 90932088-90932088 on Build GRCh38
rs137864548 Chr.15: 90931773-90931773 on Build GRCh38
rs762994556 Chr.15: 90931979-90931979 on Build GRCh38
rs771591823 Chr.15: 90931712-90931712 on Build GRCh38
rs773755660 Chr.15: 90931709-90931709 on Build GRCh38
rs757146652 Chr.15: 90931751-90931751 on Build GRCh38
rs780159584 Chr.15: 90931680-90931680 on Build GRCh38
rs762569636 Chr.15: 90931823-90931823 on Build GRCh38
rs746943969 Chr.15: 90931644-90931644 on Build GRCh38
rs768239108 Chr.15: 90931973-90931973 on Build GRCh38
rs752762730 Chr.15: 90932017-90932017 on Build GRCh38
rs751404108 Chr.15: 90931750-90931750 on Build GRCh38
rs773978137 Chr.15: 90931976-90931976 on Build GRCh38
rs932374912 Chr.15: 90931931-90931931 on Build GRCh38
rs377242953 Chr.15: 90931832-90931832 on Build GRCh38
rs762914251 Chr.15: 90931813-90931814 on Build GRCh38
rs773098621 Chr.15: 90931971-90931971 on Build GRCh38
rs111756313 Chr.15: 90932035-90932035 on Build GRCh38
rs776163027 Chr.15: 90931713-90931713 on Build GRCh38
rs369274774 Chr.15: 90932042-90932042 on Build GRCh38
rs371290863 Chr.15: 90931939-90931939 on Build GRCh38
rs764996470 Chr.15: 90931770-90931770 on Build GRCh38
rs775019544 Chr.15: 90931806-90931806 on Build GRCh38
rs199900582 Chr.15: 90931700-90931700 on Build GRCh38
rs538623921 Chr.15: 90931790-90931790 on Build GRCh38
rs767405507 Chr.15: 90931847-90931847 on Build GRCh38
rs748828424 Chr.15: 90931969-90931969 on Build GRCh38
rs764882893 Chr.15: 90931725-90931725 on Build GRCh38
rs752276722 Chr.15: 90931726-90931726 on Build GRCh38
rs34967259 Chr.15: 90932044-90932044 on Build GRCh38
rs761592237 Chr.15: 90931986-90931986 on Build GRCh38
rs758657403 Chr.15: 90931927-90931927 on Build GRCh38
rs766259113 Chr.15: 90931829-90931830 on Build GRCh38
rs752850621 Chr.15: 90931923-90931923 on Build GRCh38
rs144631975 Chr.15: 90931785-90931785 on Build GRCh38
rs746267075 Chr.15: 90932054-90932054 on Build GRCh38
rs769180797 Chr.15: 90931802-90931802 on Build GRCh38
rs781390517 Chr.15: 90932052-90932052 on Build GRCh38
rs761258820 Chr.15: 90931630-90931630 on Build GRCh38
rs138320574 Chr.15: 90931942-90931942 on Build GRCh38
rs750404897 Chr.15: 90931809-90931811 on Build GRCh38
rs200680048 Chr.15: 90931950-90931950 on Build GRCh38
rs778239014 Chr.15: 90931784-90931784 on Build GRCh38
rs747316900 Chr.15: 90931934-90931934 on Build GRCh38
rs375560756 Chr.15: 90931992-90931992 on Build GRCh38
rs772563160 Chr.15: 90931703-90931703 on Build GRCh38
rs1033079309 Chr.15: 90931692-90931692 on Build GRCh38
rs150464100 Chr.15: 90931928-90931928 on Build GRCh38
rs141515875 Chr.15: 90931794-90931794 on Build GRCh38
rs752839383 Chr.15: 90932061-90932063 on Build GRCh38
rs761655471 Chr.15: 90931747-90931748 on Build GRCh38
rs1032987575 Chr.15: 90932032-90932032 on Build GRCh38
rs368232713 Chr.15: 90932008-90932008 on Build GRCh38
rs974810102 Chr.15: 90931658-90931658 on Build GRCh38
rs903150688 Chr.15: 90931652-90931652 on Build GRCh38
rs779018863 Chr.15: 90931694-90931694 on Build GRCh38
rs772475217 Chr.15: 90931780-90931780 on Build GRCh38
rs375962566 Chr.15: 90931807-90931807 on Build GRCh38
rs371199456 Chr.15: 90931659-90931659 on Build GRCh38
rs761245831 Chr.15: 90931710-90931710 on Build GRCh38
rs921011364 Chr.15: 90931904-90931904 on Build GRCh38
rs140820485 Chr.15: 90931781-90931781 on Build GRCh38
rs766367179 Chr.15: 90932010-90932010 on Build GRCh38
rs779117867 Chr.15: 90931777-90931777 on Build GRCh38
rs912023235 Chr.15: 90931943-90931943 on Build GRCh38
rs761610413 Chr.15: 90931846-90931846 on Build GRCh38
rs372856910 Chr.15: 90931721-90931721 on Build GRCh38
rs753899170 Chr.15: 90931763-90931763 on Build GRCh38
rs763844356 Chr.15: 90931745-90931745 on Build GRCh38
rs781667437 Chr.15: 90931948-90931948 on Build GRCh38
rs768857344 Chr.15: 90931691-90931691 on Build GRCh38
rs758569594 Chr.15: 90932024-90932024 on Build GRCh38
rs996629730 Chr.15: 90931999-90931999 on Build GRCh38
rs780997119 Chr.15: 90931759-90931759 on Build GRCh38
rs138596450 Chr.15: 90931788-90931788 on Build GRCh38
rs748441338 Chr.15: 90931698-90931698 on Build GRCh38
rs201409981 Chr.15: 90931849-90931849 on Build GRCh38
rs773106026 Chr.15: 90931994-90931994 on Build GRCh38
rs575157554 Chr.15: 90931829-90931829 on Build GRCh38
rs756539452 Chr.15: 90932069-90932069 on Build GRCh38
rs868576419 Chr.15: 90931987-90931987 on Build GRCh38

View Full Product Details