Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 813055-813549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRR25
Entrez Gene ID
388199
Gene Name
proline rich 25
Gene Aliases
gs64
UniGene
Hs.528461
Gene Chromosome Location
Chr.16: 805443-813861 on Build GRCh38
Transcripts
NM_001013638.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4129453 c.720C>A p.H240Q Chr.16: 813372-813372 on Build GRCh38
4988137 c.734C>T p.P245L Chr.16: 813386-813386 on Build GRCh38
7237454 c.846G>C p.L282F Chr.16: 813498-813498 on Build GRCh38
4129454 c.896G>A p.G299E Chr.16: 813548-813548 on Build GRCh38
50711 c.748A>G p.T250A Chr.16: 813400-813400 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190742719 Chr.16: 813193-813193 on Build GRCh38
rs757048420 Chr.16: 813184-813184 on Build GRCh38
rs756880248 Chr.16: 813245-813245 on Build GRCh38
rs748783476 Chr.16: 813483-813484 on Build GRCh38
rs543041067 Chr.16: 813275-813275 on Build GRCh38
rs79943061 Chr.16: 813271-813271 on Build GRCh38
rs761223517 Chr.16: 813406-813406 on Build GRCh38
rs370642584 Chr.16: 813520-813520 on Build GRCh38
rs199580467 Chr.16: 813369-813369 on Build GRCh38
rs779575399 Chr.16: 813489-813489 on Build GRCh38
rs759066280 Chr.16: 813349-813349 on Build GRCh38
rs74001042 Chr.16: 813175-813175 on Build GRCh38
rs750838761 Chr.16: 813399-813399 on Build GRCh38
rs767347971 Chr.16: 813309-813309 on Build GRCh38
rs201601610 Chr.16: 813418-813418 on Build GRCh38
rs542054372 Chr.16: 813467-813467 on Build GRCh38
rs892945624 Chr.16: 813528-813528 on Build GRCh38
rs780856528 Chr.16: 813255-813255 on Build GRCh38
rs776108016 Chr.16: 813515-813515 on Build GRCh38
rs745757995 Chr.16: 813479-813479 on Build GRCh38
rs781705726 Chr.16: 813316-813316 on Build GRCh38
rs769241973 Chr.16: 813393-813393 on Build GRCh38
rs577082648 Chr.16: 813154-813154 on Build GRCh38
rs766908425 Chr.16: 813449-813449 on Build GRCh38
rs559368481 Chr.16: 813181-813181 on Build GRCh38
rs200304320 Chr.16: 813372-813372 on Build GRCh38
rs760676030 Chr.16: 813337-813337 on Build GRCh38
rs749462085 Chr.16: 813321-813321 on Build GRCh38
rs370207104 Chr.16: 813506-813506 on Build GRCh38
rs113727612 Chr.16: 813227-813227 on Build GRCh38
rs866813201 Chr.16: 813421-813421 on Build GRCh38
rs934980854 Chr.16: 813281-813281 on Build GRCh38
rs770880971 Chr.16: 813378-813378 on Build GRCh38
rs773978873 Chr.16: 813327-813327 on Build GRCh38
rs748677553 Chr.16: 813345-813345 on Build GRCh38
rs774135279 Chr.16: 813502-813502 on Build GRCh38
rs201246779 Chr.16: 813386-813386 on Build GRCh38
rs181972012 Chr.16: 813409-813409 on Build GRCh38
rs200763302 Chr.16: 813446-813446 on Build GRCh38
rs748379245 Chr.16: 813373-813373 on Build GRCh38
rs945703726 Chr.16: 813223-813223 on Build GRCh38
rs138882550 Chr.16: 813240-813240 on Build GRCh38
rs1006601812 Chr.16: 813325-813325 on Build GRCh38
rs57505050 Chr.16: 813096-813096 on Build GRCh38
rs751711111 Chr.16: 813526-813526 on Build GRCh38
rs771813446 Chr.16: 813335-813335 on Build GRCh38
rs368891456 Chr.16: 813461-813461 on Build GRCh38
rs762221677 Chr.16: 813352-813352 on Build GRCh38
rs761605341 Chr.16: 813263-813263 on Build GRCh38
rs990897531 Chr.16: 813395-813395 on Build GRCh38
rs386787600 Chr.16: 813192-813193 on Build GRCh38
rs13331006 Chr.16: 813192-813192 on Build GRCh38
rs142195552 Chr.16: 813422-813422 on Build GRCh38
rs377391659 Chr.16: 813512-813512 on Build GRCh38
rs752338037 Chr.16: 813351-813351 on Build GRCh38
rs757038239 Chr.16: 813470-813470 on Build GRCh38
rs757089417 Chr.16: 813433-813433 on Build GRCh38
rs138733834 Chr.16: 813356-813372 on Build GRCh38
rs759061692 Chr.16: 813389-813389 on Build GRCh38
rs879914117 Chr.16: 813439-813439 on Build GRCh38
rs371962006 Chr.16: 813358-813374 on Build GRCh38
rs898058570 Chr.16: 813452-813452 on Build GRCh38
rs774875194 Chr.16: 813443-813443 on Build GRCh38
rs747878563 Chr.16: 813503-813503 on Build GRCh38
rs964960046 Chr.16: 813142-813142 on Build GRCh38
rs748920054 Chr.16: 813444-813444 on Build GRCh38
rs780232229 Chr.16: 813365-813365 on Build GRCh38
rs747944976 Chr.16: 813330-813330 on Build GRCh38
rs757599272 Chr.16: 813312-813312 on Build GRCh38
rs1050614942 Chr.16: 813087-813087 on Build GRCh38
rs746432200 Chr.16: 813319-813319 on Build GRCh38
rs372178405 Chr.16: 813430-813430 on Build GRCh38
rs768945027 Chr.16: 813323-813323 on Build GRCh38
rs13337795 Chr.16: 813201-813201 on Build GRCh38
rs770253386 Chr.16: 813346-813362 on Build GRCh38
rs770363780 Chr.16: 813344-813344 on Build GRCh38
rs528059818 Chr.16: 813300-813300 on Build GRCh38
rs763002488 Chr.16: 813519-813519 on Build GRCh38
rs942456953 Chr.16: 813427-813427 on Build GRCh38
rs768329630 Chr.16: 813501-813501 on Build GRCh38
rs1017737912 Chr.16: 813111-813111 on Build GRCh38
rs756935372 Chr.16: 813299-813299 on Build GRCh38
rs765838293 Chr.16: 813252-813252 on Build GRCh38
rs746893100 Chr.16: 813377-813377 on Build GRCh38
rs777551939 Chr.16: 813374-813374 on Build GRCh38
rs781090060 Chr.16: 813434-813434 on Build GRCh38
rs200955215 Chr.16: 813310-813310 on Build GRCh38
rs764909225 Chr.16: 813350-813350 on Build GRCh38
rs750892184 Chr.16: 813360-813360 on Build GRCh38
rs13333991 Chr.16: 813498-813498 on Build GRCh38
rs756700254 Chr.16: 813363-813363 on Build GRCh38
rs758323835 Chr.16: 813097-813097 on Build GRCh38
rs200007995 Chr.16: 813482-813482 on Build GRCh38
rs949324457 Chr.16: 813233-813233 on Build GRCh38
rs756254702 Chr.16: 813320-813320 on Build GRCh38
rs768057791 Chr.16: 813358-813358 on Build GRCh38
rs536452586 Chr.16: 813348-813348 on Build GRCh38
rs188105234 Chr.16: 813504-813504 on Build GRCh38
rs185354741 Chr.16: 813466-813466 on Build GRCh38
rs778928894 Chr.16: 813420-813420 on Build GRCh38
rs753488297 Chr.16: 813291-813291 on Build GRCh38
rs1052048706 Chr.16: 813471-813471 on Build GRCh38
rs916377323 Chr.16: 813220-813220 on Build GRCh38
rs754666235 Chr.16: 813295-813295 on Build GRCh38
rs991070232 Chr.16: 813205-813205 on Build GRCh38
rs11860086 Chr.16: 813336-813336 on Build GRCh38
rs1046826703 Chr.16: 813472-813472 on Build GRCh38
rs777192441 Chr.16: 813455-813455 on Build GRCh38
rs373419743 Chr.16: 813510-813510 on Build GRCh38
rs374539155 Chr.16: 813397-813397 on Build GRCh38
rs761099010 Chr.16: 813448-813448 on Build GRCh38
rs765306295 Chr.16: 813464-813464 on Build GRCh38
rs13337837 Chr.16: 813398-813398 on Build GRCh38
rs758636037 Chr.16: 813428-813428 on Build GRCh38
rs564138147 Chr.16: 813279-813279 on Build GRCh38
rs367751056 Chr.16: 813367-813367 on Build GRCh38
rs770369670 Chr.16: 813523-813543 on Build GRCh38
rs767522181 Chr.16: 813265-813265 on Build GRCh38
rs376725139 Chr.16: 813285-813285 on Build GRCh38
rs962361925 Chr.16: 813147-813147 on Build GRCh38
rs990058492 Chr.16: 813218-813218 on Build GRCh38
rs745857557 Chr.16: 813442-813442 on Build GRCh38
rs780851874 Chr.16: 813476-813476 on Build GRCh38
rs542084015 Chr.16: 813396-813396 on Build GRCh38

View Full Product Details