Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 813441-813907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRR25
Entrez Gene ID
388199
Gene Name
proline rich 25
Gene Aliases
gs64
UniGene
Hs.528461
Gene Chromosome Location
Chr.16: 805443-813861 on Build GRCh38
Transcripts
NM_001013638.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7237454 c.846G>C p.L282F Chr.16: 813498-813498 on Build GRCh38
4129454 c.896G>A p.G299E Chr.16: 813548-813548 on Build GRCh38
4428166 c.929C>T p.P310L Chr.16: 813581-813581 on Build GRCh38
4995697 c.959C>T p.S320F Chr.16: 813611-813611 on Build GRCh38
6369931 c.1027G>A p.A343T Chr.16: 813679-813679 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748783476 Chr.16: 813483-813484 on Build GRCh38
rs370642584 Chr.16: 813520-813520 on Build GRCh38
rs779575399 Chr.16: 813489-813489 on Build GRCh38
rs938292768 Chr.16: 813793-813793 on Build GRCh38
rs893877556 Chr.16: 813680-813680 on Build GRCh38
rs759386822 Chr.16: 813797-813797 on Build GRCh38
rs767364579 Chr.16: 813577-813579 on Build GRCh38
rs868608979 Chr.16: 813847-813847 on Build GRCh38
rs752290423 Chr.16: 813595-813595 on Build GRCh38
rs892945624 Chr.16: 813528-813528 on Build GRCh38
rs542054372 Chr.16: 813467-813467 on Build GRCh38
rs956552894 Chr.16: 813729-813729 on Build GRCh38
rs773302543 Chr.16: 813632-813632 on Build GRCh38
rs776108016 Chr.16: 813515-813515 on Build GRCh38
rs745757995 Chr.16: 813479-813479 on Build GRCh38
rs780354281 Chr.16: 813704-813704 on Build GRCh38
rs779833961 Chr.16: 813542-813542 on Build GRCh38
rs1030803736 Chr.16: 813716-813716 on Build GRCh38
rs776983375 Chr.16: 813874-813874 on Build GRCh38
rs370207104 Chr.16: 813506-813506 on Build GRCh38
rs572812936 Chr.16: 813795-813795 on Build GRCh38
rs1012200314 Chr.16: 813694-813694 on Build GRCh38
rs967699057 Chr.16: 813538-813538 on Build GRCh38
rs757481495 Chr.16: 813529-813529 on Build GRCh38
rs750214656 Chr.16: 813537-813537 on Build GRCh38
rs774135279 Chr.16: 813502-813502 on Build GRCh38
rs570851908 Chr.16: 813621-813621 on Build GRCh38
rs956854992 Chr.16: 813574-813574 on Build GRCh38
rs201985000 Chr.16: 813552-813552 on Build GRCh38
rs754981597 Chr.16: 813584-813584 on Build GRCh38
rs553050258 Chr.16: 813698-813698 on Build GRCh38
rs372901067 Chr.16: 813533-813533 on Build GRCh38
rs755976527 Chr.16: 813540-813540 on Build GRCh38
rs751711111 Chr.16: 813526-813526 on Build GRCh38
rs368891456 Chr.16: 813461-813461 on Build GRCh38
rs759509394 Chr.16: 813641-813641 on Build GRCh38
rs773434320 Chr.16: 813801-813801 on Build GRCh38
rs568379488 Chr.16: 813702-813702 on Build GRCh38
rs753709829 Chr.16: 813637-813637 on Build GRCh38
rs569940908 Chr.16: 813608-813608 on Build GRCh38
rs771979067 Chr.16: 813772-813772 on Build GRCh38
rs914918387 Chr.16: 813592-813592 on Build GRCh38
rs761980482 Chr.16: 813577-813577 on Build GRCh38
rs762957772 Chr.16: 813805-813805 on Build GRCh38
rs200432009 Chr.16: 813705-813705 on Build GRCh38
rs377391659 Chr.16: 813512-813512 on Build GRCh38
rs771187077 Chr.16: 813871-813871 on Build GRCh38
rs757038239 Chr.16: 813470-813470 on Build GRCh38
rs530298017 Chr.16: 813559-813559 on Build GRCh38
rs545677951 Chr.16: 813743-813743 on Build GRCh38
rs760738293 Chr.16: 813733-813768 on Build GRCh38
rs775122613 Chr.16: 813713-813713 on Build GRCh38
rs73493265 Chr.16: 813548-813548 on Build GRCh38
rs952996950 Chr.16: 813730-813730 on Build GRCh38
rs763660095 Chr.16: 813673-813673 on Build GRCh38
rs762757256 Chr.16: 813878-813878 on Build GRCh38
rs73493267 Chr.16: 813869-813869 on Build GRCh38
rs772266877 Chr.16: 813778-813778 on Build GRCh38
rs747878563 Chr.16: 813503-813503 on Build GRCh38
rs528136367 Chr.16: 813546-813546 on Build GRCh38
rs752734405 Chr.16: 813654-813654 on Build GRCh38
rs779254123 Chr.16: 813837-813837 on Build GRCh38
rs1005560874 Chr.16: 813628-813628 on Build GRCh38
rs757838402 Chr.16: 813666-813666 on Build GRCh38
rs540240413 Chr.16: 813855-813855 on Build GRCh38
rs756953222 Chr.16: 813701-813701 on Build GRCh38
rs763452267 Chr.16: 813563-813563 on Build GRCh38
rs374481521 Chr.16: 813564-813564 on Build GRCh38
rs922336739 Chr.16: 813609-813609 on Build GRCh38
rs77176097 Chr.16: 813774-813774 on Build GRCh38
rs760373385 Chr.16: 813579-813579 on Build GRCh38
rs763002488 Chr.16: 813519-813519 on Build GRCh38
rs779029940 Chr.16: 813639-813639 on Build GRCh38
rs200715647 Chr.16: 813620-813620 on Build GRCh38
rs774433518 Chr.16: 813570-813570 on Build GRCh38
rs768329630 Chr.16: 813501-813501 on Build GRCh38
rs775374083 Chr.16: 813877-813877 on Build GRCh38
rs374546092 Chr.16: 813810-813810 on Build GRCh38
rs767132748 Chr.16: 813820-813820 on Build GRCh38
rs764059378 Chr.16: 813884-813884 on Build GRCh38
rs1030273788 Chr.16: 813721-813721 on Build GRCh38
rs759066564 Chr.16: 813576-813577 on Build GRCh38
rs1015929806 Chr.16: 813861-813861 on Build GRCh38
rs971273249 Chr.16: 813885-813885 on Build GRCh38
rs548399002 Chr.16: 813582-813582 on Build GRCh38
rs201117285 Chr.16: 813610-813610 on Build GRCh38
rs13333991 Chr.16: 813498-813498 on Build GRCh38
rs747152856 Chr.16: 813787-813787 on Build GRCh38
rs767292892 Chr.16: 813534-813534 on Build GRCh38
rs200007995 Chr.16: 813482-813482 on Build GRCh38
rs771311788 Chr.16: 813792-813792 on Build GRCh38
rs188105234 Chr.16: 813504-813504 on Build GRCh38
rs185354741 Chr.16: 813466-813466 on Build GRCh38
rs765186221 Chr.16: 813799-813799 on Build GRCh38
rs777273033 Chr.16: 813599-813599 on Build GRCh38
rs557277746 Chr.16: 813731-813731 on Build GRCh38
rs867608312 Chr.16: 813625-813625 on Build GRCh38
rs755331972 Chr.16: 813836-813836 on Build GRCh38
rs757970042 Chr.16: 813597-813597 on Build GRCh38
rs1052048706 Chr.16: 813471-813471 on Build GRCh38
rs748546885 Chr.16: 813838-813838 on Build GRCh38
rs761776681 Chr.16: 813626-813626 on Build GRCh38
rs770246432 Chr.16: 813560-813560 on Build GRCh38
rs760957657 Chr.16: 813633-813633 on Build GRCh38
rs769650046 Chr.16: 813876-813876 on Build GRCh38
rs767602185 Chr.16: 813578-813578 on Build GRCh38
rs367695897 Chr.16: 813588-813588 on Build GRCh38
rs751108435 Chr.16: 813817-813817 on Build GRCh38
rs747747668 Chr.16: 813554-813554 on Build GRCh38
rs985781245 Chr.16: 813734-813734 on Build GRCh38
rs1046826703 Chr.16: 813472-813472 on Build GRCh38
rs866140228 Chr.16: 813829-813829 on Build GRCh38
rs373419743 Chr.16: 813510-813510 on Build GRCh38
rs746739000 Chr.16: 813605-813605 on Build GRCh38
rs997804396 Chr.16: 813717-813717 on Build GRCh38
rs746163815 Chr.16: 813875-813875 on Build GRCh38
rs756755762 Chr.16: 813818-813818 on Build GRCh38
rs765306295 Chr.16: 813464-813464 on Build GRCh38
rs774054085 Chr.16: 813532-813533 on Build GRCh38
rs575550943 Chr.16: 813735-813735 on Build GRCh38
rs777677029 Chr.16: 813868-813868 on Build GRCh38
rs915281578 Chr.16: 813809-813809 on Build GRCh38
rs770369670 Chr.16: 813523-813543 on Build GRCh38
rs1015960817 Chr.16: 813738-813738 on Build GRCh38
rs949580914 Chr.16: 813783-813783 on Build GRCh38
rs371579704 Chr.16: 813634-813634 on Build GRCh38
rs963455539 Chr.16: 813740-813740 on Build GRCh38
rs889018396 Chr.16: 813850-813850 on Build GRCh38
rs1037944828 Chr.16: 813647-813647 on Build GRCh38
rs535020414 Chr.16: 813631-813631 on Build GRCh38
rs919471876 Chr.16: 813601-813601 on Build GRCh38
rs780851874 Chr.16: 813476-813476 on Build GRCh38
rs372301778 Chr.16: 813828-813828 on Build GRCh38

View Full Product Details