Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2759055-2759555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRRM2
Entrez Gene ID
23524
Gene Name
serine/arginine repetitive matrix 2
Gene Aliases
300-KD, CWF21, Cwc21, HSPC075, SRL300, SRm300
UniGene
Hs.433343
Gene Chromosome Location
Chr.16: 2752329-2771412 on Build GRCh38
Transcripts
NM_016333.3 XM_005255226.1 XM_006720874.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1227583 c.739C>T p.R247W Chr.16: 2759401-2759401 on Build GRCh38
4059713 c.733C>T p.R245* Chr.16: 2759395-2759395 on Build GRCh38
6143910 c.657-1G>T p.? Chr.16: 2759139-2759139 on Build GRCh38
7255256 c.680G>T p.R227L Chr.16: 2759163-2759163 on Build GRCh38
3690886 c.734G>A p.R245Q Chr.16: 2759396-2759396 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs946200782 Chr.16: 2759433-2759433 on Build GRCh38
rs369302841 Chr.16: 2759195-2759195 on Build GRCh38
rs746749243 Chr.16: 2759140-2759140 on Build GRCh38
rs767973282 Chr.16: 2759109-2759109 on Build GRCh38
rs368438407 Chr.16: 2759105-2759105 on Build GRCh38
rs772221460 Chr.16: 2759360-2759360 on Build GRCh38
rs781006645 Chr.16: 2759325-2759325 on Build GRCh38
rs747331898 Chr.16: 2759374-2759374 on Build GRCh38
rs762505544 Chr.16: 2759307-2759307 on Build GRCh38
rs375502139 Chr.16: 2759121-2759121 on Build GRCh38
rs548087176 Chr.16: 2759489-2759489 on Build GRCh38
rs370809600 Chr.16: 2759520-2759520 on Build GRCh38
rs764710884 Chr.16: 2759182-2759182 on Build GRCh38
rs778433960 Chr.16: 2759128-2759128 on Build GRCh38
rs945778167 Chr.16: 2759227-2759227 on Build GRCh38
rs765849114 Chr.16: 2759308-2759308 on Build GRCh38
rs747274700 Chr.16: 2759449-2759449 on Build GRCh38
rs1028036014 Chr.16: 2759432-2759433 on Build GRCh38
rs781209469 Chr.16: 2759307-2759308 on Build GRCh38
rs766640520 Chr.16: 2759101-2759101 on Build GRCh38
rs758049508 Chr.16: 2759193-2759193 on Build GRCh38
rs755527110 Chr.16: 2759136-2759136 on Build GRCh38
rs114827582 Chr.16: 2759297-2759297 on Build GRCh38
rs181779611 Chr.16: 2759225-2759225 on Build GRCh38
rs761938794 Chr.16: 2759085-2759085 on Build GRCh38
rs578021877 Chr.16: 2759443-2759443 on Build GRCh38
rs1009264179 Chr.16: 2759367-2759367 on Build GRCh38
rs570499242 Chr.16: 2759176-2759176 on Build GRCh38
rs1055492882 Chr.16: 2759257-2759257 on Build GRCh38
rs774609095 Chr.16: 2759215-2759215 on Build GRCh38
rs201096433 Chr.16: 2759143-2759143 on Build GRCh38
rs369676177 Chr.16: 2759202-2759202 on Build GRCh38
rs773888417 Chr.16: 2759525-2759525 on Build GRCh38
rs759893980 Chr.16: 2759175-2759175 on Build GRCh38
rs752421519 Chr.16: 2759420-2759420 on Build GRCh38
rs752474487 Chr.16: 2759333-2759333 on Build GRCh38
rs759717037 Chr.16: 2759218-2759218 on Build GRCh38
rs749350640 Chr.16: 2759209-2759209 on Build GRCh38
rs763833569 Chr.16: 2759430-2759430 on Build GRCh38
rs147445433 Chr.16: 2759158-2759158 on Build GRCh38
rs773484091 Chr.16: 2759164-2759164 on Build GRCh38
rs765627602 Chr.16: 2759126-2759127 on Build GRCh38
rs754330004 Chr.16: 2759306-2759306 on Build GRCh38
rs76786072 Chr.16: 2759445-2759445 on Build GRCh38
rs770133215 Chr.16: 2759524-2759524 on Build GRCh38
rs775690276 Chr.16: 2759361-2759361 on Build GRCh38
rs938326612 Chr.16: 2759254-2759254 on Build GRCh38
rs367845541 Chr.16: 2759221-2759221 on Build GRCh38
rs1041867423 Chr.16: 2759231-2759231 on Build GRCh38
rs377552572 Chr.16: 2759082-2759082 on Build GRCh38
rs777167753 Chr.16: 2759387-2759387 on Build GRCh38
rs749240100 Chr.16: 2759487-2759487 on Build GRCh38
rs755953001 Chr.16: 2759424-2759424 on Build GRCh38
rs781203735 Chr.16: 2759199-2759199 on Build GRCh38
rs761236436 Chr.16: 2759181-2759181 on Build GRCh38
rs777718531 Chr.16: 2759339-2759339 on Build GRCh38
rs762296235 Chr.16: 2759392-2759392 on Build GRCh38
rs771119109 Chr.16: 2759169-2759169 on Build GRCh38
rs754557612 Chr.16: 2759302-2759307 on Build GRCh38
rs773680637 Chr.16: 2759397-2759397 on Build GRCh38
rs370966028 Chr.16: 2759098-2759098 on Build GRCh38
rs138293757 Chr.16: 2759395-2759395 on Build GRCh38
rs748140896 Chr.16: 2759152-2759152 on Build GRCh38
rs374586831 Chr.16: 2759258-2759258 on Build GRCh38
rs776921584 Chr.16: 2759518-2759518 on Build GRCh38
rs201317797 Chr.16: 2759148-2759148 on Build GRCh38
rs202002640 Chr.16: 2759401-2759401 on Build GRCh38
rs767239423 Chr.16: 2759411-2759411 on Build GRCh38
rs536153993 Chr.16: 2759275-2759275 on Build GRCh38
rs372623157 Chr.16: 2759358-2759358 on Build GRCh38
rs375507849 Chr.16: 2759200-2759200 on Build GRCh38
rs745499842 Chr.16: 2759134-2759134 on Build GRCh38
rs113301435 Chr.16: 2759220-2759220 on Build GRCh38
rs72768761 Chr.16: 2759103-2759103 on Build GRCh38
rs377740821 Chr.16: 2759219-2759219 on Build GRCh38
rs544572991 Chr.16: 2759341-2759341 on Build GRCh38
rs777924298 Chr.16: 2759345-2759345 on Build GRCh38
rs756176894 Chr.16: 2759423-2759423 on Build GRCh38
rs751224645 Chr.16: 2759317-2759317 on Build GRCh38
rs550406093 Chr.16: 2759167-2759167 on Build GRCh38
rs113294042 Chr.16: 2759261-2759261 on Build GRCh38
rs769028384 Chr.16: 2759375-2759375 on Build GRCh38
rs774469947 Chr.16: 2759171-2759171 on Build GRCh38
rs1015496573 Chr.16: 2759455-2759455 on Build GRCh38
rs368595576 Chr.16: 2759294-2759294 on Build GRCh38
rs199955975 Chr.16: 2759113-2759113 on Build GRCh38
rs748093655 Chr.16: 2759409-2759419 on Build GRCh38
rs199522494 Chr.16: 2759221-2759222 on Build GRCh38
rs528408344 Chr.16: 2759471-2759471 on Build GRCh38
rs200729280 Chr.16: 2759135-2759135 on Build GRCh38
rs190699786 Chr.16: 2759444-2759444 on Build GRCh38
rs754739314 Chr.16: 2759324-2759324 on Build GRCh38
rs751223077 Chr.16: 2759196-2759196 on Build GRCh38
rs749231720 Chr.16: 2759340-2759340 on Build GRCh38
rs961271037 Chr.16: 2759468-2759468 on Build GRCh38
rs765459608 Chr.16: 2759087-2759087 on Build GRCh38
rs987896693 Chr.16: 2759112-2759112 on Build GRCh38
rs753171202 Chr.16: 2759179-2759179 on Build GRCh38
rs749886915 Chr.16: 2759191-2759191 on Build GRCh38
rs768361237 Chr.16: 2759145-2759145 on Build GRCh38
rs750813861 Chr.16: 2759191-2759192 on Build GRCh38
rs780637952 Chr.16: 2759201-2759201 on Build GRCh38
rs777600039 Chr.16: 2759430-2759431 on Build GRCh38
rs955493620 Chr.16: 2759347-2759347 on Build GRCh38
rs202233656 Chr.16: 2759197-2759197 on Build GRCh38
rs199511291 Chr.16: 2759210-2759210 on Build GRCh38
rs111721270 Chr.16: 2759405-2759405 on Build GRCh38
rs764515417 Chr.16: 2759302-2759302 on Build GRCh38
rs374564867 Chr.16: 2759224-2759224 on Build GRCh38
rs773541132 Chr.16: 2759094-2759094 on Build GRCh38
rs894164407 Chr.16: 2759531-2759531 on Build GRCh38
rs759069190 Chr.16: 2759526-2759526 on Build GRCh38
rs778926980 Chr.16: 2759355-2759355 on Build GRCh38
rs371448888 Chr.16: 2759188-2759188 on Build GRCh38
rs778919400 Chr.16: 2759435-2759435 on Build GRCh38
rs368275451 Chr.16: 2759450-2759450 on Build GRCh38
rs938805059 Chr.16: 2759336-2759336 on Build GRCh38
rs183454041 Chr.16: 2759507-2759507 on Build GRCh38
rs908437160 Chr.16: 2759272-2759272 on Build GRCh38
rs747144306 Chr.16: 2759432-2759432 on Build GRCh38
rs762095152 Chr.16: 2759114-2759115 on Build GRCh38
rs1006040964 Chr.16: 2759119-2759119 on Build GRCh38
rs757848799 Chr.16: 2759193-2759193 on Build GRCh38
rs756006206 Chr.16: 2759334-2759334 on Build GRCh38
rs990813132 Chr.16: 2759417-2759417 on Build GRCh38
rs907492920 Chr.16: 2759204-2759204 on Build GRCh38
rs754229725 Chr.16: 2759203-2759203 on Build GRCh38
rs370053653 Chr.16: 2759241-2759241 on Build GRCh38
rs1041361976 Chr.16: 2759088-2759088 on Build GRCh38
rs1018257369 Chr.16: 2759396-2759396 on Build GRCh38
rs879705289 Chr.16: 2759464-2759464 on Build GRCh38

View Full Product Details