Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2967704-2968198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KREMEN2
Entrez Gene ID
79412
Gene Name
kringle containing transmembrane protein 2
Gene Aliases
KRM2
UniGene
Hs.661128
Gene Chromosome Location
Chr.16: 2964216-2968383 on Build GRCh38
Transcripts
NM_001253725.1 NM_001253726.1 NM_024507.3 NM_172229.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1189150 c.1213G>C p.A405P Chr.16: 2967844-2967844 on Build GRCh38
6522516 c.1293C>A p.S431S Chr.16: 2967924-2967924 on Build GRCh38
5456536 c.1207C>T p.P403S Chr.16: 2967838-2967838 on Build GRCh38
6697767 c.1323C>T p.A441A Chr.16: 2967954-2967954 on Build GRCh38
969789 c.1371G>A p.S457S Chr.16: 2968002-2968002 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755438945 Chr.16: 2967927-2967927 on Build GRCh38
rs998032332 Chr.16: 2967824-2967824 on Build GRCh38
rs898442466 Chr.16: 2967888-2967888 on Build GRCh38
rs750140044 Chr.16: 2968023-2968023 on Build GRCh38
rs765382890 Chr.16: 2967882-2967882 on Build GRCh38
rs192742743 Chr.16: 2968002-2968002 on Build GRCh38
rs747033806 Chr.16: 2967953-2967953 on Build GRCh38
rs760104507 Chr.16: 2967981-2967981 on Build GRCh38
rs779963648 Chr.16: 2967952-2967952 on Build GRCh38
rs775100993 Chr.16: 2968098-2968098 on Build GRCh38
rs11866302 Chr.16: 2967853-2967853 on Build GRCh38
rs768463228 Chr.16: 2967959-2967959 on Build GRCh38
rs939519944 Chr.16: 2968036-2968036 on Build GRCh38
rs758118306 Chr.16: 2968026-2968026 on Build GRCh38
rs369237826 Chr.16: 2967997-2967997 on Build GRCh38
rs368944492 Chr.16: 2967964-2967964 on Build GRCh38
rs775520120 Chr.16: 2967843-2967843 on Build GRCh38
rs935044083 Chr.16: 2967756-2967756 on Build GRCh38
rs1003559589 Chr.16: 2968119-2968119 on Build GRCh38
rs766711665 Chr.16: 2967902-2967902 on Build GRCh38
rs745593081 Chr.16: 2967828-2967828 on Build GRCh38
rs1018988137 Chr.16: 2967966-2967966 on Build GRCh38
rs866940296 Chr.16: 2968012-2968012 on Build GRCh38
rs924183320 Chr.16: 2968030-2968030 on Build GRCh38
rs868797771 Chr.16: 2967915-2967915 on Build GRCh38
rs754734893 Chr.16: 2967733-2967734 on Build GRCh38
rs11861396 Chr.16: 2967780-2967780 on Build GRCh38
rs758916318 Chr.16: 2967735-2967735 on Build GRCh38
rs752152507 Chr.16: 2967906-2967906 on Build GRCh38
rs1022943694 Chr.16: 2968051-2968051 on Build GRCh38
rs570763542 Chr.16: 2968038-2968038 on Build GRCh38
rs758902019 Chr.16: 2967724-2967724 on Build GRCh38
rs780947988 Chr.16: 2968069-2968069 on Build GRCh38
rs992339098 Chr.16: 2968039-2968039 on Build GRCh38
rs1056625315 Chr.16: 2967845-2967845 on Build GRCh38
rs760507895 Chr.16: 2967849-2967849 on Build GRCh38
rs199874007 Chr.16: 2967969-2967969 on Build GRCh38
rs745680796 Chr.16: 2967946-2967946 on Build GRCh38
rs1004735522 Chr.16: 2967861-2967861 on Build GRCh38
rs1038469891 Chr.16: 2968031-2968031 on Build GRCh38
rs768640489 Chr.16: 2968058-2968058 on Build GRCh38
rs939860701 Chr.16: 2968013-2968013 on Build GRCh38
rs188657773 Chr.16: 2967974-2967974 on Build GRCh38
rs1052175579 Chr.16: 2967759-2967759 on Build GRCh38
rs777119720 Chr.16: 2967781-2967781 on Build GRCh38
rs751859934 Chr.16: 2967977-2967977 on Build GRCh38
rs774303459 Chr.16: 2967810-2967810 on Build GRCh38
rs756940584 Chr.16: 2967933-2967933 on Build GRCh38
rs746721693 Chr.16: 2968040-2968040 on Build GRCh38
rs915531563 Chr.16: 2967786-2967786 on Build GRCh38
rs916370117 Chr.16: 2968057-2968057 on Build GRCh38
rs898180874 Chr.16: 2968123-2968123 on Build GRCh38
rs747683556 Chr.16: 2967775-2967775 on Build GRCh38
rs770031834 Chr.16: 2967967-2967967 on Build GRCh38
rs201657950 Chr.16: 2967948-2967948 on Build GRCh38
rs771223154 Chr.16: 2968013-2968014 on Build GRCh38
rs767847888 Chr.16: 2967989-2967989 on Build GRCh38
rs547172813 Chr.16: 2967785-2967785 on Build GRCh38
rs749459840 Chr.16: 2967932-2967933 on Build GRCh38
rs946961396 Chr.16: 2967800-2967800 on Build GRCh38
rs921212346 Chr.16: 2967987-2967987 on Build GRCh38
rs778668696 Chr.16: 2967945-2967945 on Build GRCh38
rs947775196 Chr.16: 2968103-2968103 on Build GRCh38
rs780472627 Chr.16: 2967739-2967739 on Build GRCh38
rs750788121 Chr.16: 2967889-2967889 on Build GRCh38
rs750770074 Chr.16: 2967734-2967734 on Build GRCh38
rs780468793 Chr.16: 2967737-2967737 on Build GRCh38
rs762426768 Chr.16: 2968140-2968140 on Build GRCh38
rs778842189 Chr.16: 2968121-2968121 on Build GRCh38
rs1042585587 Chr.16: 2967821-2967821 on Build GRCh38
rs199819672 Chr.16: 2967965-2967965 on Build GRCh38
rs781109968 Chr.16: 2967835-2967835 on Build GRCh38
rs994470655 Chr.16: 2967918-2967918 on Build GRCh38
rs748278158 Chr.16: 2968070-2968070 on Build GRCh38
rs117373306 Chr.16: 2967851-2967851 on Build GRCh38
rs987890635 Chr.16: 2967957-2967957 on Build GRCh38
rs939506323 Chr.16: 2967838-2967838 on Build GRCh38
rs758718074 Chr.16: 2967898-2967898 on Build GRCh38
rs772168114 Chr.16: 2967837-2967837 on Build GRCh38
rs907187514 Chr.16: 2968115-2968115 on Build GRCh38
rs1047429303 Chr.16: 2968173-2968173 on Build GRCh38

View Full Product Details