Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3224258-3224752 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF200
Entrez Gene ID
7752
Gene Name
zinc finger protein 200
Gene Aliases
-
UniGene
Hs.632222
Gene Chromosome Location
Chr.16: 3222325-3236235 on Build GRCh38
Transcripts
NM_001145446.1 NM_001145447.1 NM_001145448.1 NM_003454.3 NM_198087.2 NM_198088.2 XM_005255556.1 XM_006720940.2 XM_006720941.2 XM_017023647.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
435198 c.703G>C p.D235H Chr.16: 3224374-3224374 on Build GRCh38
258427 c.507A>T p.E169D Chr.16: 3224570-3224570 on Build GRCh38
6052070 c.558G>T p.M186I Chr.16: 3224519-3224519 on Build GRCh38
238476 c.528T>C p.Y176Y Chr.16: 3224549-3224549 on Build GRCh38
2918898 c.590A>T p.D197V Chr.16: 3224487-3224487 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776609976 Chr.16: 3224582-3224582 on Build GRCh38
rs781386830 Chr.16: 3224512-3224512 on Build GRCh38
rs778658297 Chr.16: 3224534-3224534 on Build GRCh38
rs758936606 Chr.16: 3224403-3224403 on Build GRCh38
rs761950506 Chr.16: 3224470-3224470 on Build GRCh38
rs745705226 Chr.16: 3224550-3224550 on Build GRCh38
rs750812653 Chr.16: 3224482-3224482 on Build GRCh38
rs139877194 Chr.16: 3224297-3224297 on Build GRCh38
rs145395897 Chr.16: 3224344-3224344 on Build GRCh38
rs867869038 Chr.16: 3224457-3224457 on Build GRCh38
rs947223799 Chr.16: 3224463-3224463 on Build GRCh38
rs772155931 Chr.16: 3224450-3224450 on Build GRCh38
rs774375276 Chr.16: 3224653-3224653 on Build GRCh38
rs781695047 Chr.16: 3224626-3224626 on Build GRCh38
rs770519795 Chr.16: 3224533-3224533 on Build GRCh38
rs760450410 Chr.16: 3224643-3224644 on Build GRCh38
rs368623860 Chr.16: 3224655-3224655 on Build GRCh38
rs146563741 Chr.16: 3224356-3224356 on Build GRCh38
rs375643387 Chr.16: 3224622-3224623 on Build GRCh38
rs537270121 Chr.16: 3224357-3224357 on Build GRCh38
rs117151961 Chr.16: 3224286-3224286 on Build GRCh38
rs760544631 Chr.16: 3224572-3224572 on Build GRCh38
rs769154131 Chr.16: 3224422-3224422 on Build GRCh38
rs759556381 Chr.16: 3224352-3224352 on Build GRCh38
rs78240929 Chr.16: 3224469-3224469 on Build GRCh38
rs968971839 Chr.16: 3224718-3224718 on Build GRCh38
rs779162806 Chr.16: 3224379-3224379 on Build GRCh38
rs958421323 Chr.16: 3224675-3224675 on Build GRCh38
rs957416578 Chr.16: 3224725-3224725 on Build GRCh38
rs4577090 Chr.16: 3224487-3224487 on Build GRCh38
rs368931922 Chr.16: 3224608-3224608 on Build GRCh38
rs117357111 Chr.16: 3224398-3224398 on Build GRCh38
rs755440895 Chr.16: 3224615-3224615 on Build GRCh38
rs371227621 Chr.16: 3224595-3224595 on Build GRCh38
rs1004174590 Chr.16: 3224681-3224681 on Build GRCh38
rs779544740 Chr.16: 3224303-3224303 on Build GRCh38
rs748764909 Chr.16: 3224529-3224529 on Build GRCh38
rs78570235 Chr.16: 3224628-3224628 on Build GRCh38
rs903073794 Chr.16: 3224508-3224508 on Build GRCh38
rs1027667734 Chr.16: 3224634-3224634 on Build GRCh38
rs769461560 Chr.16: 3224327-3224327 on Build GRCh38
rs775538837 Chr.16: 3224461-3224461 on Build GRCh38
rs780576932 Chr.16: 3224489-3224489 on Build GRCh38
rs200496883 Chr.16: 3224322-3224322 on Build GRCh38
rs772715804 Chr.16: 3224343-3224343 on Build GRCh38
rs978595749 Chr.16: 3224730-3224730 on Build GRCh38
rs567943222 Chr.16: 3224716-3224716 on Build GRCh38
rs778776875 Chr.16: 3224647-3224647 on Build GRCh38
rs751860360 Chr.16: 3224613-3224613 on Build GRCh38
rs1023776161 Chr.16: 3224305-3224305 on Build GRCh38
rs140076513 Chr.16: 3224704-3224704 on Build GRCh38
rs769397036 Chr.16: 3224594-3224594 on Build GRCh38
rs760906126 Chr.16: 3224462-3224462 on Build GRCh38
rs755704232 Chr.16: 3224509-3224509 on Build GRCh38
rs951620433 Chr.16: 3224664-3224664 on Build GRCh38
rs901106018 Chr.16: 3224616-3224616 on Build GRCh38
rs745503240 Chr.16: 3224649-3224649 on Build GRCh38
rs747150914 Chr.16: 3224349-3224349 on Build GRCh38
rs752762933 Chr.16: 3224280-3224280 on Build GRCh38
rs371628901 Chr.16: 3224294-3224294 on Build GRCh38
rs772510762 Chr.16: 3224314-3224314 on Build GRCh38
rs563505894 Chr.16: 3224639-3224639 on Build GRCh38
rs1056780222 Chr.16: 3224530-3224530 on Build GRCh38
rs757702140 Chr.16: 3224372-3224372 on Build GRCh38
rs374158620 Chr.16: 3224312-3224312 on Build GRCh38
rs1024388850 Chr.16: 3224685-3224685 on Build GRCh38
rs764420030 Chr.16: 3224361-3224361 on Build GRCh38
rs749302233 Chr.16: 3224527-3224527 on Build GRCh38
rs780491330 Chr.16: 3224413-3224413 on Build GRCh38
rs151034617 Chr.16: 3224416-3224416 on Build GRCh38
rs895544593 Chr.16: 3224281-3224281 on Build GRCh38
rs878931621 Chr.16: 3224690-3224690 on Build GRCh38
rs1047176968 Chr.16: 3224622-3224622 on Build GRCh38
rs775628418 Chr.16: 3224563-3224563 on Build GRCh38
rs754005156 Chr.16: 3224363-3224363 on Build GRCh38
rs774405089 Chr.16: 3224350-3224350 on Build GRCh38
rs748878377 Chr.16: 3224428-3224428 on Build GRCh38
rs778087363 Chr.16: 3224334-3224334 on Build GRCh38
rs752054883 Chr.16: 3224502-3224502 on Build GRCh38
rs770554903 Chr.16: 3224441-3224441 on Build GRCh38
rs772382623 Chr.16: 3224621-3224624 on Build GRCh38
rs777547082 Chr.16: 3224426-3224426 on Build GRCh38
rs775191798 Chr.16: 3224623-3224624 on Build GRCh38
rs924526475 Chr.16: 3224731-3224731 on Build GRCh38
rs185505006 Chr.16: 3224693-3224693 on Build GRCh38
rs1005613414 Chr.16: 3224646-3224646 on Build GRCh38
rs776612634 Chr.16: 3224660-3224660 on Build GRCh38
rs574753988 Chr.16: 3224603-3224603 on Build GRCh38
rs368487467 Chr.16: 3224619-3224619 on Build GRCh38
rs201377318 Chr.16: 3224612-3224612 on Build GRCh38
rs149793470 Chr.16: 3224488-3224488 on Build GRCh38
rs771990277 Chr.16: 3224557-3224557 on Build GRCh38
rs4589551 Chr.16: 3224485-3224485 on Build GRCh38
rs141010410 Chr.16: 3224446-3224446 on Build GRCh38
rs200936424 Chr.16: 3224650-3224650 on Build GRCh38
rs944090880 Chr.16: 3224378-3224378 on Build GRCh38
rs200302589 Chr.16: 3224658-3224658 on Build GRCh38
rs189621979 Chr.16: 3224713-3224713 on Build GRCh38
rs757403207 Chr.16: 3224476-3224476 on Build GRCh38
rs201398970 Chr.16: 3224661-3224661 on Build GRCh38
rs765495787 Chr.16: 3224601-3224601 on Build GRCh38
rs539008419 Chr.16: 3224621-3224622 on Build GRCh38
rs539585480 Chr.16: 3224444-3224444 on Build GRCh38
rs910607971 Chr.16: 3224540-3224540 on Build GRCh38
rs193284245 Chr.16: 3224511-3224511 on Build GRCh38
rs921348008 Chr.16: 3224709-3224709 on Build GRCh38
rs542164536 Chr.16: 3224504-3224504 on Build GRCh38

View Full Product Details