Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 23688236-23688777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLK1
Entrez Gene ID
5347
Gene Name
polo like kinase 1
Gene Aliases
PLK, STPK13
UniGene
Hs.592049
Gene Chromosome Location
Chr.16: 23678772-23690367 on Build GRCh38
Transcripts
NM_005030.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6555519 c.1264G>T p.G422C Chr.16: 23688739-23688739 on Build GRCh38
5820908 c.1200T>A p.A400A Chr.16: 23688675-23688675 on Build GRCh38
7299388 c.1264G>A p.G422S Chr.16: 23688739-23688739 on Build GRCh38
5499251 c.1265G>A p.G422D Chr.16: 23688740-23688740 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764164601 Chr.16: 23688645-23688648 on Build GRCh38
rs893416644 Chr.16: 23688325-23688325 on Build GRCh38
rs761629266 Chr.16: 23688720-23688720 on Build GRCh38
rs950218297 Chr.16: 23688522-23688522 on Build GRCh38
rs548840490 Chr.16: 23688444-23688444 on Build GRCh38
rs1054907482 Chr.16: 23688603-23688604 on Build GRCh38
rs776583708 Chr.16: 23688649-23688650 on Build GRCh38
rs914584460 Chr.16: 23688608-23688608 on Build GRCh38
rs891315667 Chr.16: 23688353-23688353 on Build GRCh38
rs756655328 Chr.16: 23688630-23688630 on Build GRCh38
rs552293848 Chr.16: 23688635-23688635 on Build GRCh38
rs573003997 Chr.16: 23688320-23688320 on Build GRCh38
rs961778861 Chr.16: 23688424-23688427 on Build GRCh38
rs903258006 Chr.16: 23688299-23688299 on Build GRCh38
rs752889775 Chr.16: 23688627-23688627 on Build GRCh38
rs950542725 Chr.16: 23688388-23688388 on Build GRCh38
rs994540511 Chr.16: 23688352-23688352 on Build GRCh38
rs913337163 Chr.16: 23688514-23688514 on Build GRCh38
rs755705570 Chr.16: 23688494-23688494 on Build GRCh38
rs374194609 Chr.16: 23688665-23688665 on Build GRCh38
rs771773851 Chr.16: 23688661-23688661 on Build GRCh38
rs878941582 Chr.16: 23688269-23688269 on Build GRCh38
rs368823668 Chr.16: 23688650-23688650 on Build GRCh38
rs767691148 Chr.16: 23688626-23688626 on Build GRCh38
rs1026030819 Chr.16: 23688374-23688374 on Build GRCh38
rs570069218 Chr.16: 23688607-23688607 on Build GRCh38
rs930316046 Chr.16: 23688643-23688643 on Build GRCh38
rs915804727 Chr.16: 23688513-23688513 on Build GRCh38
rs143382893 Chr.16: 23688729-23688729 on Build GRCh38
rs564856373 Chr.16: 23688447-23688447 on Build GRCh38
rs371567827 Chr.16: 23688443-23688443 on Build GRCh38
rs932613060 Chr.16: 23688610-23688610 on Build GRCh38
rs866791256 Chr.16: 23688472-23688472 on Build GRCh38
rs745524508 Chr.16: 23688660-23688660 on Build GRCh38
rs561105453 Chr.16: 23688499-23688499 on Build GRCh38
rs35070139 Chr.16: 23688586-23688587 on Build GRCh38
rs780702557 Chr.16: 23688558-23688558 on Build GRCh38
rs376439699 Chr.16: 23688756-23688756 on Build GRCh38
rs540416218 Chr.16: 23688335-23688335 on Build GRCh38
rs759630351 Chr.16: 23688618-23688618 on Build GRCh38
rs761831538 Chr.16: 23688389-23688389 on Build GRCh38
rs202103204 Chr.16: 23688631-23688631 on Build GRCh38
rs1054700771 Chr.16: 23688324-23688324 on Build GRCh38
rs988854139 Chr.16: 23688504-23688504 on Build GRCh38
rs201491344 Chr.16: 23688685-23688685 on Build GRCh38
rs1019188887 Chr.16: 23688736-23688736 on Build GRCh38
rs1018806117 Chr.16: 23688391-23688391 on Build GRCh38
rs16972803 Chr.16: 23688596-23688596 on Build GRCh38
rs530358788 Chr.16: 23688416-23688416 on Build GRCh38
rs201301705 Chr.16: 23688632-23688632 on Build GRCh38
rs1001823826 Chr.16: 23688301-23688301 on Build GRCh38
rs529896437 Chr.16: 23688534-23688534 on Build GRCh38
rs996060524 Chr.16: 23688753-23688753 on Build GRCh38
rs141358144 Chr.16: 23688476-23688476 on Build GRCh38
rs746563979 Chr.16: 23688673-23688673 on Build GRCh38
rs993605006 Chr.16: 23688436-23688436 on Build GRCh38
rs924732224 Chr.16: 23688568-23688568 on Build GRCh38
rs541328261 Chr.16: 23688488-23688488 on Build GRCh38
rs2230914 Chr.16: 23688738-23688738 on Build GRCh38
rs1008378948 Chr.16: 23688356-23688356 on Build GRCh38
rs147815783 Chr.16: 23688377-23688377 on Build GRCh38
rs760834089 Chr.16: 23688627-23688628 on Build GRCh38
rs35008953 Chr.16: 23688662-23688662 on Build GRCh38
rs565735478 Chr.16: 23688691-23688691 on Build GRCh38

View Full Product Details