Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30187622-30188125 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO1A
Entrez Gene ID
11151
Gene Name
coronin 1A
Gene Aliases
CLABP, CLIPINA, HCORO1, IMD8, TACO, p57
UniGene
Hs.415067
Gene Chromosome Location
Chr.16: 30183410-30189076 on Build GRCh38
Transcripts
NM_001193333.2 NM_007074.3 XM_011545714.2 XM_017022885.1 XM_017022886.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4674223 c.844G>A p.V282I Chr.16: 30187812-30187812 on Build GRCh38
4674222 c.842T>C p.I281T Chr.16: 30187810-30187810 on Build GRCh38
3948461 c.862-10C>T p.? Chr.16: 30187932-30187932 on Build GRCh38
6634192 c.904C>A p.P302T Chr.16: 30187984-30187984 on Build GRCh38
3356822 c.987G>T p.V329V Chr.16: 30188067-30188067 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775164094 Chr.16: 30187879-30187879 on Build GRCh38
rs529237177 Chr.16: 30187839-30187839 on Build GRCh38
rs753504025 Chr.16: 30187837-30187837 on Build GRCh38
rs779111977 Chr.16: 30188100-30188101 on Build GRCh38
rs752456221 Chr.16: 30187667-30187667 on Build GRCh38
rs951099808 Chr.16: 30187799-30187799 on Build GRCh38
rs777900392 Chr.16: 30187852-30187852 on Build GRCh38
rs767960904 Chr.16: 30187908-30187908 on Build GRCh38
rs775074204 Chr.16: 30187808-30187808 on Build GRCh38
rs748379111 Chr.16: 30187733-30187733 on Build GRCh38
rs1046484053 Chr.16: 30187707-30187707 on Build GRCh38
rs534787658 Chr.16: 30187915-30187918 on Build GRCh38
rs113875403 Chr.16: 30187805-30187805 on Build GRCh38
rs201641696 Chr.16: 30187894-30187894 on Build GRCh38
rs763606361 Chr.16: 30187812-30187812 on Build GRCh38
rs945378319 Chr.16: 30187980-30187980 on Build GRCh38
rs751460210 Chr.16: 30187855-30187855 on Build GRCh38
rs754748246 Chr.16: 30187855-30187856 on Build GRCh38
rs367708753 Chr.16: 30187715-30187715 on Build GRCh38
rs551863566 Chr.16: 30187853-30187853 on Build GRCh38
rs376463194 Chr.16: 30187848-30187848 on Build GRCh38
rs199960741 Chr.16: 30188076-30188076 on Build GRCh38
rs757576567 Chr.16: 30188053-30188053 on Build GRCh38
rs565893372 Chr.16: 30187844-30187844 on Build GRCh38
rs761189964 Chr.16: 30188042-30188042 on Build GRCh38
rs894345541 Chr.16: 30187748-30187748 on Build GRCh38
rs199650298 Chr.16: 30187811-30187811 on Build GRCh38
rs752677244 Chr.16: 30187845-30187845 on Build GRCh38
rs200153667 Chr.16: 30187871-30187871 on Build GRCh38
rs758965652 Chr.16: 30187691-30187691 on Build GRCh38
rs764085231 Chr.16: 30188043-30188043 on Build GRCh38
rs1026962040 Chr.16: 30188092-30188092 on Build GRCh38
rs199744765 Chr.16: 30187892-30187892 on Build GRCh38
rs373178145 Chr.16: 30187928-30187928 on Build GRCh38
rs201613058 Chr.16: 30188100-30188100 on Build GRCh38
rs766409965 Chr.16: 30187850-30187850 on Build GRCh38
rs757631851 Chr.16: 30188005-30188005 on Build GRCh38
rs761473049 Chr.16: 30187714-30187715 on Build GRCh38
rs764684366 Chr.16: 30187787-30187789 on Build GRCh38
rs906618469 Chr.16: 30188007-30188007 on Build GRCh38
rs879035805 Chr.16: 30187859-30187859 on Build GRCh38
rs1003538544 Chr.16: 30187695-30187695 on Build GRCh38
rs755665808 Chr.16: 30187675-30187675 on Build GRCh38
rs749973304 Chr.16: 30187844-30187845 on Build GRCh38
rs913867712 Chr.16: 30187974-30187974 on Build GRCh38
rs753219676 Chr.16: 30187913-30187913 on Build GRCh38
rs759940433 Chr.16: 30188018-30188018 on Build GRCh38
rs750715781 Chr.16: 30187687-30187687 on Build GRCh38
rs201442400 Chr.16: 30187696-30187696 on Build GRCh38
rs766738360 Chr.16: 30187898-30187898 on Build GRCh38
rs747483457 Chr.16: 30187701-30187701 on Build GRCh38
rs779884676 Chr.16: 30188099-30188099 on Build GRCh38
rs373553554 Chr.16: 30187904-30187904 on Build GRCh38
rs771972084 Chr.16: 30187943-30187943 on Build GRCh38
rs759161131 Chr.16: 30187644-30187644 on Build GRCh38
rs867215542 Chr.16: 30187668-30187668 on Build GRCh38
rs761372261 Chr.16: 30187997-30187997 on Build GRCh38
rs200963965 Chr.16: 30187744-30187744 on Build GRCh38
rs144860668 Chr.16: 30188025-30188025 on Build GRCh38
rs894750739 Chr.16: 30188031-30188031 on Build GRCh38
rs769837988 Chr.16: 30187735-30187735 on Build GRCh38
rs749524712 Chr.16: 30187855-30187855 on Build GRCh38
rs765640170 Chr.16: 30187685-30187685 on Build GRCh38
rs767238508 Chr.16: 30187858-30187862 on Build GRCh38
rs770912789 Chr.16: 30187856-30187856 on Build GRCh38
rs1042118963 Chr.16: 30187907-30187907 on Build GRCh38
rs200859430 Chr.16: 30187923-30187923 on Build GRCh38
rs746960406 Chr.16: 30187966-30187966 on Build GRCh38
rs762867240 Chr.16: 30187849-30187850 on Build GRCh38
rs778793192 Chr.16: 30187862-30187862 on Build GRCh38
rs776148815 Chr.16: 30187979-30187979 on Build GRCh38
rs758727900 Chr.16: 30187838-30187838 on Build GRCh38
rs745611340 Chr.16: 30187929-30187929 on Build GRCh38
rs377351892 Chr.16: 30188082-30188082 on Build GRCh38
rs372809960 Chr.16: 30187903-30187903 on Build GRCh38
rs759536346 Chr.16: 30187804-30187804 on Build GRCh38
rs745805583 Chr.16: 30187869-30187869 on Build GRCh38
rs753915570 Chr.16: 30188044-30188044 on Build GRCh38
rs754074996 Chr.16: 30187860-30187860 on Build GRCh38
rs376748014 Chr.16: 30188096-30188096 on Build GRCh38
rs1026931027 Chr.16: 30187781-30187781 on Build GRCh38
rs376912665 Chr.16: 30188034-30188034 on Build GRCh38
rs535362943 Chr.16: 30187977-30187977 on Build GRCh38
rs139024575 Chr.16: 30187760-30187760 on Build GRCh38
rs148629867 Chr.16: 30188016-30188016 on Build GRCh38
rs547256571 Chr.16: 30187840-30187840 on Build GRCh38
rs779895187 Chr.16: 30187951-30187951 on Build GRCh38
rs767866898 Chr.16: 30188029-30188029 on Build GRCh38
rs149867063 Chr.16: 30187772-30187772 on Build GRCh38
rs1002648704 Chr.16: 30187730-30187730 on Build GRCh38
rs368258518 Chr.16: 30187874-30187874 on Build GRCh38
rs187747730 Chr.16: 30187742-30187742 on Build GRCh38
rs760150292 Chr.16: 30187891-30187891 on Build GRCh38
rs368862536 Chr.16: 30187920-30187920 on Build GRCh38
rs375783340 Chr.16: 30187919-30187919 on Build GRCh38
rs767135347 Chr.16: 30187645-30187645 on Build GRCh38
rs769394096 Chr.16: 30188011-30188011 on Build GRCh38
rs879178253 Chr.16: 30187849-30187849 on Build GRCh38
rs867131789 Chr.16: 30187942-30187942 on Build GRCh38

View Full Product Details