Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 55566590-55567089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPNS2
Entrez Gene ID
84290
Gene Name
calpain small subunit 2
Gene Aliases
-
UniGene
Hs.660027
Gene Chromosome Location
Chr.16: 55566672-55567680 on Build GRCh38
Transcripts
NM_032330.1 NM_017839.4 XM_011523169.2 XM_017023376.1

Gene Symbol
LPCAT2
Entrez Gene ID
54947
Gene Name
lysophosphatidylcholine acyltransferase 2
Gene Aliases
AGPAT11, AYTL1, LysoPAFAT
UniGene
Hs.460857
Gene Chromosome Location
Chr.16: 55509001-55586670 on Build GRCh38
Transcripts
NM_032330.1 NM_017839.4 XM_011523169.2 XM_017023376.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5966198 c.16G>C p.A6P Chr.16: 55566772-55566772 on Build GRCh38
4061275 c.38G>A p.R13Q Chr.16: 55566794-55566794 on Build GRCh38
971572 c.244C>T p.R82W Chr.16: 55567000-55567000 on Build GRCh38
1727915 c.307A>T p.I103F Chr.16: 55567063-55567063 on Build GRCh38
3969593 c.114A>G p.G38G Chr.16: 55566870-55566870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753645944 Chr.16: 55566691-55566691 on Build GRCh38
rs756657253 Chr.16: 55566792-55566792 on Build GRCh38
rs775422621 Chr.16: 55566941-55566941 on Build GRCh38
rs760381122 Chr.16: 55566745-55566745 on Build GRCh38
rs750022525 Chr.16: 55567025-55567025 on Build GRCh38
rs552622469 Chr.16: 55566732-55566732 on Build GRCh38
rs758007513 Chr.16: 55566925-55566925 on Build GRCh38
rs750924180 Chr.16: 55566781-55566781 on Build GRCh38
rs558335848 Chr.16: 55566631-55566631 on Build GRCh38
rs530829455 Chr.16: 55566963-55566963 on Build GRCh38
rs76369384 Chr.16: 55567051-55567051 on Build GRCh38
rs529079551 Chr.16: 55567001-55567001 on Build GRCh38
rs746326321 Chr.16: 55566715-55566715 on Build GRCh38
rs750778469 Chr.16: 55566907-55566907 on Build GRCh38
rs756931412 Chr.16: 55567023-55567023 on Build GRCh38
rs757410772 Chr.16: 55567040-55567041 on Build GRCh38
rs754326715 Chr.16: 55566923-55566923 on Build GRCh38
rs763562299 Chr.16: 55567002-55567002 on Build GRCh38
rs749172950 Chr.16: 55566953-55566953 on Build GRCh38
rs768647097 Chr.16: 55566962-55566962 on Build GRCh38
rs763220275 Chr.16: 55566761-55566761 on Build GRCh38
rs781556375 Chr.16: 55566711-55566711 on Build GRCh38
rs1052466735 Chr.16: 55566700-55566700 on Build GRCh38
rs759481405 Chr.16: 55566750-55566750 on Build GRCh38
rs763026774 Chr.16: 55566850-55566850 on Build GRCh38
rs781569523 Chr.16: 55566779-55566779 on Build GRCh38
rs773195282 Chr.16: 55566738-55566738 on Build GRCh38
rs761641680 Chr.16: 55566723-55566723 on Build GRCh38
rs754428894 Chr.16: 55567000-55567000 on Build GRCh38
rs745662780 Chr.16: 55566936-55566936 on Build GRCh38
rs778611244 Chr.16: 55566794-55566794 on Build GRCh38
rs769706156 Chr.16: 55566840-55566840 on Build GRCh38
rs758187748 Chr.16: 55566813-55566813 on Build GRCh38
rs761123531 Chr.16: 55566994-55566994 on Build GRCh38
rs144503508 Chr.16: 55567053-55567053 on Build GRCh38
rs540746132 Chr.16: 55566673-55566673 on Build GRCh38
rs777300399 Chr.16: 55566926-55566926 on Build GRCh38
rs769515019 Chr.16: 55566937-55566937 on Build GRCh38
rs770666608 Chr.16: 55566817-55566817 on Build GRCh38
rs201303639 Chr.16: 55566760-55566760 on Build GRCh38
rs754784372 Chr.16: 55566905-55566929 on Build GRCh38
rs776460250 Chr.16: 55566819-55566819 on Build GRCh38
rs369944110 Chr.16: 55566739-55566739 on Build GRCh38
rs751067349 Chr.16: 55567004-55567004 on Build GRCh38
rs372309142 Chr.16: 55566914-55566914 on Build GRCh38
rs541410526 Chr.16: 55566821-55566821 on Build GRCh38
rs371505336 Chr.16: 55566940-55566940 on Build GRCh38
rs766790678 Chr.16: 55566995-55566995 on Build GRCh38
rs751423765 Chr.16: 55566809-55566819 on Build GRCh38
rs749260862 Chr.16: 55567042-55567042 on Build GRCh38
rs777293329 Chr.16: 55566814-55566814 on Build GRCh38
rs996061704 Chr.16: 55566879-55566879 on Build GRCh38
rs568915574 Chr.16: 55567045-55567045 on Build GRCh38
rs767765531 Chr.16: 55566987-55566987 on Build GRCh38
rs770704096 Chr.16: 55566748-55566748 on Build GRCh38
rs373677019 Chr.16: 55566759-55566759 on Build GRCh38
rs764296861 Chr.16: 55566768-55566768 on Build GRCh38
rs563285482 Chr.16: 55566934-55566934 on Build GRCh38
rs751857905 Chr.16: 55566710-55566710 on Build GRCh38
rs545731270 Chr.16: 55567033-55567033 on Build GRCh38
rs774457292 Chr.16: 55566871-55566871 on Build GRCh38
rs755928781 Chr.16: 55567028-55567028 on Build GRCh38
rs761987158 Chr.16: 55566972-55566972 on Build GRCh38
rs775614761 Chr.16: 55566846-55566846 on Build GRCh38
rs767810947 Chr.16: 55566906-55566906 on Build GRCh38
rs780907237 Chr.16: 55566935-55566935 on Build GRCh38
rs367628397 Chr.16: 55566939-55566939 on Build GRCh38
rs751740095 Chr.16: 55566772-55566772 on Build GRCh38
rs765344413 Chr.16: 55566757-55566757 on Build GRCh38
rs369578789 Chr.16: 55567008-55567008 on Build GRCh38
rs768612057 Chr.16: 55567048-55567048 on Build GRCh38
rs1028679923 Chr.16: 55566921-55566921 on Build GRCh38
rs778149330 Chr.16: 55566940-55566940 on Build GRCh38
rs776459253 Chr.16: 55566749-55566749 on Build GRCh38
rs867752834 Chr.16: 55566709-55566709 on Build GRCh38
rs754058757 Chr.16: 55566965-55566973 on Build GRCh38
rs762054290 Chr.16: 55566877-55566877 on Build GRCh38
rs903978134 Chr.16: 55566633-55566633 on Build GRCh38
rs778525510 Chr.16: 55566731-55566731 on Build GRCh38
rs747644217 Chr.16: 55566809-55566809 on Build GRCh38
rs764105790 Chr.16: 55566869-55566869 on Build GRCh38
rs573318769 Chr.16: 55566647-55566647 on Build GRCh38
rs757573075 Chr.16: 55566777-55566777 on Build GRCh38
rs766985836 Chr.16: 55566918-55566918 on Build GRCh38
rs371676163 Chr.16: 55566815-55566815 on Build GRCh38
rs771824489 Chr.16: 55566733-55566733 on Build GRCh38
rs992787049 Chr.16: 55566985-55566985 on Build GRCh38
rs773515095 Chr.16: 55566990-55566990 on Build GRCh38

View Full Product Details