Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57759034-57759507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIFC3
Entrez Gene ID
3801
Gene Name
kinesin family member C3
Gene Aliases
-
UniGene
Hs.23131
Gene Chromosome Location
Chr.16: 57758217-57863053 on Build GRCh38
Transcripts
NM_001130099.1 NM_001130100.1 NM_001318710.1 NM_001318711.1 NM_001318712.1 NM_001318713.1 NM_001318714.1 NM_001318715.1 NM_005550.3 NR_134678.1 XM_005255937.1 XM_005255944.1 XM_006721188.1 XM_011523075.1 XM_011523076.1 XM_011523077.1 XM_011523078.1 XM_011523079.1 XM_017023219.1 XM_017023220.1 XM_017023221.1 XM_017023222.1 XM_017023223.1 XM_017023224.1 XM_017023225.1 XM_017023226.1 XM_017023227.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6324914 c.2304C>T p.A768A Chr.16: 57759152-57759152 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1056322470 Chr.16: 57759074-57759074 on Build GRCh38
rs891236876 Chr.16: 57759374-57759374 on Build GRCh38
rs980947634 Chr.16: 57759244-57759244 on Build GRCh38
rs782350950 Chr.16: 57759347-57759347 on Build GRCh38
rs71281832 Chr.16: 57759310-57759311 on Build GRCh38
rs117994346 Chr.16: 57759081-57759081 on Build GRCh38
rs35885527 Chr.16: 57759439-57759439 on Build GRCh38
rs554058092 Chr.16: 57759147-57759147 on Build GRCh38
rs997343821 Chr.16: 57759130-57759130 on Build GRCh38
rs920262530 Chr.16: 57759192-57759192 on Build GRCh38
rs996832351 Chr.16: 57759359-57759359 on Build GRCh38
rs961660717 Chr.16: 57759173-57759173 on Build GRCh38
rs1043613267 Chr.16: 57759346-57759346 on Build GRCh38
rs1056267333 Chr.16: 57759299-57759299 on Build GRCh38
rs369249757 Chr.16: 57759312-57759312 on Build GRCh38
rs186055080 Chr.16: 57759061-57759061 on Build GRCh38
rs782789315 Chr.16: 57759451-57759451 on Build GRCh38
rs903819700 Chr.16: 57759100-57759100 on Build GRCh38
rs534195343 Chr.16: 57759161-57759161 on Build GRCh38
rs782193223 Chr.16: 57759145-57759145 on Build GRCh38
rs1015749885 Chr.16: 57759158-57759158 on Build GRCh38
rs545243418 Chr.16: 57759418-57759418 on Build GRCh38
rs961502342 Chr.16: 57759426-57759426 on Build GRCh38
rs578107203 Chr.16: 57759352-57759352 on Build GRCh38
rs1027214457 Chr.16: 57759477-57759477 on Build GRCh38
rs1015802319 Chr.16: 57759411-57759411 on Build GRCh38
rs538586803 Chr.16: 57759094-57759094 on Build GRCh38
rs1008311669 Chr.16: 57759385-57759385 on Build GRCh38
rs145568783 Chr.16: 57759484-57759484 on Build GRCh38
rs1028731498 Chr.16: 57759363-57759363 on Build GRCh38
rs563173403 Chr.16: 57759479-57759479 on Build GRCh38
rs892292942 Chr.16: 57759301-57759301 on Build GRCh38
rs782170928 Chr.16: 57759268-57759268 on Build GRCh38
rs782596635 Chr.16: 57759119-57759119 on Build GRCh38
rs71387193 Chr.16: 57759234-57759234 on Build GRCh38
rs148205466 Chr.16: 57759067-57759067 on Build GRCh38
rs190752355 Chr.16: 57759148-57759148 on Build GRCh38
rs781900784 Chr.16: 57759140-57759140 on Build GRCh38
rs1009437710 Chr.16: 57759092-57759092 on Build GRCh38
rs926363519 Chr.16: 57759248-57759248 on Build GRCh38
rs141124624 Chr.16: 57759174-57759174 on Build GRCh38
rs951814320 Chr.16: 57759478-57759478 on Build GRCh38
rs1022752877 Chr.16: 57759093-57759093 on Build GRCh38
rs548533967 Chr.16: 57759060-57759060 on Build GRCh38
rs544708292 Chr.16: 57759311-57759311 on Build GRCh38
rs926289328 Chr.16: 57759485-57759485 on Build GRCh38
rs549608225 Chr.16: 57759080-57759080 on Build GRCh38

View Full Product Details