Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 58283618-58284128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC113
Entrez Gene ID
29070
Gene Name
coiled-coil domain containing 113
Gene Aliases
HSPC065
UniGene
Hs.11614
Gene Chromosome Location
Chr.16: 58196863-58283836 on Build GRCh38
Transcripts
NM_001142302.1 NM_014157.3 XM_011523038.2 XM_011523039.2 XM_011523040.2 XM_011523041.2 XM_017023176.1 XM_017023177.1 NM_001080492.1 NM_001305173.1 NM_001305174.1

Gene Symbol
PRSS54
Entrez Gene ID
221191
Gene Name
protease, serine 54
Gene Aliases
CT67, KLKBL4
UniGene
Hs.411239
Gene Chromosome Location
Chr.16: 58279997-58295047 on Build GRCh38
Transcripts
NM_001142302.1 NM_014157.3 XM_011523038.2 XM_011523039.2 XM_011523040.2 XM_011523041.2 XM_017023176.1 XM_017023177.1 NM_001080492.1 NM_001305173.1 NM_001305174.1

SNP Detail

SNP ID Chromosome Location
rs1029651535 Chr.16: 58283653-58283654 on Build GRCh38
rs902367941 Chr.16: 58283900-58283901 on Build GRCh38
rs185527082 Chr.16: 58283945-58283945 on Build GRCh38
rs1054723819 Chr.16: 58283850-58283850 on Build GRCh38
rs770095199 Chr.16: 58283897-58283897 on Build GRCh38
rs985470558 Chr.16: 58283708-58283708 on Build GRCh38
rs530048458 Chr.16: 58283683-58283683 on Build GRCh38
rs568103701 Chr.16: 58283909-58283909 on Build GRCh38
rs16960042 Chr.16: 58283857-58283857 on Build GRCh38
rs370814362 Chr.16: 58283828-58283828 on Build GRCh38
rs900022163 Chr.16: 58283896-58283896 on Build GRCh38
rs954004598 Chr.16: 58283677-58283677 on Build GRCh38
rs887071970 Chr.16: 58283912-58283912 on Build GRCh38
rs962778490 Chr.16: 58283946-58283946 on Build GRCh38
rs935007345 Chr.16: 58283816-58283816 on Build GRCh38
rs910089450 Chr.16: 58283719-58283719 on Build GRCh38
rs1040263434 Chr.16: 58283895-58283895 on Build GRCh38
rs139892519 Chr.16: 58283915-58283915 on Build GRCh38
rs775822567 Chr.16: 58284079-58284079 on Build GRCh38
rs1006854729 Chr.16: 58283997-58283997 on Build GRCh38
rs1016975240 Chr.16: 58284024-58284024 on Build GRCh38
rs927593990 Chr.16: 58283767-58283767 on Build GRCh38
rs893555084 Chr.16: 58283829-58283829 on Build GRCh38
rs963281231 Chr.16: 58284038-58284038 on Build GRCh38
rs370137789 Chr.16: 58283656-58283657 on Build GRCh38
rs775897480 Chr.16: 58283647-58283647 on Build GRCh38
rs1029926869 Chr.16: 58283903-58283903 on Build GRCh38
rs559364065 Chr.16: 58283651-58283651 on Build GRCh38
rs546671993 Chr.16: 58283748-58283748 on Build GRCh38
rs973035656 Chr.16: 58284068-58284068 on Build GRCh38
rs776707295 Chr.16: 58283746-58283746 on Build GRCh38
rs982071972 Chr.16: 58283752-58283752 on Build GRCh38
rs558710778 Chr.16: 58283995-58283995 on Build GRCh38
rs1029724886 Chr.16: 58283935-58283935 on Build GRCh38
rs962995769 Chr.16: 58283736-58283736 on Build GRCh38
rs1010859912 Chr.16: 58283860-58283860 on Build GRCh38
rs144058836 Chr.16: 58283709-58283709 on Build GRCh38

View Full Product Details