Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66579728-66580229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMTM2
Entrez Gene ID
146225
Gene Name
CKLF like MARVEL transmembrane domain containing 2
Gene Aliases
CKLFSF2
UniGene
Hs.195685
Gene Chromosome Location
Chr.16: 66579448-66588275 on Build GRCh38
Transcripts
NM_001199317.1 NM_144673.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4481652 c.252C>T p.L84L Chr.16: 66579859-66579859 on Build GRCh38
1709266 c.443C>T p.S148F Chr.16: 66580183-66580183 on Build GRCh38
4480934 c.246C>T p.F82F Chr.16: 66579853-66579853 on Build GRCh38
1493714 c.328G>A p.V110M Chr.16: 66580068-66580068 on Build GRCh38
4415170 c.365T>C p.I122T Chr.16: 66580105-66580105 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146637368 Chr.16: 66579864-66579864 on Build GRCh38
rs370543135 Chr.16: 66579773-66579773 on Build GRCh38
rs781633308 Chr.16: 66579798-66579798 on Build GRCh38
rs776968032 Chr.16: 66580111-66580111 on Build GRCh38
rs371708621 Chr.16: 66580062-66580062 on Build GRCh38
rs748608792 Chr.16: 66580195-66580195 on Build GRCh38
rs754210253 Chr.16: 66580030-66580030 on Build GRCh38
rs758546843 Chr.16: 66580051-66580051 on Build GRCh38
rs754776259 Chr.16: 66580179-66580179 on Build GRCh38
rs769582853 Chr.16: 66579940-66579940 on Build GRCh38
rs887206753 Chr.16: 66579868-66579868 on Build GRCh38
rs761550400 Chr.16: 66579874-66579874 on Build GRCh38
rs746230824 Chr.16: 66579832-66579833 on Build GRCh38
rs749169475 Chr.16: 66579943-66579943 on Build GRCh38
rs753650515 Chr.16: 66580168-66580168 on Build GRCh38
rs2290182 Chr.16: 66580105-66580105 on Build GRCh38
rs767042332 Chr.16: 66580003-66580003 on Build GRCh38
rs9925443 Chr.16: 66579788-66579788 on Build GRCh38
rs777736525 Chr.16: 66580060-66580060 on Build GRCh38
rs778876981 Chr.16: 66580185-66580185 on Build GRCh38
rs1002723323 Chr.16: 66580035-66580035 on Build GRCh38
rs763472578 Chr.16: 66580193-66580193 on Build GRCh38
rs771566416 Chr.16: 66580100-66580100 on Build GRCh38
rs765794143 Chr.16: 66580044-66580044 on Build GRCh38
rs748124434 Chr.16: 66580099-66580099 on Build GRCh38
rs760310900 Chr.16: 66580107-66580107 on Build GRCh38
rs757484990 Chr.16: 66580161-66580161 on Build GRCh38
rs751655164 Chr.16: 66580159-66580159 on Build GRCh38
rs201667312 Chr.16: 66579924-66579924 on Build GRCh38
rs766689376 Chr.16: 66579923-66579923 on Build GRCh38
rs943700981 Chr.16: 66580011-66580011 on Build GRCh38
rs770560267 Chr.16: 66580110-66580110 on Build GRCh38
rs368740252 Chr.16: 66580116-66580116 on Build GRCh38
rs763382465 Chr.16: 66580136-66580136 on Build GRCh38
rs769598007 Chr.16: 66579827-66579827 on Build GRCh38
rs771909732 Chr.16: 66580203-66580203 on Build GRCh38
rs375827407 Chr.16: 66580150-66580150 on Build GRCh38
rs111592429 Chr.16: 66579821-66579821 on Build GRCh38
rs772287237 Chr.16: 66579822-66579822 on Build GRCh38
rs934164394 Chr.16: 66579847-66579848 on Build GRCh38
rs768778425 Chr.16: 66579988-66579988 on Build GRCh38
rs781611810 Chr.16: 66580162-66580162 on Build GRCh38
rs760272770 Chr.16: 66580025-66580025 on Build GRCh38
rs752752134 Chr.16: 66580050-66580050 on Build GRCh38
rs778900087 Chr.16: 66579932-66579932 on Build GRCh38
rs371994585 Chr.16: 66579931-66579931 on Build GRCh38
rs151139084 Chr.16: 66579775-66579775 on Build GRCh38
rs778989765 Chr.16: 66579818-66579818 on Build GRCh38
rs756562089 Chr.16: 66579805-66579805 on Build GRCh38
rs200227017 Chr.16: 66579772-66579772 on Build GRCh38
rs761454694 Chr.16: 66580001-66580001 on Build GRCh38
rs750727871 Chr.16: 66579888-66579888 on Build GRCh38
rs757423151 Chr.16: 66580064-66580064 on Build GRCh38
rs765287841 Chr.16: 66579753-66579753 on Build GRCh38
rs1023671203 Chr.16: 66580189-66580189 on Build GRCh38
rs1013528492 Chr.16: 66580158-66580158 on Build GRCh38
rs903927625 Chr.16: 66579768-66579768 on Build GRCh38
rs771721025 Chr.16: 66579937-66579938 on Build GRCh38
rs914269289 Chr.16: 66579785-66579785 on Build GRCh38
rs772884517 Chr.16: 66580014-66580014 on Build GRCh38
rs748940720 Chr.16: 66579800-66579800 on Build GRCh38
rs375959404 Chr.16: 66579913-66579913 on Build GRCh38
rs753163111 Chr.16: 66580129-66580129 on Build GRCh38
rs112263070 Chr.16: 66579895-66579895 on Build GRCh38
rs749149673 Chr.16: 66580068-66580068 on Build GRCh38
rs577701143 Chr.16: 66579816-66579816 on Build GRCh38
rs768690571 Chr.16: 66580070-66580070 on Build GRCh38
rs752135930 Chr.16: 66579771-66579771 on Build GRCh38
rs775457714 Chr.16: 66579848-66579848 on Build GRCh38
rs371415575 Chr.16: 66579799-66579799 on Build GRCh38
rs368665656 Chr.16: 66579813-66579813 on Build GRCh38
rs2290183 Chr.16: 66579938-66579938 on Build GRCh38
rs1013433548 Chr.16: 66579801-66579801 on Build GRCh38
rs750115610 Chr.16: 66579957-66579957 on Build GRCh38
rs760255688 Chr.16: 66580185-66580188 on Build GRCh38
rs367871753 Chr.16: 66580027-66580027 on Build GRCh38
rs767256157 Chr.16: 66579887-66579887 on Build GRCh38
rs751207201 Chr.16: 66579779-66579779 on Build GRCh38
rs964418824 Chr.16: 66580096-66580096 on Build GRCh38
rs141329430 Chr.16: 66580086-66580086 on Build GRCh38
rs775159571 Chr.16: 66579941-66579941 on Build GRCh38
rs2290181 Chr.16: 66580194-66580194 on Build GRCh38
rs751031016 Chr.16: 66579911-66579911 on Build GRCh38
rs774757992 Chr.16: 66579809-66579809 on Build GRCh38
rs1030920227 Chr.16: 66579787-66579787 on Build GRCh38
rs759434314 Chr.16: 66579825-66579825 on Build GRCh38
rs908738542 Chr.16: 66580185-66580186 on Build GRCh38
rs755473676 Chr.16: 66580036-66580036 on Build GRCh38
rs774948803 Chr.16: 66580008-66580009 on Build GRCh38
rs1044280613 Chr.16: 66580074-66580074 on Build GRCh38
rs751117468 Chr.16: 66579869-66579869 on Build GRCh38
rs879123868 Chr.16: 66580012-66580012 on Build GRCh38
rs376462155 Chr.16: 66580114-66580114 on Build GRCh38
rs773982696 Chr.16: 66579998-66579998 on Build GRCh38
rs756538946 Chr.16: 66579919-66579919 on Build GRCh38
rs779892904 Chr.16: 66580067-66580067 on Build GRCh38
rs771408245 Chr.16: 66579784-66579794 on Build GRCh38

View Full Product Details