Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66935649-66936176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CES2
Entrez Gene ID
8824
Gene Name
carboxylesterase 2
Gene Aliases
CE-2, CES2A1, PCE-2, iCE
UniGene
Hs.282975
Gene Chromosome Location
Chr.16: 66934444-66945096 on Build GRCh38
Transcripts
NM_003869.5 NM_198061.2 NR_036684.1 XM_011523421.2 XM_017023818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4671805 c.260delG p.G88fs*25 Chr.16: 66935703-66935703 on Build GRCh38
7123376 c.253C>G p.L85V Chr.16: 66935696-66935696 on Build GRCh38
4579149 c.252T>G p.L84L Chr.16: 66935695-66935695 on Build GRCh38
972252 c.228G>A p.A76A Chr.16: 66935671-66935671 on Build GRCh38
6627396 c.265C>T p.Q89* Chr.16: 66935708-66935708 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs541665662 Chr.16: 66935981-66935981 on Build GRCh38
rs769055756 Chr.16: 66935788-66935788 on Build GRCh38
rs574995668 Chr.16: 66935735-66935735 on Build GRCh38
rs113299322 Chr.16: 66935975-66935975 on Build GRCh38
rs746811378 Chr.16: 66935754-66935754 on Build GRCh38
rs200867094 Chr.16: 66935691-66935691 on Build GRCh38
rs945117078 Chr.16: 66936023-66936023 on Build GRCh38
rs561884800 Chr.16: 66935988-66935988 on Build GRCh38
rs1029165841 Chr.16: 66935910-66935910 on Build GRCh38
rs913726937 Chr.16: 66936140-66936140 on Build GRCh38
rs772469119 Chr.16: 66936042-66936042 on Build GRCh38
rs546208879 Chr.16: 66935874-66935874 on Build GRCh38
rs112435663 Chr.16: 66935925-66935925 on Build GRCh38
rs560097079 Chr.16: 66935920-66935920 on Build GRCh38
rs757128362 Chr.16: 66935757-66935757 on Build GRCh38
rs202017139 Chr.16: 66935709-66935709 on Build GRCh38
rs952039750 Chr.16: 66936106-66936106 on Build GRCh38
rs778766860 Chr.16: 66935821-66935823 on Build GRCh38
rs375428335 Chr.16: 66935746-66935746 on Build GRCh38
rs561593977 Chr.16: 66935785-66935785 on Build GRCh38
rs1000406021 Chr.16: 66935881-66935882 on Build GRCh38
rs777745155 Chr.16: 66935684-66935684 on Build GRCh38
rs927112487 Chr.16: 66935762-66935762 on Build GRCh38
rs751306096 Chr.16: 66935837-66935837 on Build GRCh38
rs111299112 Chr.16: 66936040-66936040 on Build GRCh38
rs1027708917 Chr.16: 66936103-66936103 on Build GRCh38
rs575360275 Chr.16: 66935832-66935832 on Build GRCh38
rs11863141 Chr.16: 66935689-66935689 on Build GRCh38
rs960600098 Chr.16: 66936138-66936138 on Build GRCh38
rs979602655 Chr.16: 66936154-66936154 on Build GRCh38
rs774982107 Chr.16: 66935706-66935706 on Build GRCh38
rs762294467 Chr.16: 66935794-66935794 on Build GRCh38
rs577343202 Chr.16: 66935868-66935868 on Build GRCh38
rs375639497 Chr.16: 66935670-66935670 on Build GRCh38
rs750406965 Chr.16: 66935849-66935849 on Build GRCh38
rs781129066 Chr.16: 66935761-66935761 on Build GRCh38
rs1019049802 Chr.16: 66935932-66935932 on Build GRCh38
rs932289726 Chr.16: 66936054-66936054 on Build GRCh38
rs763343151 Chr.16: 66935698-66935698 on Build GRCh38
rs747937135 Chr.16: 66935677-66935677 on Build GRCh38
rs917605433 Chr.16: 66935957-66935971 on Build GRCh38
rs759974133 Chr.16: 66935865-66935865 on Build GRCh38
rs945195326 Chr.16: 66936143-66936143 on Build GRCh38
rs764412510 Chr.16: 66935703-66935703 on Build GRCh38
rs555099248 Chr.16: 66935731-66935731 on Build GRCh38
rs781075718 Chr.16: 66935842-66935842 on Build GRCh38
rs755634639 Chr.16: 66935851-66935851 on Build GRCh38
rs774786032 Chr.16: 66935793-66935793 on Build GRCh38
rs575430709 Chr.16: 66936059-66936059 on Build GRCh38
rs553039710 Chr.16: 66935914-66935914 on Build GRCh38
rs546575549 Chr.16: 66936073-66936073 on Build GRCh38
rs544349251 Chr.16: 66935921-66935921 on Build GRCh38
rs564445583 Chr.16: 66935924-66935924 on Build GRCh38
rs775034770 Chr.16: 66935696-66935696 on Build GRCh38
rs532806533 Chr.16: 66936068-66936068 on Build GRCh38
rs766429513 Chr.16: 66935831-66935831 on Build GRCh38
rs751539813 Chr.16: 66936102-66936102 on Build GRCh38
rs926447283 Chr.16: 66935845-66935845 on Build GRCh38
rs773898941 Chr.16: 66935811-66935811 on Build GRCh38
rs762472447 Chr.16: 66935712-66935712 on Build GRCh38
rs375480722 Chr.16: 66935732-66935732 on Build GRCh38
rs758169695 Chr.16: 66935679-66935679 on Build GRCh38
rs747007078 Chr.16: 66935686-66935686 on Build GRCh38
rs563825352 Chr.16: 66936060-66936060 on Build GRCh38
rs766595893 Chr.16: 66935733-66935733 on Build GRCh38
rs779492419 Chr.16: 66935857-66935857 on Build GRCh38
rs951989104 Chr.16: 66935959-66935959 on Build GRCh38
rs757073211 Chr.16: 66935841-66935841 on Build GRCh38
rs112096129 Chr.16: 66936137-66936137 on Build GRCh38
rs566356963 Chr.16: 66935669-66935669 on Build GRCh38
rs760788132 Chr.16: 66935812-66935812 on Build GRCh38
rs953506729 Chr.16: 66935726-66935726 on Build GRCh38
rs980692902 Chr.16: 66935965-66935965 on Build GRCh38
rs753284380 Chr.16: 66935683-66935691 on Build GRCh38
rs1029113648 Chr.16: 66935724-66935724 on Build GRCh38
rs765508888 Chr.16: 66935833-66935833 on Build GRCh38
rs992044979 Chr.16: 66936139-66936139 on Build GRCh38
rs141056810 Chr.16: 66935671-66935671 on Build GRCh38
rs770423168 Chr.16: 66935688-66935688 on Build GRCh38
rs913684940 Chr.16: 66936016-66936016 on Build GRCh38
rs1043443293 Chr.16: 66936027-66936027 on Build GRCh38
rs1033620978 Chr.16: 66936134-66936134 on Build GRCh38
rs573046366 Chr.16: 66935976-66935976 on Build GRCh38
rs745340469 Chr.16: 66935767-66935767 on Build GRCh38
rs903535850 Chr.16: 66936043-66936043 on Build GRCh38
rs772671317 Chr.16: 66935799-66935799 on Build GRCh38
rs183709379 Chr.16: 66936111-66936111 on Build GRCh38
rs775183302 Chr.16: 66935784-66935784 on Build GRCh38
rs769437765 Chr.16: 66935695-66935695 on Build GRCh38
rs757366476 Chr.16: 66936121-66936121 on Build GRCh38
rs767760548 Chr.16: 66935668-66935668 on Build GRCh38
rs368771436 Chr.16: 66935742-66935742 on Build GRCh38
rs370203590 Chr.16: 66935723-66935723 on Build GRCh38
rs757225384 Chr.16: 66935802-66935803 on Build GRCh38
rs962427073 Chr.16: 66935943-66935943 on Build GRCh38
rs769225861 Chr.16: 66935777-66935777 on Build GRCh38
rs972126713 Chr.16: 66935951-66935951 on Build GRCh38

View Full Product Details