Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84422783-84423323 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2C2
Entrez Gene ID
9914
Gene Name
ATPase secretory pathway Ca2+ transporting 2
Gene Aliases
SPCA2
UniGene
Hs.6168
Gene Chromosome Location
Chr.16: 84368523-84465777 on Build GRCh38
Transcripts
NM_001286527.2 NM_001291454.1 NM_014861.3 XM_011523486.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6607881 c.857_858insT p.L287fs*93 Chr.16: 84423201-84423202 on Build GRCh38
2694484 c.898C>G p.L300V Chr.16: 84423242-84423242 on Build GRCh38
6002080 c.919G>A p.G307S Chr.16: 84423263-84423263 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs200133776 Chr.16: 84423257-84423257 on Build GRCh38
rs961365977 Chr.16: 84422819-84422819 on Build GRCh38
rs753817918 Chr.16: 84423235-84423235 on Build GRCh38
rs199706243 Chr.16: 84423186-84423186 on Build GRCh38
rs141220263 Chr.16: 84422966-84422966 on Build GRCh38
rs752047785 Chr.16: 84423149-84423149 on Build GRCh38
rs752538856 Chr.16: 84423244-84423244 on Build GRCh38
rs774803166 Chr.16: 84423207-84423207 on Build GRCh38
rs745608003 Chr.16: 84423182-84423182 on Build GRCh38
rs934415658 Chr.16: 84422904-84422904 on Build GRCh38
rs867892114 Chr.16: 84422975-84422975 on Build GRCh38
rs758169029 Chr.16: 84423057-84423057 on Build GRCh38
rs1050457706 Chr.16: 84422969-84422969 on Build GRCh38
rs370676847 Chr.16: 84423272-84423272 on Build GRCh38
rs757789081 Chr.16: 84422855-84422855 on Build GRCh38
rs376042309 Chr.16: 84423161-84423161 on Build GRCh38
rs56138311 Chr.16: 84422847-84422847 on Build GRCh38
rs750950827 Chr.16: 84423228-84423228 on Build GRCh38
rs772170428 Chr.16: 84422859-84422859 on Build GRCh38
rs758188902 Chr.16: 84423248-84423248 on Build GRCh38
rs375177699 Chr.16: 84423276-84423276 on Build GRCh38
rs777247679 Chr.16: 84423289-84423289 on Build GRCh38
rs72806604 Chr.16: 84423299-84423299 on Build GRCh38
rs776795996 Chr.16: 84422995-84422995 on Build GRCh38
rs768891653 Chr.16: 84423247-84423249 on Build GRCh38
rs1028722302 Chr.16: 84422912-84422912 on Build GRCh38
rs1004506432 Chr.16: 84422862-84422862 on Build GRCh38
rs1014983887 Chr.16: 84422866-84422866 on Build GRCh38
rs1011354898 Chr.16: 84423077-84423077 on Build GRCh38
rs374724976 Chr.16: 84423242-84423242 on Build GRCh38
rs370341123 Chr.16: 84422844-84422844 on Build GRCh38
rs914448233 Chr.16: 84422950-84422950 on Build GRCh38
rs955900564 Chr.16: 84423274-84423274 on Build GRCh38
rs924457034 Chr.16: 84423066-84423066 on Build GRCh38
rs139304279 Chr.16: 84423067-84423068 on Build GRCh38
rs562766153 Chr.16: 84423053-84423053 on Build GRCh38
rs776344425 Chr.16: 84423138-84423138 on Build GRCh38
rs571714004 Chr.16: 84423285-84423285 on Build GRCh38
rs746424579 Chr.16: 84422905-84422908 on Build GRCh38
rs180970469 Chr.16: 84422978-84422978 on Build GRCh38
rs953110089 Chr.16: 84422922-84422922 on Build GRCh38
rs772540698 Chr.16: 84423281-84423281 on Build GRCh38
rs201235841 Chr.16: 84423220-84423220 on Build GRCh38
rs1015720458 Chr.16: 84422807-84422807 on Build GRCh38
rs185063 Chr.16: 84423099-84423099 on Build GRCh38
rs752430512 Chr.16: 84423055-84423055 on Build GRCh38
rs368568706 Chr.16: 84423279-84423279 on Build GRCh38
rs752905350 Chr.16: 84423301-84423301 on Build GRCh38
rs200531713 Chr.16: 84423194-84423194 on Build GRCh38
rs1050007297 Chr.16: 84423097-84423097 on Build GRCh38
rs868521142 Chr.16: 84422833-84422833 on Build GRCh38
rs932341296 Chr.16: 84423088-84423088 on Build GRCh38
rs527505377 Chr.16: 84423159-84423159 on Build GRCh38
rs959661518 Chr.16: 84423035-84423035 on Build GRCh38
rs541138041 Chr.16: 84423010-84423010 on Build GRCh38
rs763671229 Chr.16: 84423062-84423062 on Build GRCh38
rs763289392 Chr.16: 84423141-84423141 on Build GRCh38
rs989947903 Chr.16: 84422931-84422931 on Build GRCh38
rs754068095 Chr.16: 84423167-84423167 on Build GRCh38
rs755245896 Chr.16: 84423241-84423241 on Build GRCh38
rs908632665 Chr.16: 84423178-84423178 on Build GRCh38
rs754769417 Chr.16: 84423170-84423170 on Build GRCh38
rs67306513 Chr.16: 84422806-84422806 on Build GRCh38
rs1028671399 Chr.16: 84423059-84423059 on Build GRCh38
rs559224106 Chr.16: 84423290-84423290 on Build GRCh38
rs369118852 Chr.16: 84423136-84423136 on Build GRCh38
rs938753841 Chr.16: 84422827-84422827 on Build GRCh38
rs553849921 Chr.16: 84422870-84422870 on Build GRCh38
rs780786968 Chr.16: 84423267-84423267 on Build GRCh38
rs746590672 Chr.16: 84423183-84423183 on Build GRCh38
rs185395719 Chr.16: 84423120-84423120 on Build GRCh38
rs991453440 Chr.16: 84423001-84423001 on Build GRCh38
rs543875636 Chr.16: 84423044-84423044 on Build GRCh38
rs764500182 Chr.16: 84423145-84423145 on Build GRCh38
rs200584392 Chr.16: 84423173-84423173 on Build GRCh38
rs778685103 Chr.16: 84423170-84423170 on Build GRCh38
rs750126488 Chr.16: 84423177-84423177 on Build GRCh38
rs77672821 Chr.16: 84423021-84423021 on Build GRCh38
rs776198967 Chr.16: 84423187-84423187 on Build GRCh38
rs887304636 Chr.16: 84423030-84423030 on Build GRCh38
rs146980662 Chr.16: 84423152-84423152 on Build GRCh38
rs901439095 Chr.16: 84423051-84423051 on Build GRCh38
rs748337996 Chr.16: 84423232-84423232 on Build GRCh38
rs376861806 Chr.16: 84423197-84423197 on Build GRCh38
rs928677130 Chr.16: 84422816-84422816 on Build GRCh38
rs781190394 Chr.16: 84423114-84423114 on Build GRCh38
rs991443575 Chr.16: 84423146-84423146 on Build GRCh38
rs34915415 Chr.16: 84422833-84422834 on Build GRCh38
rs181716436 Chr.16: 84422849-84422849 on Build GRCh38
rs531523544 Chr.16: 84422845-84422845 on Build GRCh38
rs247814 Chr.16: 84423036-84423036 on Build GRCh38
rs776781110 Chr.16: 84423296-84423296 on Build GRCh38
rs750546400 Chr.16: 84423156-84423156 on Build GRCh38
rs751168529 Chr.16: 84423086-84423086 on Build GRCh38
rs397943449 Chr.16: 84422844-84422845 on Build GRCh38
rs375151758 Chr.16: 84422998-84422998 on Build GRCh38
rs549964766 Chr.16: 84422809-84422809 on Build GRCh38
rs186566012 Chr.16: 84422861-84422861 on Build GRCh38
rs538910781 Chr.16: 84423277-84423277 on Build GRCh38
rs932923262 Chr.16: 84423208-84423208 on Build GRCh38
rs901392520 Chr.16: 84423200-84423200 on Build GRCh38
rs965767725 Chr.16: 84422886-84422886 on Build GRCh38
rs770472116 Chr.16: 84423263-84423263 on Build GRCh38
rs766535136 Chr.16: 84423234-84423234 on Build GRCh38
rs1027407303 Chr.16: 84422834-84422834 on Build GRCh38
rs915964866 Chr.16: 84423016-84423016 on Build GRCh38
rs1048540126 Chr.16: 84423029-84423029 on Build GRCh38
rs577866736 Chr.16: 84422888-84422888 on Build GRCh38
rs777623630 Chr.16: 84423181-84423181 on Build GRCh38
rs372268142 Chr.16: 84423284-84423284 on Build GRCh38
rs1041269228 Chr.16: 84423046-84423046 on Build GRCh38
rs967682638 Chr.16: 84422972-84422972 on Build GRCh38
rs1050070221 Chr.16: 84423224-84423224 on Build GRCh38
rs79539860 Chr.16: 84422979-84422979 on Build GRCh38
rs1021865554 Chr.16: 84423083-84423083 on Build GRCh38
rs148216584 Chr.16: 84422962-84422962 on Build GRCh38
rs535847572 Chr.16: 84422910-84422910 on Build GRCh38
rs761158336 Chr.16: 84422940-84422940 on Build GRCh38
rs1015771345 Chr.16: 84422980-84422980 on Build GRCh38
rs772007243 Chr.16: 84423180-84423180 on Build GRCh38
rs375259761 Chr.16: 84423262-84423262 on Build GRCh38
rs990328726 Chr.16: 84423078-84423078 on Build GRCh38
rs189404488 Chr.16: 84422977-84422977 on Build GRCh38
rs371572479 Chr.16: 84423222-84423222 on Build GRCh38
rs1035575041 Chr.16: 84423102-84423102 on Build GRCh38

View Full Product Details