Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 1877121-1877619 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPA1
Entrez Gene ID
6117
Gene Name
replication protein A1
Gene Aliases
HSSB, MST075, REPA1, RF-A, RP-A, RPA70
UniGene
Hs.461925
Gene Chromosome Location
Chr.17: 1829979-1899554 on Build GRCh38
Transcripts
NM_002945.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1224170 c.589T>G p.W197G Chr.17: 1877213-1877213 on Build GRCh38
5749810 c.605G>C p.R202P Chr.17: 1877229-1877229 on Build GRCh38
1224171 c.629G>A p.R210H Chr.17: 1877253-1877253 on Build GRCh38
4779747 c.659A>T p.K220M Chr.17: 1877283-1877283 on Build GRCh38
5364047 c.685G>A p.E229K Chr.17: 1877309-1877309 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759142510 Chr.17: 1877228-1877228 on Build GRCh38
rs968092784 Chr.17: 1877333-1877333 on Build GRCh38
rs780060924 Chr.17: 1877275-1877275 on Build GRCh38
rs991976668 Chr.17: 1877559-1877559 on Build GRCh38
rs937083504 Chr.17: 1877444-1877444 on Build GRCh38
rs41544523 Chr.17: 1877354-1877354 on Build GRCh38
rs768518718 Chr.17: 1877340-1877340 on Build GRCh38
rs543693164 Chr.17: 1877447-1877447 on Build GRCh38
rs577883096 Chr.17: 1877429-1877429 on Build GRCh38
rs17292084 Chr.17: 1877400-1877400 on Build GRCh38
rs908983987 Chr.17: 1877415-1877415 on Build GRCh38
rs373046252 Chr.17: 1877189-1877189 on Build GRCh38
rs41558914 Chr.17: 1877425-1877425 on Build GRCh38
rs190043329 Chr.17: 1877402-1877402 on Build GRCh38
rs1051803359 Chr.17: 1877457-1877457 on Build GRCh38
rs770232450 Chr.17: 1877310-1877310 on Build GRCh38
rs570728892 Chr.17: 1877197-1877197 on Build GRCh38
rs776881865 Chr.17: 1877412-1877412 on Build GRCh38
rs536536201 Chr.17: 1877336-1877336 on Build GRCh38
rs914165085 Chr.17: 1877424-1877424 on Build GRCh38
rs941755265 Chr.17: 1877422-1877422 on Build GRCh38
rs772983955 Chr.17: 1877364-1877364 on Build GRCh38
rs968143255 Chr.17: 1877474-1877474 on Build GRCh38
rs369200380 Chr.17: 1877185-1877185 on Build GRCh38
rs761567513 Chr.17: 1877253-1877253 on Build GRCh38
rs879082244 Chr.17: 1877445-1877445 on Build GRCh38
rs1029882563 Chr.17: 1877265-1877265 on Build GRCh38
rs562191056 Chr.17: 1877468-1877468 on Build GRCh38
rs376851045 Chr.17: 1877308-1877308 on Build GRCh38
rs956763526 Chr.17: 1877548-1877548 on Build GRCh38
rs765477442 Chr.17: 1877257-1877257 on Build GRCh38
rs748498375 Chr.17: 1877203-1877203 on Build GRCh38
rs111514401 Chr.17: 1877198-1877198 on Build GRCh38
rs770743630 Chr.17: 1877212-1877212 on Build GRCh38
rs949779309 Chr.17: 1877538-1877538 on Build GRCh38
rs780359272 Chr.17: 1877220-1877220 on Build GRCh38
rs897325561 Chr.17: 1877430-1877430 on Build GRCh38
rs374637130 Chr.17: 1877359-1877359 on Build GRCh38
rs1043884344 Chr.17: 1877204-1877204 on Build GRCh38
rs56133432 Chr.17: 1877469-1877470 on Build GRCh38
rs994309675 Chr.17: 1877441-1877441 on Build GRCh38
rs180732111 Chr.17: 1877202-1877202 on Build GRCh38
rs750543655 Chr.17: 1877182-1877182 on Build GRCh38
rs41543120 Chr.17: 1877333-1877333 on Build GRCh38
rs767011932 Chr.17: 1877320-1877323 on Build GRCh38
rs758506055 Chr.17: 1877271-1877271 on Build GRCh38
rs778020667 Chr.17: 1877588-1877588 on Build GRCh38
rs1031045926 Chr.17: 1877520-1877520 on Build GRCh38
rs996852025 Chr.17: 1877381-1877381 on Build GRCh38
rs745448338 Chr.17: 1877329-1877329 on Build GRCh38
rs746510180 Chr.17: 1877338-1877338 on Build GRCh38
rs764083881 Chr.17: 1877240-1877240 on Build GRCh38
rs535175685 Chr.17: 1877397-1877397 on Build GRCh38
rs760596122 Chr.17: 1877234-1877234 on Build GRCh38
rs964098643 Chr.17: 1877373-1877373 on Build GRCh38
rs879179755 Chr.17: 1877443-1877443 on Build GRCh38
rs747548683 Chr.17: 1877252-1877252 on Build GRCh38
rs779086063 Chr.17: 1877162-1877162 on Build GRCh38
rs770101441 Chr.17: 1877581-1877581 on Build GRCh38
rs1018213128 Chr.17: 1877469-1877469 on Build GRCh38
rs1018328012 Chr.17: 1877318-1877318 on Build GRCh38
rs774104695 Chr.17: 1877219-1877219 on Build GRCh38
rs980029752 Chr.17: 1877440-1877440 on Build GRCh38
rs778723583 Chr.17: 1877328-1877328 on Build GRCh38
rs201396404 Chr.17: 1877355-1877355 on Build GRCh38
rs750584253 Chr.17: 1877270-1877270 on Build GRCh38
rs947713491 Chr.17: 1877398-1877398 on Build GRCh38
rs751641978 Chr.17: 1877278-1877278 on Build GRCh38
rs923767844 Chr.17: 1877339-1877339 on Build GRCh38
rs570041689 Chr.17: 1877304-1877304 on Build GRCh38
rs1055588954 Chr.17: 1877497-1877497 on Build GRCh38
rs1005751298 Chr.17: 1877147-1877147 on Build GRCh38
rs1016743456 Chr.17: 1877366-1877366 on Build GRCh38
rs917000604 Chr.17: 1877507-1877507 on Build GRCh38
rs796445126 Chr.17: 1877475-1877475 on Build GRCh38
rs755486139 Chr.17: 1877290-1877290 on Build GRCh38
rs1047463290 Chr.17: 1877343-1877343 on Build GRCh38
rs776513896 Chr.17: 1877342-1877342 on Build GRCh38
rs879360378 Chr.17: 1877358-1877358 on Build GRCh38
rs201881937 Chr.17: 1877229-1877229 on Build GRCh38
rs370128129 Chr.17: 1877467-1877467 on Build GRCh38
rs1005827499 Chr.17: 1877360-1877360 on Build GRCh38

View Full Product Details