Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4307015-4307562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBE2G1
Entrez Gene ID
7326
Gene Name
ubiquitin conjugating enzyme E2 G1
Gene Aliases
E217K, UBC7, UBE2G
UniGene
Hs.741319
Gene Chromosome Location
Chr.17: 4269217-4366674 on Build GRCh38
Transcripts
NM_003342.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5386448 c.75T>G p.F25L Chr.17: 4307095-4307095 on Build GRCh38
5942752 c.125T>A p.I42N Chr.17: 4307045-4307045 on Build GRCh38
979840 c.110G>A p.R37Q Chr.17: 4307060-4307060 on Build GRCh38
3717401 c.93T>A p.D31E Chr.17: 4307077-4307077 on Build GRCh38
979839 c.145C>A p.L49I Chr.17: 4307025-4307025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952371238 Chr.17: 4307473-4307473 on Build GRCh38
rs529349391 Chr.17: 4307398-4307398 on Build GRCh38
rs974912516 Chr.17: 4307457-4307457 on Build GRCh38
rs1002981800 Chr.17: 4307214-4307214 on Build GRCh38
rs995127697 Chr.17: 4307466-4307466 on Build GRCh38
rs948890942 Chr.17: 4307419-4307419 on Build GRCh38
rs1026899690 Chr.17: 4307468-4307468 on Build GRCh38
rs758488536 Chr.17: 4307201-4307201 on Build GRCh38
rs542670223 Chr.17: 4307431-4307431 on Build GRCh38
rs1048095712 Chr.17: 4307202-4307202 on Build GRCh38
rs978808505 Chr.17: 4307067-4307067 on Build GRCh38
rs1054720135 Chr.17: 4307384-4307384 on Build GRCh38
rs775975578 Chr.17: 4307128-4307128 on Build GRCh38
rs995441337 Chr.17: 4307283-4307283 on Build GRCh38
rs528449461 Chr.17: 4307474-4307474 on Build GRCh38
rs780297703 Chr.17: 4307224-4307224 on Build GRCh38
rs956183770 Chr.17: 4307095-4307095 on Build GRCh38
rs893310987 Chr.17: 4307147-4307147 on Build GRCh38
rs904466929 Chr.17: 4307360-4307360 on Build GRCh38
rs774741645 Chr.17: 4307501-4307501 on Build GRCh38
rs907335868 Chr.17: 4307445-4307445 on Build GRCh38
rs763731364 Chr.17: 4307122-4307122 on Build GRCh38
rs1056952597 Chr.17: 4307268-4307268 on Build GRCh38
rs985255754 Chr.17: 4307305-4307305 on Build GRCh38
rs781117825 Chr.17: 4307047-4307047 on Build GRCh38
rs182977693 Chr.17: 4307350-4307350 on Build GRCh38
rs924654030 Chr.17: 4307336-4307336 on Build GRCh38
rs1019416962 Chr.17: 4307523-4307523 on Build GRCh38
rs367927285 Chr.17: 4307062-4307062 on Build GRCh38
rs76073694 Chr.17: 4307385-4307385 on Build GRCh38
rs939755475 Chr.17: 4307261-4307261 on Build GRCh38
rs987522594 Chr.17: 4307440-4307440 on Build GRCh38
rs113198399 Chr.17: 4307537-4307537 on Build GRCh38
rs886858832 Chr.17: 4307217-4307217 on Build GRCh38
rs964865428 Chr.17: 4307456-4307456 on Build GRCh38
rs375277575 Chr.17: 4307386-4307386 on Build GRCh38
rs1015273082 Chr.17: 4307238-4307238 on Build GRCh38
rs1012848307 Chr.17: 4307041-4307041 on Build GRCh38
rs1023284553 Chr.17: 4307307-4307307 on Build GRCh38
rs115786458 Chr.17: 4307294-4307294 on Build GRCh38
rs201603499 Chr.17: 4307072-4307072 on Build GRCh38
rs750846978 Chr.17: 4307145-4307145 on Build GRCh38
rs755504527 Chr.17: 4307427-4307427 on Build GRCh38
rs1037151652 Chr.17: 4307458-4307458 on Build GRCh38
rs769874508 Chr.17: 4307060-4307060 on Build GRCh38
rs764745148 Chr.17: 4307142-4307142 on Build GRCh38
rs1019040540 Chr.17: 4307451-4307451 on Build GRCh38
rs571911442 Chr.17: 4307173-4307173 on Build GRCh38
rs559338189 Chr.17: 4307462-4307462 on Build GRCh38
rs1006706130 Chr.17: 4307489-4307489 on Build GRCh38
rs1002645147 Chr.17: 4307449-4307449 on Build GRCh38
rs976542287 Chr.17: 4307335-4307336 on Build GRCh38
rs558142938 Chr.17: 4307234-4307234 on Build GRCh38
rs760596506 Chr.17: 4307104-4307104 on Build GRCh38
rs912061564 Chr.17: 4307163-4307163 on Build GRCh38
rs895665845 Chr.17: 4307289-4307289 on Build GRCh38
rs866438117 Chr.17: 4307461-4307461 on Build GRCh38
rs547220830 Chr.17: 4307368-4307368 on Build GRCh38
rs541730829 Chr.17: 4307198-4307198 on Build GRCh38
rs368988618 Chr.17: 4307068-4307068 on Build GRCh38
rs966139006 Chr.17: 4307321-4307321 on Build GRCh38
rs371350329 Chr.17: 4307303-4307303 on Build GRCh38
rs750708035 Chr.17: 4307135-4307135 on Build GRCh38
rs974977832 Chr.17: 4307191-4307191 on Build GRCh38
rs943588400 Chr.17: 4307184-4307184 on Build GRCh38
rs555406620 Chr.17: 4307050-4307050 on Build GRCh38

View Full Product Details