Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7260523-7261029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN7
Entrez Gene ID
1366
Gene Name
claudin 7
Gene Aliases
CEPTRL2, CLDN-7, CPETRL2, Hs.84359, claudin-1
UniGene
Hs.513915
Gene Chromosome Location
Chr.17: 7259903-7263193 on Build GRCh38
Transcripts
NM_001185022.1 NM_001185023.1 NM_001307.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6440114 c.270_271GG>AA p.G91S Chr.17: 7260938-7260939 on Build GRCh38
3521665 c.271G>A p.G91S Chr.17: 7260938-7260938 on Build GRCh38
6348463 c.245C>A p.A82D Chr.17: 7260964-7260964 on Build GRCh38
983903 c.315G>A p.T105T Chr.17: 7260894-7260894 on Build GRCh38
5777921 c.419A>G p.Y140C Chr.17: 7260696-7260696 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373478236 Chr.17: 7260910-7260910 on Build GRCh38
rs768087014 Chr.17: 7260796-7260796 on Build GRCh38
rs754006543 Chr.17: 7260857-7260857 on Build GRCh38
rs368954496 Chr.17: 7260661-7260661 on Build GRCh38
rs547957587 Chr.17: 7260929-7260929 on Build GRCh38
rs746209260 Chr.17: 7260873-7260873 on Build GRCh38
rs369146758 Chr.17: 7260883-7260883 on Build GRCh38
rs755817615 Chr.17: 7260588-7260588 on Build GRCh38
rs867054456 Chr.17: 7260886-7260886 on Build GRCh38
rs372273470 Chr.17: 7260609-7260609 on Build GRCh38
rs145889982 Chr.17: 7260801-7260801 on Build GRCh38
rs780034815 Chr.17: 7260750-7260750 on Build GRCh38
rs776184919 Chr.17: 7260781-7260781 on Build GRCh38
rs750305093 Chr.17: 7260727-7260727 on Build GRCh38
rs753641622 Chr.17: 7260596-7260596 on Build GRCh38
rs986873517 Chr.17: 7260958-7260958 on Build GRCh38
rs769041772 Chr.17: 7260967-7260967 on Build GRCh38
rs745832802 Chr.17: 7260602-7260602 on Build GRCh38
rs756152576 Chr.17: 7260846-7260846 on Build GRCh38
rs772811592 Chr.17: 7260992-7260992 on Build GRCh38
rs751474604 Chr.17: 7260819-7260819 on Build GRCh38
rs533639505 Chr.17: 7260990-7260990 on Build GRCh38
rs767141439 Chr.17: 7260582-7260582 on Build GRCh38
rs986821161 Chr.17: 7260699-7260699 on Build GRCh38
rs144832882 Chr.17: 7260912-7260912 on Build GRCh38
rs201396194 Chr.17: 7260692-7260692 on Build GRCh38
rs777639047 Chr.17: 7260549-7260549 on Build GRCh38
rs908450344 Chr.17: 7260925-7260925 on Build GRCh38
rs752341778 Chr.17: 7260584-7260584 on Build GRCh38
rs999467896 Chr.17: 7260772-7260772 on Build GRCh38
rs148970901 Chr.17: 7260642-7260642 on Build GRCh38
rs749475956 Chr.17: 7260764-7260764 on Build GRCh38
rs963756863 Chr.17: 7260824-7260824 on Build GRCh38
rs547019974 Chr.17: 7260570-7260570 on Build GRCh38
rs769406740 Chr.17: 7260894-7260894 on Build GRCh38
rs746299150 Chr.17: 7260780-7260780 on Build GRCh38
rs778015963 Chr.17: 7260759-7260759 on Build GRCh38
rs767306728 Chr.17: 7260594-7260594 on Build GRCh38
rs756198131 Chr.17: 7260756-7260756 on Build GRCh38
rs17849410 Chr.17: 7260718-7260718 on Build GRCh38
rs149308129 Chr.17: 7260947-7260947 on Build GRCh38
rs377295133 Chr.17: 7260664-7260664 on Build GRCh38
rs754608450 Chr.17: 7260581-7260581 on Build GRCh38
rs948772806 Chr.17: 7261003-7261003 on Build GRCh38
rs777744847 Chr.17: 7260990-7260995 on Build GRCh38
rs370514369 Chr.17: 7260577-7260577 on Build GRCh38
rs772901497 Chr.17: 7260904-7260904 on Build GRCh38
rs755530318 Chr.17: 7260581-7260583 on Build GRCh38
rs748196864 Chr.17: 7260754-7260754 on Build GRCh38
rs760178187 Chr.17: 7260792-7260792 on Build GRCh38
rs767332527 Chr.17: 7260998-7260998 on Build GRCh38
rs768742180 Chr.17: 7260553-7260553 on Build GRCh38
rs374338331 Chr.17: 7260566-7260566 on Build GRCh38
rs1019664423 Chr.17: 7260981-7260981 on Build GRCh38
rs560743926 Chr.17: 7260948-7260948 on Build GRCh38
rs777728095 Chr.17: 7260854-7260854 on Build GRCh38
rs142682069 Chr.17: 7260887-7260887 on Build GRCh38
rs760131614 Chr.17: 7260675-7260675 on Build GRCh38
rs774011626 Chr.17: 7260994-7260994 on Build GRCh38
rs755628370 Chr.17: 7260818-7260818 on Build GRCh38
rs778759624 Chr.17: 7260551-7260551 on Build GRCh38
rs758959441 Chr.17: 7260914-7260914 on Build GRCh38
rs752095132 Chr.17: 7260601-7260601 on Build GRCh38
rs200397491 Chr.17: 7260564-7260564 on Build GRCh38
rs781781930 Chr.17: 7260636-7260639 on Build GRCh38
rs750701862 Chr.17: 7260941-7260941 on Build GRCh38
rs140980249 Chr.17: 7260712-7260712 on Build GRCh38
rs143759813 Chr.17: 7260647-7260647 on Build GRCh38
rs749098075 Chr.17: 7260550-7260550 on Build GRCh38
rs369509007 Chr.17: 7260791-7260791 on Build GRCh38
rs763776370 Chr.17: 7260707-7260707 on Build GRCh38
rs761667333 Chr.17: 7260736-7260739 on Build GRCh38
rs747479920 Chr.17: 7260882-7260882 on Build GRCh38
rs367911021 Chr.17: 7260571-7260571 on Build GRCh38
rs151076113 Chr.17: 7260677-7260677 on Build GRCh38
rs774841188 Chr.17: 7260773-7260773 on Build GRCh38
rs771268160 Chr.17: 7260766-7260766 on Build GRCh38
rs568192125 Chr.17: 7260945-7260945 on Build GRCh38
rs954296922 Chr.17: 7260953-7260953 on Build GRCh38
rs1004744599 Chr.17: 7260628-7260628 on Build GRCh38
rs750216913 Chr.17: 7260805-7260805 on Build GRCh38
rs781744000 Chr.17: 7260968-7260968 on Build GRCh38
rs754980773 Chr.17: 7260620-7260620 on Build GRCh38
rs770049907 Chr.17: 7260634-7260634 on Build GRCh38
rs945643569 Chr.17: 7260758-7260758 on Build GRCh38
rs760140090 Chr.17: 7260693-7260693 on Build GRCh38
rs878992849 Chr.17: 7260987-7260987 on Build GRCh38
rs111255205 Chr.17: 7260993-7260993 on Build GRCh38
rs769316794 Chr.17: 7260991-7260991 on Build GRCh38
rs779314588 Chr.17: 7260862-7260862 on Build GRCh38
rs576767384 Chr.17: 7260995-7260995 on Build GRCh38
rs762687113 Chr.17: 7260909-7260909 on Build GRCh38
rs778857857 Chr.17: 7260599-7260599 on Build GRCh38
rs1022174664 Chr.17: 7260748-7260748 on Build GRCh38
rs929702652 Chr.17: 7260929-7260943 on Build GRCh38
rs761455420 Chr.17: 7260892-7260892 on Build GRCh38
rs568777982 Chr.17: 7260878-7260878 on Build GRCh38
rs781260339 Chr.17: 7260631-7260631 on Build GRCh38
rs1057085097 Chr.17: 7260684-7260684 on Build GRCh38
rs756267361 Chr.17: 7260771-7260772 on Build GRCh38
rs757414858 Chr.17: 7260859-7260859 on Build GRCh38
rs762778751 Chr.17: 7260808-7260808 on Build GRCh38
rs758773722 Chr.17: 7260944-7260944 on Build GRCh38
rs921359121 Chr.17: 7260656-7260656 on Build GRCh38
rs761388518 Chr.17: 7260800-7260800 on Build GRCh38
rs779224586 Chr.17: 7260933-7260933 on Build GRCh38
rs757459277 Chr.17: 7260926-7260926 on Build GRCh38
rs774931052 Chr.17: 7260669-7260669 on Build GRCh38
rs374786723 Chr.17: 7260580-7260580 on Build GRCh38
rs758320145 Chr.17: 7260734-7260734 on Build GRCh38
rs749461011 Chr.17: 7261007-7261008 on Build GRCh38
rs145020937 Chr.17: 7260719-7260719 on Build GRCh38
rs776791392 Chr.17: 7260563-7260563 on Build GRCh38
rs757054554 Chr.17: 7260598-7260598 on Build GRCh38
rs765023474 Chr.17: 7260725-7260725 on Build GRCh38
rs201481204 Chr.17: 7260798-7260798 on Build GRCh38

View Full Product Details