Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 12717211-12717728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYOCD
Entrez Gene ID
93649
Gene Name
myocardin
Gene Aliases
MYCD
UniGene
Hs.462257
Gene Chromosome Location
Chr.17: 12665890-12768949 on Build GRCh38
Transcripts
NM_001146312.2 NM_153604.3 XM_005256863.1 XM_005256864.1 XM_017025342.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3969819 c.196C>T p.R66C Chr.17: 12717364-12717364 on Build GRCh38
337462 c.243C>T p.H81H Chr.17: 12717411-12717411 on Build GRCh38
5588389 c.219C>A p.N73K Chr.17: 12717387-12717387 on Build GRCh38
1686249 c.188C>T p.S63F Chr.17: 12717356-12717356 on Build GRCh38
1686248 c.188C>T p.S63F Chr.17: 12717356-12717356 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560474889 Chr.17: 12717533-12717533 on Build GRCh38
rs754668002 Chr.17: 12717445-12717445 on Build GRCh38
rs112304255 Chr.17: 12717553-12717553 on Build GRCh38
rs35598669 Chr.17: 12717374-12717375 on Build GRCh38
rs865842559 Chr.17: 12717285-12717285 on Build GRCh38
rs1017466710 Chr.17: 12717532-12717532 on Build GRCh38
rs376571806 Chr.17: 12717427-12717427 on Build GRCh38
rs570974150 Chr.17: 12717691-12717691 on Build GRCh38
rs555047637 Chr.17: 12717500-12717500 on Build GRCh38
rs114839607 Chr.17: 12717394-12717394 on Build GRCh38
rs767804135 Chr.17: 12717393-12717393 on Build GRCh38
rs187220743 Chr.17: 12717632-12717632 on Build GRCh38
rs895310273 Chr.17: 12717655-12717655 on Build GRCh38
rs760467742 Chr.17: 12717400-12717400 on Build GRCh38
rs895331457 Chr.17: 12717237-12717237 on Build GRCh38
rs922487528 Chr.17: 12717481-12717481 on Build GRCh38
rs1025683321 Chr.17: 12717283-12717283 on Build GRCh38
rs574088661 Chr.17: 12717365-12717365 on Build GRCh38
rs1055973987 Chr.17: 12717651-12717651 on Build GRCh38
rs771322950 Chr.17: 12717370-12717370 on Build GRCh38
rs372478735 Chr.17: 12717364-12717364 on Build GRCh38
rs983169242 Chr.17: 12717341-12717341 on Build GRCh38
rs201202686 Chr.17: 12717356-12717356 on Build GRCh38
rs982389467 Chr.17: 12717568-12717568 on Build GRCh38
rs941100454 Chr.17: 12717618-12717618 on Build GRCh38
rs778335241 Chr.17: 12717459-12717459 on Build GRCh38
rs958082210 Chr.17: 12717406-12717406 on Build GRCh38
rs950686766 Chr.17: 12717652-12717652 on Build GRCh38
rs543795827 Chr.17: 12717657-12717657 on Build GRCh38
rs973292326 Chr.17: 12717264-12717264 on Build GRCh38
rs375113399 Chr.17: 12717472-12717472 on Build GRCh38
rs1012498560 Chr.17: 12717242-12717242 on Build GRCh38
rs12944823 Chr.17: 12717444-12717444 on Build GRCh38
rs749588446 Chr.17: 12717465-12717465 on Build GRCh38
rs758098578 Chr.17: 12717338-12717338 on Build GRCh38
rs145044197 Chr.17: 12717467-12717467 on Build GRCh38
rs549485517 Chr.17: 12717669-12717669 on Build GRCh38
rs531169226 Chr.17: 12717636-12717636 on Build GRCh38
rs529531835 Chr.17: 12717382-12717382 on Build GRCh38
rs140365970 Chr.17: 12717387-12717387 on Build GRCh38
rs775018858 Chr.17: 12717329-12717330 on Build GRCh38
rs757785621 Chr.17: 12717443-12717443 on Build GRCh38
rs994659110 Chr.17: 12717641-12717641 on Build GRCh38
rs376272243 Chr.17: 12717294-12717294 on Build GRCh38
rs1009023142 Chr.17: 12717509-12717509 on Build GRCh38
rs763934435 Chr.17: 12717404-12717404 on Build GRCh38
rs1027594652 Chr.17: 12717648-12717648 on Build GRCh38
rs745336949 Chr.17: 12717329-12717329 on Build GRCh38
rs548751076 Chr.17: 12717612-12717612 on Build GRCh38
rs759496322 Chr.17: 12717390-12717390 on Build GRCh38
rs779520845 Chr.17: 12717344-12717344 on Build GRCh38
rs12602716 Chr.17: 12717580-12717580 on Build GRCh38
rs770212120 Chr.17: 12717360-12717360 on Build GRCh38
rs753607516 Chr.17: 12717416-12717416 on Build GRCh38

View Full Product Details