Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 15588407-15588915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDRT1
Entrez Gene ID
374286
Gene Name
CMT1A duplicated region transcript 1
Gene Aliases
C170RF1, C17ORF1, C17ORF1A, FBXW10B, FBXW10P1, HREP, SM25H2
UniGene
Hs.548021
Gene Chromosome Location
Chr.17: 15565482-15619704 on Build GRCh38
Transcripts
NM_001282540.1 NM_006382.3

SNP Detail

SNP ID Chromosome Location
rs1036630422 Chr.17: 15588458-15588459 on Build GRCh38
rs2261062 Chr.17: 15588894-15588894 on Build GRCh38
rs930441831 Chr.17: 15588800-15588800 on Build GRCh38
rs151444 Chr.17: 15588527-15588527 on Build GRCh38
rs537079878 Chr.17: 15588886-15588886 on Build GRCh38
rs979010720 Chr.17: 15588758-15588758 on Build GRCh38
rs758388327 Chr.17: 15588686-15588686 on Build GRCh38
rs905878109 Chr.17: 15588560-15588560 on Build GRCh38
rs111806138 Chr.17: 15588642-15588642 on Build GRCh38
rs189567700 Chr.17: 15588740-15588740 on Build GRCh38
rs9909520 Chr.17: 15588565-15588565 on Build GRCh38
rs940466712 Chr.17: 15588795-15588795 on Build GRCh38
rs111558793 Chr.17: 15588516-15588516 on Build GRCh38
rs543923980 Chr.17: 15588879-15588881 on Build GRCh38
rs562041081 Chr.17: 15588686-15588686 on Build GRCh38
rs911543753 Chr.17: 15588785-15588785 on Build GRCh38
rs528618372 Chr.17: 15588629-15588629 on Build GRCh38
rs898992045 Chr.17: 15588487-15588487 on Build GRCh38
rs8076058 Chr.17: 15588799-15588799 on Build GRCh38
rs2123374 Chr.17: 15588832-15588832 on Build GRCh38
rs556936443 Chr.17: 15588892-15588892 on Build GRCh38
rs2261061 Chr.17: 15588742-15588742 on Build GRCh38
rs542120745 Chr.17: 15588648-15588648 on Build GRCh38
rs565877589 Chr.17: 15588787-15588787 on Build GRCh38
rs879039328 Chr.17: 15588855-15588855 on Build GRCh38
rs201749508 Chr.17: 15588495-15588495 on Build GRCh38
rs2261060 Chr.17: 15588688-15588688 on Build GRCh38
rs1029214798 Chr.17: 15588760-15588760 on Build GRCh38
rs960421764 Chr.17: 15588609-15588609 on Build GRCh38
rs530853221 Chr.17: 15588720-15588720 on Build GRCh38
rs953603465 Chr.17: 15588769-15588769 on Build GRCh38
rs546744729 Chr.17: 15588778-15588778 on Build GRCh38
rs533490433 Chr.17: 15588743-15588743 on Build GRCh38
rs1035691749 Chr.17: 15588594-15588594 on Build GRCh38
rs151443 Chr.17: 15588796-15588796 on Build GRCh38
rs1044866654 Chr.17: 15588833-15588833 on Build GRCh38
rs182183252 Chr.17: 15588885-15588885 on Build GRCh38

View Full Product Details