Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28742312-28742821 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YTHDF2
Entrez Gene ID
51441
Gene Name
YTH N6-methyladenosine RNA binding protein 2
Gene Aliases
CAHL, HGRG8, NY-REN-2
UniGene
Hs.532286
Gene Chromosome Location
Chr.1: 28736621-28769775 on Build GRCh38
Transcripts
NM_001172828.1 NM_001173128.1 NM_016258.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1492062 c.453C>G p.S151R Chr.1: 28742723-28742723 on Build GRCh38
6009185 c.477C>T p.S159S Chr.1: 28742747-28742747 on Build GRCh38
7081247 c.201C>T p.S67S Chr.1: 28742471-28742471 on Build GRCh38
5823935 c.535G>T p.A179S Chr.1: 28742805-28742805 on Build GRCh38
242294 c.236delG p.G79fs*8 Chr.1: 28742506-28742506 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765032904 Chr.1: 28742378-28742378 on Build GRCh38
rs752023542 Chr.1: 28742439-28742439 on Build GRCh38
rs773120628 Chr.1: 28742644-28742644 on Build GRCh38
rs772231330 Chr.1: 28742605-28742605 on Build GRCh38
rs765938444 Chr.1: 28742531-28742531 on Build GRCh38
rs764184098 Chr.1: 28742385-28742387 on Build GRCh38
rs377196196 Chr.1: 28742500-28742500 on Build GRCh38
rs915236261 Chr.1: 28742556-28742556 on Build GRCh38
rs779219050 Chr.1: 28742725-28742725 on Build GRCh38
rs764108934 Chr.1: 28742364-28742364 on Build GRCh38
rs777998164 Chr.1: 28742466-28742466 on Build GRCh38
rs769728659 Chr.1: 28742636-28742636 on Build GRCh38
rs773823654 Chr.1: 28742656-28742656 on Build GRCh38
rs772993578 Chr.1: 28742768-28742768 on Build GRCh38
rs774054308 Chr.1: 28742788-28742788 on Build GRCh38
rs11553689 Chr.1: 28742501-28742501 on Build GRCh38
rs761671171 Chr.1: 28742377-28742377 on Build GRCh38
rs372495672 Chr.1: 28742740-28742740 on Build GRCh38
rs767472994 Chr.1: 28742385-28742385 on Build GRCh38
rs569693049 Chr.1: 28742627-28742627 on Build GRCh38
rs934528708 Chr.1: 28742737-28742737 on Build GRCh38
rs767140624 Chr.1: 28742794-28742794 on Build GRCh38
rs764504425 Chr.1: 28742688-28742688 on Build GRCh38
rs749602577 Chr.1: 28742568-28742568 on Build GRCh38
rs778969888 Chr.1: 28742597-28742597 on Build GRCh38
rs1026461457 Chr.1: 28742346-28742346 on Build GRCh38
rs754268030 Chr.1: 28742709-28742709 on Build GRCh38
rs749456973 Chr.1: 28742468-28742468 on Build GRCh38
rs758556248 Chr.1: 28742735-28742735 on Build GRCh38
rs779553234 Chr.1: 28742426-28742426 on Build GRCh38
rs753436207 Chr.1: 28742779-28742779 on Build GRCh38
rs769922823 Chr.1: 28742759-28742759 on Build GRCh38
rs201869265 Chr.1: 28742471-28742471 on Build GRCh38
rs746011009 Chr.1: 28742598-28742598 on Build GRCh38
rs762653488 Chr.1: 28742649-28742649 on Build GRCh38
rs202034293 Chr.1: 28742524-28742524 on Build GRCh38
rs768752102 Chr.1: 28742497-28742497 on Build GRCh38
rs756472537 Chr.1: 28742561-28742561 on Build GRCh38
rs746039159 Chr.1: 28742729-28742729 on Build GRCh38
rs770588985 Chr.1: 28742786-28742786 on Build GRCh38
rs751151205 Chr.1: 28742548-28742548 on Build GRCh38
rs765085924 Chr.1: 28742519-28742519 on Build GRCh38
rs775354068 Chr.1: 28742384-28742385 on Build GRCh38
rs759294135 Chr.1: 28742358-28742358 on Build GRCh38
rs759126815 Chr.1: 28742789-28742789 on Build GRCh38
rs912177633 Chr.1: 28742430-28742430 on Build GRCh38
rs374492880 Chr.1: 28742678-28742678 on Build GRCh38
rs528989825 Chr.1: 28742680-28742680 on Build GRCh38
rs752116682 Chr.1: 28742399-28742399 on Build GRCh38
rs753750682 Chr.1: 28742372-28742372 on Build GRCh38
rs776350974 Chr.1: 28742352-28742352 on Build GRCh38
rs763496880 Chr.1: 28742700-28742700 on Build GRCh38
rs977579815 Chr.1: 28742504-28742504 on Build GRCh38
rs759075709 Chr.1: 28742668-28742668 on Build GRCh38
rs760602900 Chr.1: 28742357-28742357 on Build GRCh38
rs778053711 Chr.1: 28742564-28742564 on Build GRCh38
rs756670969 Chr.1: 28742682-28742682 on Build GRCh38
rs894195277 Chr.1: 28742633-28742633 on Build GRCh38
rs757877517 Chr.1: 28742393-28742393 on Build GRCh38
rs781663576 Chr.1: 28742449-28742449 on Build GRCh38
rs755569731 Chr.1: 28742441-28742441 on Build GRCh38
rs781678699 Chr.1: 28742553-28742553 on Build GRCh38
rs200088823 Chr.1: 28742621-28742621 on Build GRCh38
rs770091939 Chr.1: 28742460-28742460 on Build GRCh38
rs371160991 Chr.1: 28742669-28742669 on Build GRCh38
rs944926320 Chr.1: 28742461-28742461 on Build GRCh38
rs200930096 Chr.1: 28742615-28742615 on Build GRCh38
rs532078034 Chr.1: 28742580-28742580 on Build GRCh38
rs374197182 Chr.1: 28742744-28742744 on Build GRCh38
rs76268173 Chr.1: 28742480-28742480 on Build GRCh38
rs368431005 Chr.1: 28742718-28742718 on Build GRCh38
rs748558755 Chr.1: 28742456-28742456 on Build GRCh38
rs750047986 Chr.1: 28742389-28742389 on Build GRCh38
rs759529001 Chr.1: 28742482-28742482 on Build GRCh38
rs375674944 Chr.1: 28742558-28742558 on Build GRCh38
rs982349515 Chr.1: 28742398-28742398 on Build GRCh38
rs762613721 Chr.1: 28742526-28742526 on Build GRCh38
rs759340655 Chr.1: 28742552-28742552 on Build GRCh38
rs754022556 Chr.1: 28742474-28742476 on Build GRCh38

View Full Product Details