Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 17248764-17249186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS3
Entrez Gene ID
8533
Gene Name
COP9 signalosome subunit 3
Gene Aliases
CSN3, SGN3
UniGene
Hs.6076
Gene Chromosome Location
Chr.17: 17246624-17281303 on Build GRCh38
Transcripts
NM_001199125.1 NM_001316354.1 NM_001316355.1 NM_001316356.1 NM_001316357.1 NM_001316358.1 NM_003653.3 XM_005256837.4 XM_005256840.3 XM_005256842.4 XM_017025246.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6379240 c.1045G>T p.A349S Chr.17: 17249018-17249018 on Build GRCh38
372419 c.1127T>C p.I376T Chr.17: 17248936-17248936 on Build GRCh38
6634087 c.1088A>C p.N363T Chr.17: 17248975-17248975 on Build GRCh38
7114167 c.1129G>C p.D377H Chr.17: 17248934-17248934 on Build GRCh38
6634085 c.1124A>C p.N375T Chr.17: 17248939-17248939 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs907273033 Chr.17: 17249087-17249087 on Build GRCh38
rs779076700 Chr.17: 17249085-17249085 on Build GRCh38
rs148936216 Chr.17: 17248980-17248980 on Build GRCh38
rs11336908 Chr.17: 17249050-17249050 on Build GRCh38
rs929730344 Chr.17: 17249051-17249051 on Build GRCh38
rs756371016 Chr.17: 17248879-17248879 on Build GRCh38
rs202013101 Chr.17: 17249001-17249001 on Build GRCh38
rs747502184 Chr.17: 17249050-17249052 on Build GRCh38
rs368885985 Chr.17: 17248908-17248908 on Build GRCh38
rs955234252 Chr.17: 17248919-17248919 on Build GRCh38
rs749362131 Chr.17: 17248892-17248892 on Build GRCh38
rs137898777 Chr.17: 17249094-17249094 on Build GRCh38
rs986630105 Chr.17: 17248955-17248955 on Build GRCh38
rs760118803 Chr.17: 17249045-17249045 on Build GRCh38
rs1055737924 Chr.17: 17249161-17249161 on Build GRCh38
rs530130850 Chr.17: 17249089-17249089 on Build GRCh38
rs79888342 Chr.17: 17249062-17249062 on Build GRCh38
rs778653192 Chr.17: 17248928-17248928 on Build GRCh38
rs986431854 Chr.17: 17248864-17248864 on Build GRCh38
rs775388275 Chr.17: 17248951-17248951 on Build GRCh38
rs754120112 Chr.17: 17249074-17249074 on Build GRCh38
rs546444024 Chr.17: 17249036-17249036 on Build GRCh38
rs564942402 Chr.17: 17249050-17249051 on Build GRCh38
rs912113493 Chr.17: 17248874-17248874 on Build GRCh38
rs12162147 Chr.17: 17249004-17249004 on Build GRCh38
rs200374983 Chr.17: 17249068-17249068 on Build GRCh38
rs778185979 Chr.17: 17248881-17248881 on Build GRCh38
rs771719189 Chr.17: 17248943-17248943 on Build GRCh38
rs752154824 Chr.17: 17249105-17249105 on Build GRCh38
rs143738729 Chr.17: 17248942-17248942 on Build GRCh38
rs559532101 Chr.17: 17249051-17249052 on Build GRCh38
rs753085602 Chr.17: 17249050-17249050 on Build GRCh38
rs553573176 Chr.17: 17249049-17249049 on Build GRCh38
rs150284267 Chr.17: 17248857-17248857 on Build GRCh38
rs755680016 Chr.17: 17249049-17249049 on Build GRCh38
rs1048129382 Chr.17: 17249054-17249054 on Build GRCh38
rs756602053 Chr.17: 17249064-17249064 on Build GRCh38
rs376927180 Chr.17: 17249142-17249142 on Build GRCh38
rs1055987290 Chr.17: 17249098-17249098 on Build GRCh38
rs1041861539 Chr.17: 17249075-17249075 on Build GRCh38
rs772637302 Chr.17: 17249049-17249050 on Build GRCh38
rs537192939 Chr.17: 17248859-17248859 on Build GRCh38
rs199692441 Chr.17: 17248983-17248983 on Build GRCh38
rs373209658 Chr.17: 17249082-17249082 on Build GRCh38
rs747840057 Chr.17: 17249045-17249046 on Build GRCh38
rs563472230 Chr.17: 17249051-17249051 on Build GRCh38
rs146811066 Chr.17: 17248962-17248962 on Build GRCh38
rs201493311 Chr.17: 17249015-17249015 on Build GRCh38
rs776324980 Chr.17: 17248998-17248998 on Build GRCh38
rs774671125 Chr.17: 17249039-17249039 on Build GRCh38
rs1016607409 Chr.17: 17248818-17248818 on Build GRCh38

View Full Product Details