Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28369429-28369937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VTN
Entrez Gene ID
7448
Gene Name
vitronectin
Gene Aliases
V75, VN, VNT
UniGene
Hs.2257
Gene Chromosome Location
Chr.17: 28367277-28370352 on Build GRCh38
Transcripts
NM_000638.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6820910 c.190C>T p.R64C Chr.17: 28369846-28369846 on Build GRCh38
6523380 c.392A>T p.K131M Chr.17: 28369644-28369644 on Build GRCh38
3983321 c.315G>C p.Q105H Chr.17: 28369721-28369721 on Build GRCh38
5524179 c.194G>A p.G65E Chr.17: 28369842-28369842 on Build GRCh38
3819092 c.430C>T p.P144S Chr.17: 28369606-28369606 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782075144 Chr.17: 28369798-28369798 on Build GRCh38
rs1044296654 Chr.17: 28369542-28369542 on Build GRCh38
rs1027478384 Chr.17: 28369775-28369775 on Build GRCh38
rs781910413 Chr.17: 28369802-28369802 on Build GRCh38
rs781814928 Chr.17: 28369670-28369670 on Build GRCh38
rs782162923 Chr.17: 28369510-28369510 on Build GRCh38
rs758080124 Chr.17: 28369881-28369881 on Build GRCh38
rs144093348 Chr.17: 28369665-28369665 on Build GRCh38
rs2071378 Chr.17: 28369456-28369456 on Build GRCh38
rs532276485 Chr.17: 28369483-28369483 on Build GRCh38
rs782542357 Chr.17: 28369552-28369552 on Build GRCh38
rs782708368 Chr.17: 28369675-28369675 on Build GRCh38
rs145095194 Chr.17: 28369845-28369845 on Build GRCh38
rs782649817 Chr.17: 28369472-28369472 on Build GRCh38
rs782791340 Chr.17: 28369660-28369660 on Build GRCh38
rs782168976 Chr.17: 28369846-28369846 on Build GRCh38
rs782631664 Chr.17: 28369836-28369836 on Build GRCh38
rs782820530 Chr.17: 28369549-28369549 on Build GRCh38
rs371699977 Chr.17: 28369725-28369725 on Build GRCh38
rs782442875 Chr.17: 28369604-28369604 on Build GRCh38
rs781791270 Chr.17: 28369904-28369904 on Build GRCh38
rs782496984 Chr.17: 28369822-28369822 on Build GRCh38
rs782508700 Chr.17: 28369728-28369728 on Build GRCh38
rs782023049 Chr.17: 28369509-28369509 on Build GRCh38
rs781909548 Chr.17: 28369462-28369462 on Build GRCh38
rs782142233 Chr.17: 28369912-28369912 on Build GRCh38
rs782617667 Chr.17: 28369844-28369844 on Build GRCh38
rs782122891 Chr.17: 28369808-28369808 on Build GRCh38
rs782665898 Chr.17: 28369754-28369754 on Build GRCh38
rs2071377 Chr.17: 28369461-28369461 on Build GRCh38
rs543411017 Chr.17: 28369663-28369663 on Build GRCh38
rs782269687 Chr.17: 28369479-28369479 on Build GRCh38
rs782595048 Chr.17: 28369588-28369588 on Build GRCh38
rs781824403 Chr.17: 28369817-28369817 on Build GRCh38
rs782379851 Chr.17: 28369885-28369885 on Build GRCh38
rs782262705 Chr.17: 28369630-28369630 on Build GRCh38
rs944354168 Chr.17: 28369816-28369816 on Build GRCh38
rs2227741 Chr.17: 28369672-28369672 on Build GRCh38
rs112691790 Chr.17: 28369823-28369823 on Build GRCh38
rs35552133 Chr.17: 28369887-28369888 on Build GRCh38
rs546780648 Chr.17: 28369751-28369752 on Build GRCh38
rs148666516 Chr.17: 28369789-28369789 on Build GRCh38
rs557265411 Chr.17: 28369627-28369627 on Build GRCh38
rs112887300 Chr.17: 28369659-28369659 on Build GRCh38
rs924108792 Chr.17: 28369850-28369850 on Build GRCh38
rs373650198 Chr.17: 28369550-28369550 on Build GRCh38
rs782737815 Chr.17: 28369901-28369901 on Build GRCh38
rs45518737 Chr.17: 28369469-28369469 on Build GRCh38
rs782255864 Chr.17: 28369490-28369490 on Build GRCh38
rs782372739 Chr.17: 28369765-28369765 on Build GRCh38
rs782689771 Chr.17: 28369706-28369706 on Build GRCh38
rs782630613 Chr.17: 28369488-28369488 on Build GRCh38
rs782335341 Chr.17: 28369491-28369491 on Build GRCh38
rs367756510 Chr.17: 28369747-28369747 on Build GRCh38
rs781997284 Chr.17: 28369467-28369467 on Build GRCh38
rs782540967 Chr.17: 28369729-28369730 on Build GRCh38
rs782345251 Chr.17: 28369909-28369909 on Build GRCh38
rs782819003 Chr.17: 28369913-28369913 on Build GRCh38
rs781837413 Chr.17: 28369477-28369477 on Build GRCh38
rs374926698 Chr.17: 28369496-28369496 on Build GRCh38
rs184192380 Chr.17: 28369843-28369843 on Build GRCh38
rs782593575 Chr.17: 28369899-28369899 on Build GRCh38
rs782327226 Chr.17: 28369748-28369748 on Build GRCh38
rs199522124 Chr.17: 28369614-28369614 on Build GRCh38
rs782541028 Chr.17: 28369701-28369701 on Build GRCh38
rs782463783 Chr.17: 28369553-28369553 on Build GRCh38
rs150757499 Chr.17: 28369809-28369809 on Build GRCh38
rs782003778 Chr.17: 28369897-28369897 on Build GRCh38
rs369239403 Chr.17: 28369508-28369508 on Build GRCh38
rs368033486 Chr.17: 28369793-28369793 on Build GRCh38
rs781814438 Chr.17: 28369697-28369697 on Build GRCh38
rs782430869 Chr.17: 28369478-28369478 on Build GRCh38
rs375615794 Chr.17: 28369664-28369664 on Build GRCh38
rs370458755 Chr.17: 28369735-28369735 on Build GRCh38
rs782515931 Chr.17: 28369726-28369726 on Build GRCh38
rs782183692 Chr.17: 28369498-28369498 on Build GRCh38
rs781850224 Chr.17: 28369564-28369564 on Build GRCh38
rs201384220 Chr.17: 28369824-28369824 on Build GRCh38
rs782170052 Chr.17: 28369860-28369860 on Build GRCh38
rs782332650 Chr.17: 28369848-28369848 on Build GRCh38
rs970401046 Chr.17: 28369756-28369756 on Build GRCh38
rs782593190 Chr.17: 28369610-28369610 on Build GRCh38
rs782022688 Chr.17: 28369856-28369856 on Build GRCh38
rs781910250 Chr.17: 28369609-28369609 on Build GRCh38
rs200955799 Chr.17: 28369646-28369646 on Build GRCh38
rs781994851 Chr.17: 28369774-28369774 on Build GRCh38
rs541153221 Chr.17: 28369830-28369830 on Build GRCh38
rs781798269 Chr.17: 28369455-28369455 on Build GRCh38
rs370834847 Chr.17: 28369577-28369577 on Build GRCh38
rs35339100 Chr.17: 28369529-28369529 on Build GRCh38
rs377679842 Chr.17: 28369626-28369626 on Build GRCh38
rs782346117 Chr.17: 28369791-28369791 on Build GRCh38
rs1012667049 Chr.17: 28369734-28369734 on Build GRCh38
rs141253462 Chr.17: 28369799-28369799 on Build GRCh38
rs782222213 Chr.17: 28369592-28369592 on Build GRCh38
rs782005192 Chr.17: 28369541-28369541 on Build GRCh38
rs781919497 Chr.17: 28369651-28369651 on Build GRCh38
rs782653082 Chr.17: 28369887-28369887 on Build GRCh38

View Full Product Details