Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28616594-28617100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0100
Entrez Gene ID
9703
Gene Name
KIAA0100
Gene Aliases
BCOX, BCOX1, CT101, FMP27
UniGene
Hs.151761
Gene Chromosome Location
Chr.17: 28614440-28645159 on Build GRCh38
Transcripts
NM_014680.3 XM_005258073.2 XM_017025423.1 NR_040012.1

Gene Symbol
SPAG5-AS1
Entrez Gene ID
100506436
Gene Name
SPAG5 antisense RNA 1
Gene Aliases
-
UniGene
Hs.743630
Gene Chromosome Location
Chr.17: 28599133-28617377 on Build GRCh38
Transcripts
NM_014680.3 XM_005258073.2 XM_017025423.1 NR_040012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4064961 c.6086T>A p.F2029Y Chr.17: 28616689-28616689 on Build GRCh38
4064963 c.5980C>T p.R1994W Chr.17: 28616930-28616930 on Build GRCh38
3378105 c.6098G>A p.R2033Q Chr.17: 28616677-28616677 on Build GRCh38
3819114 c.6157+2T>G p.? Chr.17: 28616616-28616616 on Build GRCh38
6357212 c.5921G>A p.R1974Q Chr.17: 28616989-28616989 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760118789 Chr.17: 28616948-28616948 on Build GRCh38
rs980270920 Chr.17: 28617015-28617015 on Build GRCh38
rs752543165 Chr.17: 28617028-28617028 on Build GRCh38
rs764654510 Chr.17: 28617042-28617043 on Build GRCh38
rs938109117 Chr.17: 28616812-28616812 on Build GRCh38
rs748270972 Chr.17: 28616860-28616860 on Build GRCh38
rs925690132 Chr.17: 28616841-28616841 on Build GRCh38
rs767149095 Chr.17: 28616980-28616980 on Build GRCh38
rs754702224 Chr.17: 28616853-28616853 on Build GRCh38
rs369125189 Chr.17: 28616772-28616772 on Build GRCh38
rs768556642 Chr.17: 28616760-28616760 on Build GRCh38
rs764882466 Chr.17: 28617021-28617021 on Build GRCh38
rs778118461 Chr.17: 28617033-28617033 on Build GRCh38
rs756690315 Chr.17: 28616663-28616663 on Build GRCh38
rs373012241 Chr.17: 28616632-28616632 on Build GRCh38
rs749647969 Chr.17: 28617046-28617046 on Build GRCh38
rs375361944 Chr.17: 28616838-28616838 on Build GRCh38
rs748918145 Chr.17: 28616670-28616671 on Build GRCh38
rs772351072 Chr.17: 28616855-28616855 on Build GRCh38
rs761017343 Chr.17: 28617009-28617010 on Build GRCh38
rs950780608 Chr.17: 28616679-28616679 on Build GRCh38
rs866318934 Chr.17: 28616929-28616929 on Build GRCh38
rs750307079 Chr.17: 28616720-28616720 on Build GRCh38
rs777246681 Chr.17: 28616921-28616921 on Build GRCh38
rs761545535 Chr.17: 28616801-28616801 on Build GRCh38
rs753825105 Chr.17: 28616852-28616852 on Build GRCh38
rs775932115 Chr.17: 28616963-28616963 on Build GRCh38
rs770001686 Chr.17: 28616959-28616959 on Build GRCh38
rs755685175 Chr.17: 28616847-28616847 on Build GRCh38
rs753682478 Chr.17: 28617016-28617016 on Build GRCh38
rs551308663 Chr.17: 28616880-28616880 on Build GRCh38
rs921129618 Chr.17: 28616786-28616786 on Build GRCh38
rs139355794 Chr.17: 28616988-28616988 on Build GRCh38
rs199533260 Chr.17: 28616795-28616795 on Build GRCh38
rs372910327 Chr.17: 28616778-28616778 on Build GRCh38
rs781444088 Chr.17: 28617036-28617036 on Build GRCh38
rs371436851 Chr.17: 28617030-28617030 on Build GRCh38
rs779957103 Chr.17: 28616849-28616849 on Build GRCh38
rs747071615 Chr.17: 28616898-28616898 on Build GRCh38
rs749080893 Chr.17: 28616755-28616755 on Build GRCh38
rs754269381 Chr.17: 28616627-28616627 on Build GRCh38
rs776641490 Chr.17: 28616858-28616858 on Build GRCh38
rs754965960 Chr.17: 28617018-28617018 on Build GRCh38
rs748445942 Chr.17: 28616771-28616771 on Build GRCh38
rs763638904 Chr.17: 28616637-28616637 on Build GRCh38
rs988468386 Chr.17: 28616758-28616758 on Build GRCh38
rs746298700 Chr.17: 28617040-28617040 on Build GRCh38
rs760158902 Chr.17: 28616990-28616990 on Build GRCh38
rs771182650 Chr.17: 28616906-28616906 on Build GRCh38
rs538681329 Chr.17: 28617006-28617011 on Build GRCh38
rs769060940 Chr.17: 28617048-28617048 on Build GRCh38
rs765917708 Chr.17: 28617005-28617005 on Build GRCh38
rs867044347 Chr.17: 28616867-28616867 on Build GRCh38
rs201896448 Chr.17: 28616738-28616738 on Build GRCh38
rs780781168 Chr.17: 28616685-28616685 on Build GRCh38
rs1044619120 Chr.17: 28617052-28617052 on Build GRCh38
rs751273890 Chr.17: 28616641-28616641 on Build GRCh38
rs578041145 Chr.17: 28616781-28616781 on Build GRCh38
rs747273983 Chr.17: 28617034-28617034 on Build GRCh38
rs375780767 Chr.17: 28616768-28616768 on Build GRCh38
rs770334294 Chr.17: 28617041-28617041 on Build GRCh38
rs376115781 Chr.17: 28616792-28616792 on Build GRCh38
rs775924543 Chr.17: 28617043-28617043 on Build GRCh38
rs749938329 Chr.17: 28616840-28616840 on Build GRCh38
rs766803007 Chr.17: 28616622-28616622 on Build GRCh38
rs779754747 Chr.17: 28616748-28616748 on Build GRCh38
rs778984368 Chr.17: 28616765-28616765 on Build GRCh38
rs531142947 Chr.17: 28616885-28616885 on Build GRCh38
rs772353763 Chr.17: 28616865-28616865 on Build GRCh38
rs761302694 Chr.17: 28616970-28616970 on Build GRCh38
rs1048449244 Chr.17: 28616819-28616819 on Build GRCh38
rs895466858 Chr.17: 28617014-28617014 on Build GRCh38
rs1007304345 Chr.17: 28616661-28616661 on Build GRCh38

View Full Product Details