Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43399982-43400475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL4D
Entrez Gene ID
379
Gene Name
ADP ribosylation factor like GTPase 4D
Gene Aliases
ARF4L, ARL6
UniGene
Hs.183153
Gene Chromosome Location
Chr.17: 43398985-43401136 on Build GRCh38
Transcripts
NM_001661.3 XM_011524782.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5578064 c.341T>C p.V114A Chr.17: 43400073-43400073 on Build GRCh38
5564127 c.322C>T p.R108W Chr.17: 43400054-43400054 on Build GRCh38
243053 c.334G>A p.A112T Chr.17: 43400066-43400066 on Build GRCh38
6189943 c.573G>T p.K191N Chr.17: 43400305-43400305 on Build GRCh38
6147005 c.275G>T p.R92L Chr.17: 43400007-43400007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779778668 Chr.17: 43400222-43400222 on Build GRCh38
rs866011666 Chr.17: 43400210-43400210 on Build GRCh38
rs61749921 Chr.17: 43400302-43400302 on Build GRCh38
rs746029987 Chr.17: 43400089-43400089 on Build GRCh38
rs955391996 Chr.17: 43400043-43400043 on Build GRCh38
rs761216623 Chr.17: 43400042-43400042 on Build GRCh38
rs1036573759 Chr.17: 43400446-43400446 on Build GRCh38
rs181617545 Chr.17: 43400365-43400365 on Build GRCh38
rs997417710 Chr.17: 43400398-43400398 on Build GRCh38
rs930229985 Chr.17: 43400218-43400218 on Build GRCh38
rs780810752 Chr.17: 43400373-43400373 on Build GRCh38
rs942197588 Chr.17: 43400445-43400445 on Build GRCh38
rs3179287 Chr.17: 43400342-43400342 on Build GRCh38
rs573025377 Chr.17: 43400341-43400341 on Build GRCh38
rs1050650948 Chr.17: 43400138-43400138 on Build GRCh38
rs868719891 Chr.17: 43400100-43400100 on Build GRCh38
rs774397233 Chr.17: 43400017-43400017 on Build GRCh38
rs766095828 Chr.17: 43400152-43400152 on Build GRCh38
rs1002576957 Chr.17: 43400217-43400217 on Build GRCh38
rs751943750 Chr.17: 43400195-43400195 on Build GRCh38
rs774799110 Chr.17: 43400350-43400350 on Build GRCh38
rs941492394 Chr.17: 43400113-43400113 on Build GRCh38
rs142692407 Chr.17: 43400098-43400098 on Build GRCh38
rs369611018 Chr.17: 43400383-43400383 on Build GRCh38
rs955338315 Chr.17: 43400431-43400431 on Build GRCh38
rs147354814 Chr.17: 43400149-43400149 on Build GRCh38
rs774152596 Chr.17: 43400142-43400142 on Build GRCh38
rs781336467 Chr.17: 43400084-43400084 on Build GRCh38
rs764306538 Chr.17: 43400347-43400347 on Build GRCh38
rs113009855 Chr.17: 43400430-43400430 on Build GRCh38
rs771429264 Chr.17: 43400147-43400147 on Build GRCh38
rs770023977 Chr.17: 43400093-43400093 on Build GRCh38
rs767123284 Chr.17: 43400047-43400047 on Build GRCh38
rs935334904 Chr.17: 43400010-43400010 on Build GRCh38
rs896130940 Chr.17: 43400250-43400250 on Build GRCh38
rs931946757 Chr.17: 43400413-43400413 on Build GRCh38
rs958350187 Chr.17: 43400277-43400277 on Build GRCh38
rs368069774 Chr.17: 43400044-43400044 on Build GRCh38
rs780243844 Chr.17: 43400222-43400222 on Build GRCh38
rs142573585 Chr.17: 43400239-43400239 on Build GRCh38
rs1044804021 Chr.17: 43400316-43400316 on Build GRCh38
rs757796175 Chr.17: 43400196-43400196 on Build GRCh38
rs748124890 Chr.17: 43400146-43400146 on Build GRCh38
rs200026555 Chr.17: 43400208-43400208 on Build GRCh38
rs974266100 Chr.17: 43400186-43400186 on Build GRCh38
rs763964406 Chr.17: 43400343-43400343 on Build GRCh38
rs760421620 Chr.17: 43400053-43400053 on Build GRCh38
rs924023421 Chr.17: 43400345-43400345 on Build GRCh38
rs535882068 Chr.17: 43400346-43400346 on Build GRCh38
rs765651568 Chr.17: 43400055-43400055 on Build GRCh38
rs778115782 Chr.17: 43400071-43400071 on Build GRCh38
rs756182377 Chr.17: 43400359-43400359 on Build GRCh38
rs776470066 Chr.17: 43400153-43400153 on Build GRCh38
rs750033537 Chr.17: 43400051-43400051 on Build GRCh38
rs758871820 Chr.17: 43400070-43400070 on Build GRCh38
rs764583641 Chr.17: 43400189-43400189 on Build GRCh38
rs139560078 Chr.17: 43400339-43400339 on Build GRCh38
rs372258038 Chr.17: 43400078-43400078 on Build GRCh38
rs746640974 Chr.17: 43400328-43400329 on Build GRCh38
rs749061842 Chr.17: 43400107-43400107 on Build GRCh38
rs1059968 Chr.17: 43400004-43400004 on Build GRCh38
rs200670918 Chr.17: 43400345-43400345 on Build GRCh38
rs139507398 Chr.17: 43400200-43400200 on Build GRCh38
rs769598616 Chr.17: 43400364-43400364 on Build GRCh38
rs911312917 Chr.17: 43400101-43400101 on Build GRCh38
rs747991125 Chr.17: 43400315-43400315 on Build GRCh38
rs763210222 Chr.17: 43400183-43400183 on Build GRCh38
rs1059974 Chr.17: 43400122-43400122 on Build GRCh38
rs768447728 Chr.17: 43400344-43400345 on Build GRCh38
rs1030675121 Chr.17: 43400429-43400429 on Build GRCh38
rs750988206 Chr.17: 43400351-43400351 on Build GRCh38
rs754663381 Chr.17: 43400249-43400249 on Build GRCh38
rs377180469 Chr.17: 43400216-43400216 on Build GRCh38
rs768554539 Chr.17: 43400125-43400125 on Build GRCh38
rs753140896 Chr.17: 43400069-43400069 on Build GRCh38
rs755255761 Chr.17: 43400390-43400390 on Build GRCh38

View Full Product Details