Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44170599-44171096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB16
Entrez Gene ID
92591
Gene Name
ankyrin repeat and SOCS box containing 16
Gene Aliases
-
UniGene
Hs.534517
Gene Chromosome Location
Chr.17: 44170706-44179083 on Build GRCh38
Transcripts
NM_080863.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
706185 c.101A>T p.Q34L Chr.17: 44170890-44170890 on Build GRCh38
5508973 c.101A>C p.Q34P Chr.17: 44170890-44170890 on Build GRCh38
979893 c.231C>T p.F77F Chr.17: 44171020-44171020 on Build GRCh38
979894 c.234A>C p.Q78H Chr.17: 44171023-44171023 on Build GRCh38
4151631 c.253A>C p.M85L Chr.17: 44171042-44171042 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771660999 Chr.17: 44170908-44170908 on Build GRCh38
rs777406910 Chr.17: 44170959-44170959 on Build GRCh38
rs56265171 Chr.17: 44170837-44170837 on Build GRCh38
rs763502506 Chr.17: 44170877-44170877 on Build GRCh38
rs200827398 Chr.17: 44170782-44170782 on Build GRCh38
rs761296640 Chr.17: 44170860-44170860 on Build GRCh38
rs144229400 Chr.17: 44170871-44170871 on Build GRCh38
rs759914571 Chr.17: 44170826-44170826 on Build GRCh38
rs542752674 Chr.17: 44171047-44171047 on Build GRCh38
rs192855432 Chr.17: 44170768-44170768 on Build GRCh38
rs371159852 Chr.17: 44170958-44170958 on Build GRCh38
rs375392717 Chr.17: 44170732-44170732 on Build GRCh38
rs7218599 Chr.17: 44170978-44170978 on Build GRCh38
rs776085869 Chr.17: 44170974-44170974 on Build GRCh38
rs760686231 Chr.17: 44170992-44170992 on Build GRCh38
rs754307863 Chr.17: 44170754-44170754 on Build GRCh38
rs778649100 Chr.17: 44170905-44170905 on Build GRCh38
rs149170804 Chr.17: 44170928-44170928 on Build GRCh38
rs1027621389 Chr.17: 44170971-44170971 on Build GRCh38
rs1048746941 Chr.17: 44170991-44170991 on Build GRCh38
rs768859857 Chr.17: 44170981-44170981 on Build GRCh38
rs528495680 Chr.17: 44170898-44170898 on Build GRCh38
rs758404845 Chr.17: 44171026-44171026 on Build GRCh38
rs748110246 Chr.17: 44170907-44170907 on Build GRCh38
rs745476523 Chr.17: 44171036-44171036 on Build GRCh38
rs971252008 Chr.17: 44170720-44170720 on Build GRCh38
rs572124875 Chr.17: 44171032-44171032 on Build GRCh38
rs75781143 Chr.17: 44170950-44170950 on Build GRCh38
rs758866778 Chr.17: 44170830-44170830 on Build GRCh38
rs371686293 Chr.17: 44171068-44171068 on Build GRCh38
rs143324294 Chr.17: 44170931-44170931 on Build GRCh38
rs527398853 Chr.17: 44170723-44170723 on Build GRCh38
rs755528606 Chr.17: 44170755-44170755 on Build GRCh38
rs766824314 Chr.17: 44170753-44170753 on Build GRCh38
rs568896513 Chr.17: 44170801-44170801 on Build GRCh38
rs544986778 Chr.17: 44170984-44170984 on Build GRCh38
rs147113944 Chr.17: 44170990-44170990 on Build GRCh38
rs764528773 Chr.17: 44170929-44170929 on Build GRCh38
rs770479254 Chr.17: 44170972-44170972 on Build GRCh38
rs779630328 Chr.17: 44170759-44170759 on Build GRCh38
rs567496977 Chr.17: 44170948-44170948 on Build GRCh38
rs1027215236 Chr.17: 44170655-44170655 on Build GRCh38
rs761508752 Chr.17: 44170813-44170813 on Build GRCh38
rs754096340 Chr.17: 44170949-44170949 on Build GRCh38
rs749287866 Chr.17: 44170813-44170821 on Build GRCh38
rs781268578 Chr.17: 44170891-44170891 on Build GRCh38
rs377163601 Chr.17: 44170834-44170834 on Build GRCh38
rs775059291 Chr.17: 44171005-44171009 on Build GRCh38
rs545105977 Chr.17: 44170698-44170698 on Build GRCh38
rs370158958 Chr.17: 44170925-44170925 on Build GRCh38
rs764575988 Chr.17: 44170878-44170878 on Build GRCh38
rs774231569 Chr.17: 44170868-44170910 on Build GRCh38
rs753051378 Chr.17: 44170874-44170874 on Build GRCh38
rs766494048 Chr.17: 44171000-44171000 on Build GRCh38
rs746583463 Chr.17: 44170967-44170967 on Build GRCh38
rs776397394 Chr.17: 44170788-44170788 on Build GRCh38
rs953114278 Chr.17: 44170862-44170862 on Build GRCh38
rs144140141 Chr.17: 44170839-44170839 on Build GRCh38
rs751177767 Chr.17: 44170745-44170745 on Build GRCh38
rs140506716 Chr.17: 44170870-44170870 on Build GRCh38
rs945211411 Chr.17: 44170797-44170797 on Build GRCh38
rs952884480 Chr.17: 44170663-44170663 on Build GRCh38
rs367837547 Chr.17: 44170817-44170817 on Build GRCh38
rs766933192 Chr.17: 44170779-44170779 on Build GRCh38
rs745726300 Chr.17: 44170793-44170793 on Build GRCh38
rs753959435 Chr.17: 44171001-44171001 on Build GRCh38
rs777744675 Chr.17: 44170770-44170770 on Build GRCh38
rs777886270 Chr.17: 44170833-44170833 on Build GRCh38
rs759040207 Chr.17: 44170914-44170915 on Build GRCh38
rs138591784 Chr.17: 44171005-44171005 on Build GRCh38
rs201364601 Chr.17: 44170853-44170853 on Build GRCh38
rs769577332 Chr.17: 44170794-44170794 on Build GRCh38
rs767339837 Chr.17: 44170821-44170821 on Build GRCh38
rs749221208 Chr.17: 44170847-44170847 on Build GRCh38
rs752107616 Chr.17: 44170881-44170881 on Build GRCh38
rs769537913 Chr.17: 44171046-44171046 on Build GRCh38
rs768528706 Chr.17: 44170849-44170849 on Build GRCh38
rs775918008 Chr.17: 44170926-44170926 on Build GRCh38
rs997016890 Chr.17: 44170629-44170629 on Build GRCh38
rs143380581 Chr.17: 44171059-44171059 on Build GRCh38
rs113632650 Chr.17: 44170697-44170697 on Build GRCh38
rs866246827 Chr.17: 44171057-44171057 on Build GRCh38
rs767555969 Chr.17: 44170932-44170932 on Build GRCh38
rs754251112 Chr.17: 44170822-44170822 on Build GRCh38
rs773776656 Chr.17: 44170808-44170808 on Build GRCh38
rs760380688 Chr.17: 44170915-44170915 on Build GRCh38
rs768638576 Chr.17: 44170904-44170904 on Build GRCh38
rs369796709 Chr.17: 44170744-44170744 on Build GRCh38
rs773252427 Chr.17: 44170803-44170808 on Build GRCh38
rs146524979 Chr.17: 44170844-44170844 on Build GRCh38
rs376996083 Chr.17: 44170899-44170899 on Build GRCh38
rs781322387 Chr.17: 44170838-44170838 on Build GRCh38
rs752595469 Chr.17: 44171024-44171024 on Build GRCh38
rs765664033 Chr.17: 44170872-44170872 on Build GRCh38
rs199735813 Chr.17: 44170773-44170773 on Build GRCh38
rs376160399 Chr.17: 44171016-44171016 on Build GRCh38
rs747221966 Chr.17: 44170775-44170775 on Build GRCh38
rs145226168 Chr.17: 44170914-44170914 on Build GRCh38
rs981791311 Chr.17: 44171066-44171066 on Build GRCh38
rs762817455 Chr.17: 44170803-44170803 on Build GRCh38
rs763623202 Chr.17: 44170804-44170804 on Build GRCh38
rs770798712 Chr.17: 44170815-44170817 on Build GRCh38
rs760171144 Chr.17: 44171011-44171014 on Build GRCh38
rs560261909 Chr.17: 44170701-44170701 on Build GRCh38
rs779878771 Chr.17: 44170902-44170902 on Build GRCh38
rs200273394 Chr.17: 44170750-44170750 on Build GRCh38
rs577750620 Chr.17: 44170695-44170695 on Build GRCh38
rs750553915 Chr.17: 44170938-44170938 on Build GRCh38
rs368831858 Chr.17: 44171027-44171027 on Build GRCh38
rs780782857 Chr.17: 44171035-44171035 on Build GRCh38
rs534308231 Chr.17: 44170882-44170882 on Build GRCh38
rs533182208 Chr.17: 44170829-44170829 on Build GRCh38
rs970441000 Chr.17: 44171009-44171009 on Build GRCh38
rs773651180 Chr.17: 44171069-44171069 on Build GRCh38
rs767211105 Chr.17: 44170920-44170920 on Build GRCh38
rs905098190 Chr.17: 44170736-44170736 on Build GRCh38

View Full Product Details