Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44320270-44320775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A39
Entrez Gene ID
51629
Gene Name
solute carrier family 25 member 39
Gene Aliases
CGI-69, CGI69
UniGene
Hs.744880
Gene Chromosome Location
Chr.17: 44319625-44324870 on Build GRCh38
Transcripts
NM_001143780.2 NM_001321240.1 NM_001321241.1 NM_016016.3 XM_011524880.2 XM_011524882.2 XM_011524883.2 XM_011524884.1 XM_017024737.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6390069 c.743G>C p.G248A Chr.17: 44320656-44320656 on Build GRCh38
4130142 c.762T>G p.G254G Chr.17: 44320637-44320637 on Build GRCh38
6388508 c.742G>T p.G248C Chr.17: 44320657-44320657 on Build GRCh38
4783675 c.739T>C p.F247L Chr.17: 44320684-44320684 on Build GRCh38
4066897 c.830T>C p.V277A Chr.17: 44320384-44320384 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs4503841 Chr.17: 44320662-44320662 on Build GRCh38
rs552106177 Chr.17: 44320571-44320571 on Build GRCh38
rs200749023 Chr.17: 44320469-44320469 on Build GRCh38
rs1002967668 Chr.17: 44320570-44320571 on Build GRCh38
rs937375539 Chr.17: 44320544-44320544 on Build GRCh38
rs1035934402 Chr.17: 44320729-44320729 on Build GRCh38
rs976865852 Chr.17: 44320459-44320459 on Build GRCh38
rs777686644 Chr.17: 44320637-44320637 on Build GRCh38
rs375195968 Chr.17: 44320623-44320623 on Build GRCh38
rs780133418 Chr.17: 44320486-44320486 on Build GRCh38
rs569932603 Chr.17: 44320413-44320413 on Build GRCh38
rs531119889 Chr.17: 44320550-44320550 on Build GRCh38
rs754389470 Chr.17: 44320304-44320304 on Build GRCh38
rs908063595 Chr.17: 44320515-44320515 on Build GRCh38
rs373801902 Chr.17: 44320342-44320342 on Build GRCh38
rs761222982 Chr.17: 44320364-44320364 on Build GRCh38
rs546731911 Chr.17: 44320500-44320500 on Build GRCh38
rs369965341 Chr.17: 44320722-44320722 on Build GRCh38
rs749231936 Chr.17: 44320289-44320289 on Build GRCh38
rs766331338 Chr.17: 44320701-44320701 on Build GRCh38
rs768064559 Chr.17: 44320359-44320359 on Build GRCh38
rs368359016 Chr.17: 44320436-44320436 on Build GRCh38
rs368274431 Chr.17: 44320576-44320576 on Build GRCh38
rs760257390 Chr.17: 44320451-44320451 on Build GRCh38
rs760861201 Chr.17: 44320689-44320689 on Build GRCh38
rs888010901 Chr.17: 44320510-44320510 on Build GRCh38
rs771522660 Chr.17: 44320448-44320448 on Build GRCh38
rs767141017 Chr.17: 44320293-44320293 on Build GRCh38
rs199627269 Chr.17: 44320289-44320289 on Build GRCh38
rs567892816 Chr.17: 44320678-44320678 on Build GRCh38
rs775542651 Chr.17: 44320290-44320290 on Build GRCh38
rs776375013 Chr.17: 44320655-44320655 on Build GRCh38
rs754094717 Chr.17: 44320712-44320712 on Build GRCh38
rs746042795 Chr.17: 44320406-44320406 on Build GRCh38
rs963708401 Chr.17: 44320470-44320470 on Build GRCh38
rs146456291 Chr.17: 44320591-44320591 on Build GRCh38
rs2011951 Chr.17: 44320684-44320684 on Build GRCh38
rs376573435 Chr.17: 44320677-44320677 on Build GRCh38
rs1021550910 Chr.17: 44320391-44320391 on Build GRCh38
rs958924472 Chr.17: 44320387-44320387 on Build GRCh38
rs754833890 Chr.17: 44320625-44320625 on Build GRCh38
rs143955536 Chr.17: 44320510-44320510 on Build GRCh38
rs759620735 Chr.17: 44320713-44320713 on Build GRCh38
rs756462816 Chr.17: 44320485-44320485 on Build GRCh38
rs932298946 Chr.17: 44320502-44320502 on Build GRCh38
rs764707540 Chr.17: 44320461-44320461 on Build GRCh38
rs779445834 Chr.17: 44320301-44320301 on Build GRCh38
rs781583332 Chr.17: 44320401-44320401 on Build GRCh38
rs763226052 Chr.17: 44320455-44320455 on Build GRCh38
rs770294785 Chr.17: 44320642-44320642 on Build GRCh38
rs761436942 Chr.17: 44320446-44320446 on Build GRCh38
rs111617263 Chr.17: 44320354-44320354 on Build GRCh38
rs192077377 Chr.17: 44320549-44320549 on Build GRCh38
rs991456876 Chr.17: 44320388-44320388 on Build GRCh38
rs780506604 Chr.17: 44320732-44320732 on Build GRCh38
rs745441194 Chr.17: 44320664-44320664 on Build GRCh38
rs1050674333 Chr.17: 44320506-44320506 on Build GRCh38
rs750595785 Chr.17: 44320617-44320617 on Build GRCh38
rs545826495 Chr.17: 44320453-44320453 on Build GRCh38
rs139586541 Chr.17: 44320390-44320390 on Build GRCh38
rs758442325 Chr.17: 44320311-44320311 on Build GRCh38
rs765488036 Chr.17: 44320716-44320716 on Build GRCh38
rs766673966 Chr.17: 44320622-44320622 on Build GRCh38
rs770639629 Chr.17: 44320582-44320582 on Build GRCh38
rs765546926 Chr.17: 44320380-44320380 on Build GRCh38
rs528148193 Chr.17: 44320490-44320490 on Build GRCh38
rs759917757 Chr.17: 44320379-44320379 on Build GRCh38
rs185939414 Chr.17: 44320679-44320679 on Build GRCh38
rs756230000 Chr.17: 44320620-44320620 on Build GRCh38
rs774923729 Chr.17: 44320595-44320595 on Build GRCh38
rs139114659 Chr.17: 44320292-44320292 on Build GRCh38
rs762387970 Chr.17: 44320291-44320291 on Build GRCh38
rs983738727 Chr.17: 44320751-44320751 on Build GRCh38
rs767200003 Chr.17: 44320371-44320371 on Build GRCh38
rs781531929 Chr.17: 44320327-44320327 on Build GRCh38
rs768124989 Chr.17: 44320601-44320601 on Build GRCh38
rs762351754 Chr.17: 44320600-44320600 on Build GRCh38
rs749146182 Chr.17: 44320487-44320487 on Build GRCh38
rs751455285 Chr.17: 44320727-44320727 on Build GRCh38
rs764322498 Chr.17: 44320592-44320592 on Build GRCh38
rs753154581 Chr.17: 44320303-44320303 on Build GRCh38
rs1006341620 Chr.17: 44320338-44320338 on Build GRCh38
rs778506151 Chr.17: 44320628-44320628 on Build GRCh38
rs962225758 Chr.17: 44320724-44320724 on Build GRCh38
rs548595895 Chr.17: 44320533-44320533 on Build GRCh38
rs745370067 Chr.17: 44320743-44320743 on Build GRCh38
rs773969174 Chr.17: 44320361-44320361 on Build GRCh38
rs113531238 Chr.17: 44320353-44320353 on Build GRCh38
rs774126352 Chr.17: 44320439-44320439 on Build GRCh38
rs968638098 Chr.17: 44320649-44320649 on Build GRCh38
rs545784339 Chr.17: 44320748-44320748 on Build GRCh38
rs758392171 Chr.17: 44320634-44320634 on Build GRCh38
rs377362658 Chr.17: 44320730-44320730 on Build GRCh38
rs755305686 Chr.17: 44320299-44320299 on Build GRCh38
rs552722507 Chr.17: 44320317-44320317 on Build GRCh38
rs769660133 Chr.17: 44320337-44320337 on Build GRCh38
rs186314042 Chr.17: 44320372-44320372 on Build GRCh38
rs770710274 Chr.17: 44320406-44320407 on Build GRCh38
rs757387684 Chr.17: 44320394-44320394 on Build GRCh38
rs371523676 Chr.17: 44320676-44320676 on Build GRCh38
rs758160607 Chr.17: 44320721-44320721 on Build GRCh38
rs775984976 Chr.17: 44320454-44320454 on Build GRCh38
rs201618124 Chr.17: 44320383-44320383 on Build GRCh38
rs745703466 Chr.17: 44320336-44320336 on Build GRCh38
rs769716055 Chr.17: 44320407-44320407 on Build GRCh38
rs903024515 Chr.17: 44320594-44320594 on Build GRCh38
rs962236024 Chr.17: 44320518-44320518 on Build GRCh38
rs372262650 Chr.17: 44320728-44320728 on Build GRCh38
rs574065353 Chr.17: 44320509-44320509 on Build GRCh38
rs1003345954 Chr.17: 44320369-44320369 on Build GRCh38
rs981663689 Chr.17: 44320527-44320527 on Build GRCh38
rs767634143 Chr.17: 44320475-44320475 on Build GRCh38
rs778600104 Chr.17: 44320624-44320625 on Build GRCh38
rs754269784 Chr.17: 44320295-44320295 on Build GRCh38
rs778933722 Chr.17: 44320488-44320488 on Build GRCh38
rs201972895 Chr.17: 44320682-44320682 on Build GRCh38
rs771245625 Chr.17: 44320322-44320322 on Build GRCh38
rs777845471 Chr.17: 44320315-44320315 on Build GRCh38
rs181608971 Chr.17: 44320590-44320590 on Build GRCh38
rs1003974412 Chr.17: 44320706-44320706 on Build GRCh38
rs200025174 Chr.17: 44320572-44320572 on Build GRCh38
rs750685874 Chr.17: 44320481-44320481 on Build GRCh38
rs778132147 Chr.17: 44320389-44320389 on Build GRCh38
rs937389171 Chr.17: 44320306-44320306 on Build GRCh38
rs149738856 Chr.17: 44320641-44320641 on Build GRCh38
rs748101051 Chr.17: 44320631-44320631 on Build GRCh38
rs772882946 Chr.17: 44320450-44320450 on Build GRCh38
rs757439199 Chr.17: 44320471-44320471 on Build GRCh38
rs749021391 Chr.17: 44320432-44320432 on Build GRCh38

View Full Product Details