Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32377878-32378374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSDC1
Entrez Gene ID
55108
Gene Name
BSD domain containing 1
Gene Aliases
-
UniGene
Hs.353454
Gene Chromosome Location
Chr.1: 32364633-32394461 on Build GRCh38
Transcripts
NM_001143888.2 NM_001143889.2 NM_001143890.2 NM_001300958.1 NM_018045.7 NR_125352.1 XM_005270986.4 XM_011541677.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6294482 c.649-3C>A p.? Chr.1: 32378051-32378051 on Build GRCh38
6294481 c.649-3C>A p.? Chr.1: 32378051-32378051 on Build GRCh38
6291966 c.580-7C>T p.? Chr.1: 32378290-32378290 on Build GRCh38
6291967 c.580-7C>T p.? Chr.1: 32378290-32378290 on Build GRCh38
4393861 c.707C>T p.P236L Chr.1: 32377990-32377990 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142016597 Chr.1: 32377938-32377938 on Build GRCh38
rs766366862 Chr.1: 32378228-32378228 on Build GRCh38
rs765230852 Chr.1: 32377980-32377980 on Build GRCh38
rs898470760 Chr.1: 32378038-32378038 on Build GRCh38
rs765632276 Chr.1: 32378181-32378181 on Build GRCh38
rs759508472 Chr.1: 32378087-32378087 on Build GRCh38
rs950620368 Chr.1: 32378084-32378084 on Build GRCh38
rs527539072 Chr.1: 32378300-32378300 on Build GRCh38
rs373402760 Chr.1: 32378322-32378322 on Build GRCh38
rs144703195 Chr.1: 32378013-32378013 on Build GRCh38
rs377336946 Chr.1: 32378294-32378294 on Build GRCh38
rs148827149 Chr.1: 32378035-32378035 on Build GRCh38
rs939935631 Chr.1: 32377906-32377906 on Build GRCh38
rs768625179 Chr.1: 32377960-32377960 on Build GRCh38
rs368456990 Chr.1: 32378000-32378000 on Build GRCh38
rs757648734 Chr.1: 32378018-32378018 on Build GRCh38
rs774279370 Chr.1: 32377987-32377987 on Build GRCh38
rs560058496 Chr.1: 32378289-32378289 on Build GRCh38
rs779532794 Chr.1: 32378027-32378027 on Build GRCh38
rs768458088 Chr.1: 32378048-32378048 on Build GRCh38
rs935732795 Chr.1: 32378313-32378313 on Build GRCh38
rs867979679 Chr.1: 32378254-32378254 on Build GRCh38
rs764013404 Chr.1: 32378292-32378293 on Build GRCh38
rs141409170 Chr.1: 32377988-32377988 on Build GRCh38
rs948367676 Chr.1: 32377930-32377930 on Build GRCh38
rs760803134 Chr.1: 32378287-32378287 on Build GRCh38
rs757406630 Chr.1: 32378313-32378314 on Build GRCh38
rs754233245 Chr.1: 32378017-32378017 on Build GRCh38
rs767690873 Chr.1: 32377989-32377989 on Build GRCh38
rs749183364 Chr.1: 32378260-32378260 on Build GRCh38
rs138894665 Chr.1: 32377922-32377922 on Build GRCh38
rs771617822 Chr.1: 32377928-32377928 on Build GRCh38
rs377007471 Chr.1: 32378085-32378085 on Build GRCh38
rs754661592 Chr.1: 32378223-32378223 on Build GRCh38
rs200234101 Chr.1: 32377937-32377938 on Build GRCh38
rs562590616 Chr.1: 32378135-32378135 on Build GRCh38
rs754014924 Chr.1: 32378296-32378297 on Build GRCh38
rs765592300 Chr.1: 32378293-32378293 on Build GRCh38
rs775577822 Chr.1: 32378094-32378094 on Build GRCh38
rs189557595 Chr.1: 32378071-32378071 on Build GRCh38
rs992779914 Chr.1: 32378196-32378196 on Build GRCh38
rs572772703 Chr.1: 32377927-32377927 on Build GRCh38
rs144566892 Chr.1: 32378014-32378014 on Build GRCh38
rs760817406 Chr.1: 32378164-32378164 on Build GRCh38
rs778231390 Chr.1: 32377925-32377925 on Build GRCh38
rs750992561 Chr.1: 32378182-32378182 on Build GRCh38
rs781158297 Chr.1: 32377950-32377950 on Build GRCh38
rs746614219 Chr.1: 32378044-32378044 on Build GRCh38
rs373057622 Chr.1: 32378233-32378233 on Build GRCh38
rs760674758 Chr.1: 32378062-32378063 on Build GRCh38
rs760700040 Chr.1: 32378082-32378082 on Build GRCh38
rs150963407 Chr.1: 32378227-32378227 on Build GRCh38
rs767010333 Chr.1: 32378333-32378333 on Build GRCh38
rs946986015 Chr.1: 32377961-32377961 on Build GRCh38
rs748031007 Chr.1: 32378070-32378070 on Build GRCh38
rs997211590 Chr.1: 32378089-32378089 on Build GRCh38
rs767537137 Chr.1: 32378093-32378093 on Build GRCh38
rs979728441 Chr.1: 32378237-32378237 on Build GRCh38
rs946372123 Chr.1: 32378314-32378314 on Build GRCh38
rs371990220 Chr.1: 32378031-32378031 on Build GRCh38
rs746773505 Chr.1: 32377952-32377952 on Build GRCh38
rs777450364 Chr.1: 32378247-32378247 on Build GRCh38
rs577793168 Chr.1: 32378184-32378184 on Build GRCh38
rs761617631 Chr.1: 32377967-32377967 on Build GRCh38
rs751121688 Chr.1: 32378220-32378220 on Build GRCh38
rs372312772 Chr.1: 32378025-32378025 on Build GRCh38
rs769503914 Chr.1: 32378315-32378315 on Build GRCh38
rs144659572 Chr.1: 32378280-32378280 on Build GRCh38
rs779835045 Chr.1: 32378039-32378039 on Build GRCh38
rs775383748 Chr.1: 32378022-32378023 on Build GRCh38
rs770858246 Chr.1: 32378267-32378267 on Build GRCh38
rs569264845 Chr.1: 32378312-32378312 on Build GRCh38
rs762062939 Chr.1: 32378291-32378291 on Build GRCh38
rs779673645 Chr.1: 32378185-32378185 on Build GRCh38
rs756507297 Chr.1: 32377924-32377924 on Build GRCh38
rs555357103 Chr.1: 32377948-32377948 on Build GRCh38
rs749708975 Chr.1: 32377926-32377926 on Build GRCh38
rs866514828 Chr.1: 32378271-32378271 on Build GRCh38
rs374773298 Chr.1: 32378177-32378177 on Build GRCh38
rs139279941 Chr.1: 32378005-32378005 on Build GRCh38
rs776504485 Chr.1: 32378057-32378057 on Build GRCh38
rs768863124 Chr.1: 32378288-32378288 on Build GRCh38
rs757708121 Chr.1: 32378179-32378179 on Build GRCh38
rs769109768 Chr.1: 32378101-32378101 on Build GRCh38

View Full Product Details