Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 35737244-35737787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLSPN
Entrez Gene ID
63967
Gene Name
claspin
Gene Aliases
-
UniGene
Hs.175613
Gene Chromosome Location
Chr.1: 35720465-35769985 on Build GRCh38
Transcripts
NM_001190481.1 NM_022111.3 XM_011541938.2 XM_011541939.2 XM_011541940.2 XM_011541941.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5977957 c.3707G>C p.R1236T Chr.1: 35737379-35737379 on Build GRCh38
4672923 c.3730C>T p.P1244S Chr.1: 35737356-35737356 on Build GRCh38
4672922 c.3730C>T p.P1244S Chr.1: 35737356-35737356 on Build GRCh38
5977958 c.3707G>C p.R1236T Chr.1: 35737379-35737379 on Build GRCh38
5095287 c.3671G>A p.R1224H Chr.1: 35737415-35737415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762470559 Chr.1: 35737697-35737697 on Build GRCh38
rs140581594 Chr.1: 35737410-35737410 on Build GRCh38
rs1021309485 Chr.1: 35737706-35737706 on Build GRCh38
rs569293388 Chr.1: 35737467-35737467 on Build GRCh38
rs752512972 Chr.1: 35737330-35737330 on Build GRCh38
rs749034696 Chr.1: 35737418-35737418 on Build GRCh38
rs35527076 Chr.1: 35737602-35737603 on Build GRCh38
rs900986071 Chr.1: 35737734-35737734 on Build GRCh38
rs755561116 Chr.1: 35737376-35737376 on Build GRCh38
rs370629561 Chr.1: 35737354-35737354 on Build GRCh38
rs752088207 Chr.1: 35737370-35737370 on Build GRCh38
rs770394532 Chr.1: 35737438-35737438 on Build GRCh38
rs1039793559 Chr.1: 35737539-35737539 on Build GRCh38
rs866750053 Chr.1: 35737400-35737400 on Build GRCh38
rs55647365 Chr.1: 35737415-35737415 on Build GRCh38
rs1009468208 Chr.1: 35737693-35737693 on Build GRCh38
rs759508861 Chr.1: 35737308-35737308 on Build GRCh38
rs748461366 Chr.1: 35737413-35737413 on Build GRCh38
rs756247926 Chr.1: 35737546-35737546 on Build GRCh38
rs773902235 Chr.1: 35737440-35737440 on Build GRCh38
rs113424263 Chr.1: 35737737-35737737 on Build GRCh38
rs547820791 Chr.1: 35737421-35737421 on Build GRCh38
rs962415871 Chr.1: 35737300-35737300 on Build GRCh38
rs759421276 Chr.1: 35737444-35737444 on Build GRCh38
rs1053789241 Chr.1: 35737639-35737639 on Build GRCh38
rs765971629 Chr.1: 35737299-35737299 on Build GRCh38
rs757337254 Chr.1: 35737323-35737323 on Build GRCh38
rs893833785 Chr.1: 35737666-35737666 on Build GRCh38
rs774265866 Chr.1: 35737305-35737305 on Build GRCh38
rs934583884 Chr.1: 35737390-35737390 on Build GRCh38
rs971248899 Chr.1: 35737334-35737334 on Build GRCh38
rs937955485 Chr.1: 35737625-35737625 on Build GRCh38
rs111988969 Chr.1: 35737493-35737493 on Build GRCh38
rs570570704 Chr.1: 35737650-35737650 on Build GRCh38
rs757462389 Chr.1: 35737349-35737349 on Build GRCh38
rs1017019994 Chr.1: 35737298-35737298 on Build GRCh38
rs201797606 Chr.1: 35737503-35737503 on Build GRCh38
rs962402801 Chr.1: 35737757-35737757 on Build GRCh38
rs879216434 Chr.1: 35737403-35737403 on Build GRCh38
rs536778268 Chr.1: 35737518-35737518 on Build GRCh38
rs915738741 Chr.1: 35737326-35737326 on Build GRCh38
rs547782607 Chr.1: 35737362-35737362 on Build GRCh38
rs777276992 Chr.1: 35737416-35737416 on Build GRCh38
rs56311745 Chr.1: 35737364-35737364 on Build GRCh38
rs1016597937 Chr.1: 35737753-35737753 on Build GRCh38
rs779945185 Chr.1: 35737720-35737720 on Build GRCh38
rs899995164 Chr.1: 35737550-35737550 on Build GRCh38
rs375983992 Chr.1: 35737325-35737325 on Build GRCh38
rs772029559 Chr.1: 35737459-35737459 on Build GRCh38
rs551827393 Chr.1: 35737544-35737544 on Build GRCh38
rs998343280 Chr.1: 35737560-35737560 on Build GRCh38
rs909581143 Chr.1: 35737363-35737363 on Build GRCh38
rs532703088 Chr.1: 35737285-35737285 on Build GRCh38
rs750463495 Chr.1: 35737358-35737358 on Build GRCh38

View Full Product Details