Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63477236-63477744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACE
Entrez Gene ID
1636
Gene Name
angiotensin I converting enzyme
Gene Aliases
ACE1, CD143, DCP, DCP1, ICH, MVCD3
UniGene
Hs.298469
Gene Chromosome Location
Chr.17: 63477061-63498380 on Build GRCh38
Transcripts
NM_000789.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7318147 c.214G>A p.D72N Chr.17: 63477308-63477308 on Build GRCh38
707739 c.221A>C p.N74T Chr.17: 63477315-63477315 on Build GRCh38
7194293 c.201C>T p.A67A Chr.17: 63477295-63477295 on Build GRCh38
6429826 c.205T>C p.W69R Chr.17: 63477299-63477299 on Build GRCh38
7318146 c.214G>A p.D72N Chr.17: 63477308-63477308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748655390 Chr.17: 63477525-63477525 on Build GRCh38
rs1033534767 Chr.17: 63477403-63477403 on Build GRCh38
rs561937787 Chr.17: 63477532-63477532 on Build GRCh38
rs769922375 Chr.17: 63477304-63477304 on Build GRCh38
rs997125723 Chr.17: 63477339-63477339 on Build GRCh38
rs1051130124 Chr.17: 63477623-63477623 on Build GRCh38
rs527828087 Chr.17: 63477534-63477534 on Build GRCh38
rs1001051825 Chr.17: 63477712-63477712 on Build GRCh38
rs773257897 Chr.17: 63477305-63477305 on Build GRCh38
rs1009813622 Chr.17: 63477646-63477646 on Build GRCh38
rs747292160 Chr.17: 63477284-63477284 on Build GRCh38
rs372565955 Chr.17: 63477303-63477303 on Build GRCh38
rs1042619812 Chr.17: 63477666-63477666 on Build GRCh38
rs569066804 Chr.17: 63477697-63477697 on Build GRCh38
rs959531460 Chr.17: 63477416-63477416 on Build GRCh38
rs1051387451 Chr.17: 63477490-63477490 on Build GRCh38
rs576632634 Chr.17: 63477459-63477459 on Build GRCh38
rs781610217 Chr.17: 63477353-63477353 on Build GRCh38
rs7211207 Chr.17: 63477552-63477552 on Build GRCh38
rs752559637 Chr.17: 63477308-63477308 on Build GRCh38
rs868424949 Chr.17: 63477282-63477282 on Build GRCh38
rs749318099 Chr.17: 63477718-63477718 on Build GRCh38
rs891389285 Chr.17: 63477492-63477492 on Build GRCh38
rs950425419 Chr.17: 63477442-63477442 on Build GRCh38
rs1009655068 Chr.17: 63477361-63477361 on Build GRCh38
rs1021013249 Chr.17: 63477363-63477364 on Build GRCh38
rs968169000 Chr.17: 63477369-63477369 on Build GRCh38
rs184457276 Chr.17: 63477325-63477325 on Build GRCh38
rs866105888 Chr.17: 63477266-63477266 on Build GRCh38
rs760330379 Chr.17: 63477316-63477316 on Build GRCh38
rs921377643 Chr.17: 63477609-63477609 on Build GRCh38
rs867626302 Chr.17: 63477323-63477323 on Build GRCh38
rs780601919 Chr.17: 63477283-63477283 on Build GRCh38
rs868323704 Chr.17: 63477293-63477293 on Build GRCh38
rs867174380 Chr.17: 63477300-63477300 on Build GRCh38
rs545751216 Chr.17: 63477400-63477400 on Build GRCh38
rs779520756 Chr.17: 63477336-63477336 on Build GRCh38
rs181234076 Chr.17: 63477472-63477472 on Build GRCh38
rs903911203 Chr.17: 63477563-63477563 on Build GRCh38
rs189334234 Chr.17: 63477446-63477446 on Build GRCh38
rs1047503443 Chr.17: 63477289-63477289 on Build GRCh38
rs538128723 Chr.17: 63477721-63477721 on Build GRCh38
rs149083340 Chr.17: 63477627-63477627 on Build GRCh38
rs867394500 Chr.17: 63477301-63477301 on Build GRCh38
rs778615098 Chr.17: 63477326-63477326 on Build GRCh38
rs776943620 Chr.17: 63477287-63477287 on Build GRCh38
rs147912715 Chr.17: 63477271-63477271 on Build GRCh38
rs773409599 Chr.17: 63477505-63477505 on Build GRCh38
rs992062827 Chr.17: 63477420-63477420 on Build GRCh38
rs745572589 Chr.17: 63477276-63477276 on Build GRCh38
rs576744905 Chr.17: 63477356-63477356 on Build GRCh38
rs768833386 Chr.17: 63477348-63477348 on Build GRCh38
rs375602836 Chr.17: 63477317-63477317 on Build GRCh38
rs753783787 Chr.17: 63477322-63477322 on Build GRCh38
rs1029656360 Chr.17: 63477350-63477355 on Build GRCh38
rs747410284 Chr.17: 63477343-63477343 on Build GRCh38
rs868134438 Chr.17: 63477269-63477269 on Build GRCh38
rs932882408 Chr.17: 63477486-63477486 on Build GRCh38
rs983081002 Chr.17: 63477451-63477451 on Build GRCh38
rs908920716 Chr.17: 63477457-63477462 on Build GRCh38
rs868605010 Chr.17: 63477262-63477262 on Build GRCh38
rs887280103 Chr.17: 63477294-63477294 on Build GRCh38
rs766548114 Chr.17: 63477307-63477307 on Build GRCh38
rs886053219 Chr.17: 63477258-63477258 on Build GRCh38
rs769225220 Chr.17: 63477285-63477285 on Build GRCh38
rs771828535 Chr.17: 63477280-63477280 on Build GRCh38
rs929042659 Chr.17: 63477275-63477275 on Build GRCh38
rs749921316 Chr.17: 63477330-63477330 on Build GRCh38
rs370200820 Chr.17: 63477334-63477334 on Build GRCh38
rs917574425 Chr.17: 63477432-63477432 on Build GRCh38
rs755617014 Chr.17: 63477364-63477364 on Build GRCh38
rs547466255 Chr.17: 63477550-63477550 on Build GRCh38
rs533192764 Chr.17: 63477611-63477611 on Build GRCh38
rs1000979672 Chr.17: 63477568-63477568 on Build GRCh38
rs1038833692 Chr.17: 63477559-63477559 on Build GRCh38

View Full Product Details