Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45267318-45267810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP3K14
Entrez Gene ID
9020
Gene Name
mitogen-activated protein kinase kinase kinase 14
Gene Aliases
FTDCR1B, HS, HSNIK, NIK
UniGene
Hs.404183
Gene Chromosome Location
Chr.17: 45263119-45317064 on Build GRCh38
Transcripts
NM_003954.4 XM_011525441.2 NR_024434.2 NR_024435.2 NR_110324.1 NR_110325.1 NR_110326.1

Gene Symbol
MAP3K14-AS1
Entrez Gene ID
100133991
Gene Name
MAP3K14 antisense RNA 1
Gene Aliases
-
UniGene
Hs.668927
Gene Chromosome Location
Chr.17: 45247925-45268630 on Build GRCh38
Transcripts
NM_003954.4 XM_011525441.2 NR_024434.2 NR_024435.2 NR_110324.1 NR_110325.1 NR_110326.1

SNP Detail

SNP ID Chromosome Location
rs1032715573 Chr.17: 45267631-45267631 on Build GRCh38
rs759696785 Chr.17: 45267705-45267705 on Build GRCh38
rs148416800 Chr.17: 45267420-45267420 on Build GRCh38
rs748655204 Chr.17: 45267563-45267563 on Build GRCh38
rs746619908 Chr.17: 45267515-45267515 on Build GRCh38
rs769256349 Chr.17: 45267554-45267554 on Build GRCh38
rs773403567 Chr.17: 45267466-45267466 on Build GRCh38
rs944938258 Chr.17: 45267551-45267551 on Build GRCh38
rs34097542 Chr.17: 45267738-45267739 on Build GRCh38
rs372554542 Chr.17: 45267629-45267629 on Build GRCh38
rs17846846 Chr.17: 45267434-45267434 on Build GRCh38
rs538625946 Chr.17: 45267660-45267660 on Build GRCh38
rs745358592 Chr.17: 45267552-45267552 on Build GRCh38
rs750712807 Chr.17: 45267415-45267415 on Build GRCh38
rs757176083 Chr.17: 45267633-45267633 on Build GRCh38
rs778057997 Chr.17: 45267676-45267676 on Build GRCh38
rs753923383 Chr.17: 45267494-45267494 on Build GRCh38
rs1003435070 Chr.17: 45267741-45267741 on Build GRCh38
rs756158025 Chr.17: 45267417-45267417 on Build GRCh38
rs370000467 Chr.17: 45267663-45267663 on Build GRCh38
rs554658901 Chr.17: 45267344-45267344 on Build GRCh38
rs144935730 Chr.17: 45267550-45267551 on Build GRCh38
rs376080956 Chr.17: 45267781-45267781 on Build GRCh38
rs772435508 Chr.17: 45267571-45267571 on Build GRCh38
rs767767533 Chr.17: 45267409-45267409 on Build GRCh38
rs375783480 Chr.17: 45267693-45267693 on Build GRCh38
rs755169552 Chr.17: 45267636-45267636 on Build GRCh38
rs957112717 Chr.17: 45267385-45267385 on Build GRCh38
rs144165828 Chr.17: 45267347-45267347 on Build GRCh38
rs376749009 Chr.17: 45267548-45267548 on Build GRCh38
rs749745409 Chr.17: 45267446-45267446 on Build GRCh38
rs199865460 Chr.17: 45267738-45267738 on Build GRCh38
rs765212451 Chr.17: 45267498-45267498 on Build GRCh38
rs377070363 Chr.17: 45267393-45267393 on Build GRCh38
rs755630489 Chr.17: 45267651-45267651 on Build GRCh38
rs549267740 Chr.17: 45267491-45267491 on Build GRCh38
rs763575771 Chr.17: 45267376-45267376 on Build GRCh38
rs1035368490 Chr.17: 45267742-45267742 on Build GRCh38
rs763722669 Chr.17: 45267378-45267379 on Build GRCh38
rs138915793 Chr.17: 45267737-45267737 on Build GRCh38
rs11867907 Chr.17: 45267712-45267712 on Build GRCh38
rs371521847 Chr.17: 45267450-45267450 on Build GRCh38
rs151296471 Chr.17: 45267443-45267443 on Build GRCh38
rs146770225 Chr.17: 45267673-45267674 on Build GRCh38
rs780159331 Chr.17: 45267419-45267419 on Build GRCh38
rs756974116 Chr.17: 45267545-45267545 on Build GRCh38
rs771322208 Chr.17: 45267690-45267690 on Build GRCh38
rs543502251 Chr.17: 45267351-45267351 on Build GRCh38
rs150489166 Chr.17: 45267471-45267471 on Build GRCh38
rs56302559 Chr.17: 45267442-45267442 on Build GRCh38
rs777450057 Chr.17: 45267507-45267507 on Build GRCh38
rs369529527 Chr.17: 45267699-45267699 on Build GRCh38
rs938141306 Chr.17: 45267455-45267455 on Build GRCh38
rs139936905 Chr.17: 45267571-45267572 on Build GRCh38
rs56907763 Chr.17: 45267402-45267402 on Build GRCh38
rs766570512 Chr.17: 45267583-45267583 on Build GRCh38
rs748652165 Chr.17: 45267662-45267662 on Build GRCh38
rs751614619 Chr.17: 45267779-45267779 on Build GRCh38
rs143567739 Chr.17: 45267751-45267751 on Build GRCh38
rs892790739 Chr.17: 45267678-45267678 on Build GRCh38
rs775435880 Chr.17: 45267723-45267723 on Build GRCh38
rs752614062 Chr.17: 45267500-45267500 on Build GRCh38
rs1023427255 Chr.17: 45267704-45267704 on Build GRCh38
rs760823535 Chr.17: 45267581-45267581 on Build GRCh38
rs1005375658 Chr.17: 45267360-45267360 on Build GRCh38
rs747366448 Chr.17: 45267686-45267686 on Build GRCh38
rs758188422 Chr.17: 45267506-45267506 on Build GRCh38
rs955204029 Chr.17: 45267758-45267758 on Build GRCh38
rs751958888 Chr.17: 45267394-45267394 on Build GRCh38
rs762235528 Chr.17: 45267457-45267457 on Build GRCh38
rs986607520 Chr.17: 45267770-45267770 on Build GRCh38
rs998510224 Chr.17: 45267352-45267352 on Build GRCh38
rs576007466 Chr.17: 45267349-45267349 on Build GRCh38
rs202201840 Chr.17: 45267585-45267585 on Build GRCh38
rs149303541 Chr.17: 45267616-45267616 on Build GRCh38
rs774779223 Chr.17: 45267451-45267451 on Build GRCh38
rs148027547 Chr.17: 45267763-45267763 on Build GRCh38
rs746706058 Chr.17: 45267440-45267440 on Build GRCh38
rs149897421 Chr.17: 45267539-45267540 on Build GRCh38
rs751413709 Chr.17: 45267630-45267630 on Build GRCh38
rs55828586 Chr.17: 45267650-45267650 on Build GRCh38
rs140631051 Chr.17: 45267454-45267454 on Build GRCh38
rs776117126 Chr.17: 45267374-45267374 on Build GRCh38
rs752629082 Chr.17: 45267604-45267604 on Build GRCh38
rs771825847 Chr.17: 45267438-45267438 on Build GRCh38
rs375277208 Chr.17: 45267490-45267490 on Build GRCh38
rs773601828 Chr.17: 45267580-45267580 on Build GRCh38
rs774893057 Chr.17: 45267559-45267559 on Build GRCh38
rs373775810 Chr.17: 45267425-45267425 on Build GRCh38
rs777448729 Chr.17: 45267439-45267439 on Build GRCh38
rs367962948 Chr.17: 45267601-45267601 on Build GRCh38
rs536010742 Chr.17: 45267343-45267343 on Build GRCh38
rs376396491 Chr.17: 45267642-45267642 on Build GRCh38
rs759626619 Chr.17: 45267595-45267595 on Build GRCh38
rs143365100 Chr.17: 45267635-45267636 on Build GRCh38
rs141230125 Chr.17: 45267600-45267600 on Build GRCh38
rs146625889 Chr.17: 45267482-45267482 on Build GRCh38
rs747959258 Chr.17: 45267435-45267435 on Build GRCh38
rs767814621 Chr.17: 45267462-45267462 on Build GRCh38
rs142428947 Chr.17: 45267431-45267431 on Build GRCh38
rs35362441 Chr.17: 45267505-45267506 on Build GRCh38
rs149375313 Chr.17: 45267739-45267739 on Build GRCh38
rs147799978 Chr.17: 45267406-45267406 on Build GRCh38
rs1023921026 Chr.17: 45267381-45267381 on Build GRCh38
rs373213575 Chr.17: 45267782-45267782 on Build GRCh38
rs571831529 Chr.17: 45267495-45267495 on Build GRCh38

View Full Product Details