Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48614192-48614700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXB8
Entrez Gene ID
3218
Gene Name
homeobox B8
Gene Aliases
HOX2, HOX2D, Hox-2.4
UniGene
Hs.514292
Gene Chromosome Location
Chr.17: 48612346-48622853 on Build GRCh38
Transcripts
NM_024016.3 XM_005257286.2 XM_017024564.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6207177 c.153G>A p.P51P Chr.17: 48614552-48614552 on Build GRCh38
3518746 c.356G>A p.G119D Chr.17: 48614349-48614349 on Build GRCh38
6147388 c.89A>C p.Q30P Chr.17: 48614616-48614616 on Build GRCh38
6081575 c.133G>T p.G45C Chr.17: 48614572-48614572 on Build GRCh38
4067395 c.302A>G p.D101G Chr.17: 48614403-48614403 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745337132 Chr.17: 48614435-48614436 on Build GRCh38
rs978622495 Chr.17: 48614426-48614426 on Build GRCh38
rs771441367 Chr.17: 48614328-48614328 on Build GRCh38
rs777427813 Chr.17: 48614519-48614519 on Build GRCh38
rs905371739 Chr.17: 48614633-48614635 on Build GRCh38
rs1047300311 Chr.17: 48614590-48614590 on Build GRCh38
rs747706359 Chr.17: 48614408-48614408 on Build GRCh38
rs759929773 Chr.17: 48614335-48614335 on Build GRCh38
rs962027322 Chr.17: 48614555-48614556 on Build GRCh38
rs780049802 Chr.17: 48614395-48614395 on Build GRCh38
rs909227307 Chr.17: 48614226-48614227 on Build GRCh38
rs774570857 Chr.17: 48614598-48614598 on Build GRCh38
rs768407252 Chr.17: 48614313-48614313 on Build GRCh38
rs747697211 Chr.17: 48614322-48614322 on Build GRCh38
rs757384780 Chr.17: 48614367-48614367 on Build GRCh38
rs766421679 Chr.17: 48614475-48614475 on Build GRCh38
rs775910991 Chr.17: 48614437-48614437 on Build GRCh38
rs756203173 Chr.17: 48614459-48614459 on Build GRCh38
rs771660620 Chr.17: 48614409-48614409 on Build GRCh38
rs778737872 Chr.17: 48614406-48614406 on Build GRCh38
rs777889597 Chr.17: 48614263-48614263 on Build GRCh38
rs760867249 Chr.17: 48614619-48614619 on Build GRCh38
rs751653877 Chr.17: 48614265-48614265 on Build GRCh38
rs573071628 Chr.17: 48614339-48614339 on Build GRCh38
rs751166855 Chr.17: 48614670-48614670 on Build GRCh38
rs530515759 Chr.17: 48614391-48614391 on Build GRCh38
rs150150478 Chr.17: 48614458-48614458 on Build GRCh38
rs906478504 Chr.17: 48614417-48614417 on Build GRCh38
rs778550819 Chr.17: 48614505-48614505 on Build GRCh38
rs901140674 Chr.17: 48614534-48614534 on Build GRCh38
rs751078920 Chr.17: 48614528-48614528 on Build GRCh38
rs766921338 Chr.17: 48614235-48614235 on Build GRCh38
rs775821122 Chr.17: 48614342-48614342 on Build GRCh38
rs912830988 Chr.17: 48614216-48614216 on Build GRCh38
rs2303484 Chr.17: 48614447-48614447 on Build GRCh38
rs754841389 Chr.17: 48614486-48614486 on Build GRCh38
rs576664763 Chr.17: 48614244-48614244 on Build GRCh38
rs199840850 Chr.17: 48614350-48614350 on Build GRCh38
rs202069371 Chr.17: 48614628-48614628 on Build GRCh38
rs199877083 Chr.17: 48614612-48614612 on Build GRCh38
rs752956744 Chr.17: 48614247-48614247 on Build GRCh38
rs762149861 Chr.17: 48614599-48614599 on Build GRCh38
rs200695320 Chr.17: 48614333-48614333 on Build GRCh38
rs546892177 Chr.17: 48614593-48614593 on Build GRCh38
rs769053052 Chr.17: 48614564-48614564 on Build GRCh38
rs757221567 Chr.17: 48614267-48614267 on Build GRCh38
rs112978726 Chr.17: 48614402-48614402 on Build GRCh38
rs745749118 Chr.17: 48614273-48614273 on Build GRCh38
rs770390441 Chr.17: 48614552-48614552 on Build GRCh38
rs763342496 Chr.17: 48614440-48614440 on Build GRCh38
rs772593397 Chr.17: 48614412-48614412 on Build GRCh38
rs747874938 Chr.17: 48614512-48614512 on Build GRCh38
rs766516844 Chr.17: 48614621-48614621 on Build GRCh38
rs545574189 Chr.17: 48614262-48614262 on Build GRCh38
rs992702927 Chr.17: 48614283-48614283 on Build GRCh38
rs942777417 Chr.17: 48614456-48614456 on Build GRCh38
rs1024878375 Chr.17: 48614365-48614365 on Build GRCh38
rs370314418 Chr.17: 48614660-48614660 on Build GRCh38
rs568947604 Chr.17: 48614650-48614650 on Build GRCh38
rs61737568 Chr.17: 48614356-48614356 on Build GRCh38
rs866955706 Chr.17: 48614386-48614386 on Build GRCh38
rs764396578 Chr.17: 48614441-48614441 on Build GRCh38
rs777216868 Chr.17: 48614330-48614330 on Build GRCh38
rs537994443 Chr.17: 48614239-48614239 on Build GRCh38
rs112987208 Chr.17: 48614225-48614226 on Build GRCh38
rs201120876 Chr.17: 48614507-48614507 on Build GRCh38
rs986062551 Chr.17: 48614657-48614657 on Build GRCh38
rs541171369 Chr.17: 48614218-48614219 on Build GRCh38
rs749106433 Chr.17: 48614401-48614401 on Build GRCh38
rs989206063 Chr.17: 48614541-48614541 on Build GRCh38
rs749777413 Chr.17: 48614556-48614556 on Build GRCh38
rs1022189335 Chr.17: 48614594-48614594 on Build GRCh38
rs752597921 Chr.17: 48614472-48614472 on Build GRCh38
rs549646264 Chr.17: 48614646-48614646 on Build GRCh38
rs11267100 Chr.17: 48614229-48614230 on Build GRCh38
rs774687021 Chr.17: 48614443-48614443 on Build GRCh38
rs781223185 Chr.17: 48614271-48614271 on Build GRCh38
rs774086334 Chr.17: 48614315-48614315 on Build GRCh38
rs749011584 Chr.17: 48614311-48614311 on Build GRCh38
rs767475775 Chr.17: 48614382-48614382 on Build GRCh38
rs779959955 Chr.17: 48614303-48614303 on Build GRCh38
rs750410378 Chr.17: 48614393-48614393 on Build GRCh38
rs557823107 Chr.17: 48614243-48614243 on Build GRCh38
rs140795156 Chr.17: 48614436-48614436 on Build GRCh38
rs374945607 Chr.17: 48614513-48614513 on Build GRCh38
rs7406365 Chr.17: 48614225-48614225 on Build GRCh38
rs746444993 Chr.17: 48614416-48614416 on Build GRCh38
rs202042736 Chr.17: 48614363-48614363 on Build GRCh38
rs1034515299 Chr.17: 48614669-48614669 on Build GRCh38
rs752448777 Chr.17: 48614647-48614647 on Build GRCh38
rs753785842 Chr.17: 48614482-48614482 on Build GRCh38
rs1048674518 Chr.17: 48614299-48614299 on Build GRCh38
rs769683083 Chr.17: 48614293-48614293 on Build GRCh38
rs1050030331 Chr.17: 48614212-48614212 on Build GRCh38
rs770300550 Chr.17: 48614435-48614435 on Build GRCh38
rs761913780 Chr.17: 48614445-48614445 on Build GRCh38
rs1033989746 Chr.17: 48614256-48614256 on Build GRCh38

View Full Product Details