Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 55720998-55721492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM100
Entrez Gene ID
55273
Gene Name
transmembrane protein 100
Gene Aliases
-
UniGene
Hs.173233
Gene Chromosome Location
Chr.17: 55719627-55732121 on Build GRCh38
Transcripts
NM_001099640.1 NM_018286.2 XM_017024814.1 XM_017024815.1 XM_017024816.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6054691 c.73C>A p.P25T Chr.17: 55720998-55720998 on Build GRCh38
460286 c.24G>A p.E8E Chr.17: 55721047-55721047 on Build GRCh38
5954569 c.24G>C p.E8D Chr.17: 55721047-55721047 on Build GRCh38
1303080 c.37C>T p.P13S Chr.17: 55721034-55721034 on Build GRCh38
366242 c.54A>G p.A18A Chr.17: 55721017-55721017 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs796116488 Chr.17: 55721197-55721197 on Build GRCh38
rs546033612 Chr.17: 55721238-55721238 on Build GRCh38
rs752839114 Chr.17: 55721096-55721096 on Build GRCh38
rs752286152 Chr.17: 55721106-55721106 on Build GRCh38
rs905700433 Chr.17: 55721119-55721119 on Build GRCh38
rs767878375 Chr.17: 55721442-55721442 on Build GRCh38
rs895513768 Chr.17: 55721076-55721076 on Build GRCh38
rs968372092 Chr.17: 55721127-55721127 on Build GRCh38
rs985555981 Chr.17: 55721368-55721368 on Build GRCh38
rs780433767 Chr.17: 55721409-55721409 on Build GRCh38
rs759255007 Chr.17: 55721080-55721080 on Build GRCh38
rs994105424 Chr.17: 55721185-55721185 on Build GRCh38
rs779284458 Chr.17: 55721028-55721028 on Build GRCh38
rs968862601 Chr.17: 55721415-55721415 on Build GRCh38
rs529219754 Chr.17: 55721428-55721428 on Build GRCh38
rs955612058 Chr.17: 55721352-55721352 on Build GRCh38
rs879076398 Chr.17: 55721066-55721066 on Build GRCh38
rs137934934 Chr.17: 55721236-55721236 on Build GRCh38
rs769410353 Chr.17: 55721041-55721041 on Build GRCh38
rs1004250445 Chr.17: 55721313-55721313 on Build GRCh38
rs377523122 Chr.17: 55721099-55721099 on Build GRCh38
rs368993073 Chr.17: 55721046-55721046 on Build GRCh38
rs977094230 Chr.17: 55721437-55721437 on Build GRCh38
rs924259641 Chr.17: 55721461-55721461 on Build GRCh38
rs143402351 Chr.17: 55721371-55721371 on Build GRCh38
rs768283137 Chr.17: 55721035-55721035 on Build GRCh38
rs888453992 Chr.17: 55721296-55721296 on Build GRCh38
rs561722925 Chr.17: 55721394-55721394 on Build GRCh38
rs772892040 Chr.17: 55721044-55721044 on Build GRCh38
rs961290230 Chr.17: 55721184-55721184 on Build GRCh38
rs117852529 Chr.17: 55721203-55721203 on Build GRCh38
rs1014815102 Chr.17: 55721050-55721050 on Build GRCh38
rs367641446 Chr.17: 55721135-55721135 on Build GRCh38
rs760927413 Chr.17: 55721122-55721122 on Build GRCh38
rs79554922 Chr.17: 55721079-55721079 on Build GRCh38
rs182021312 Chr.17: 55721404-55721404 on Build GRCh38
rs143975703 Chr.17: 55721377-55721377 on Build GRCh38
rs938932719 Chr.17: 55721183-55721183 on Build GRCh38
rs750680505 Chr.17: 55721378-55721378 on Build GRCh38
rs911337135 Chr.17: 55721372-55721372 on Build GRCh38
rs771017116 Chr.17: 55721062-55721062 on Build GRCh38
rs1007724041 Chr.17: 55721392-55721392 on Build GRCh38
rs1016794940 Chr.17: 55721159-55721159 on Build GRCh38
rs916913923 Chr.17: 55721414-55721414 on Build GRCh38
rs767413875 Chr.17: 55721095-55721095 on Build GRCh38
rs189103262 Chr.17: 55721374-55721374 on Build GRCh38
rs757362112 Chr.17: 55721333-55721333 on Build GRCh38
rs780700702 Chr.17: 55721037-55721037 on Build GRCh38
rs897770456 Chr.17: 55721363-55721363 on Build GRCh38
rs914240324 Chr.17: 55721107-55721107 on Build GRCh38
rs961947688 Chr.17: 55721055-55721055 on Build GRCh38

View Full Product Details