Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76155484-76155990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF157
Entrez Gene ID
114804
Gene Name
ring finger protein 157
Gene Aliases
-
UniGene
Hs.500643
Gene Chromosome Location
Chr.17: 76142453-76240368 on Build GRCh38
Transcripts
NM_052916.2 XM_005257007.4 XM_017024117.1 XM_017024118.1 XM_017024119.1 XM_017024120.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4130650 c.1685C>A p.S562* Chr.17: 76155575-76155575 on Build GRCh38
3890473 c.1607C>T p.S536F Chr.17: 76155653-76155653 on Build GRCh38
3522495 c.1596C>T p.V532V Chr.17: 76155664-76155664 on Build GRCh38
5439120 c.1639G>T p.E547* Chr.17: 76155621-76155621 on Build GRCh38
3522493 c.1638G>A p.E546E Chr.17: 76155622-76155622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529020860 Chr.17: 76155676-76155676 on Build GRCh38
rs199974386 Chr.17: 76155689-76155689 on Build GRCh38
rs762543408 Chr.17: 76155704-76155704 on Build GRCh38
rs375411739 Chr.17: 76155667-76155667 on Build GRCh38
rs372761404 Chr.17: 76155548-76155548 on Build GRCh38
rs763672842 Chr.17: 76155516-76155516 on Build GRCh38
rs937165405 Chr.17: 76155955-76155955 on Build GRCh38
rs762426180 Chr.17: 76155567-76155569 on Build GRCh38
rs966314231 Chr.17: 76155764-76155764 on Build GRCh38
rs772746027 Chr.17: 76155550-76155550 on Build GRCh38
rs764901300 Chr.17: 76155693-76155693 on Build GRCh38
rs150633713 Chr.17: 76155610-76155610 on Build GRCh38
rs753486491 Chr.17: 76155607-76155607 on Build GRCh38
rs749433492 Chr.17: 76155577-76155577 on Build GRCh38
rs1049104967 Chr.17: 76155530-76155530 on Build GRCh38
rs758500056 Chr.17: 76155779-76155779 on Build GRCh38
rs749486875 Chr.17: 76155650-76155650 on Build GRCh38
rs188463051 Chr.17: 76155854-76155854 on Build GRCh38
rs918244270 Chr.17: 76155809-76155809 on Build GRCh38
rs759986748 Chr.17: 76155582-76155582 on Build GRCh38
rs767929929 Chr.17: 76155769-76155769 on Build GRCh38
rs1055721673 Chr.17: 76155628-76155628 on Build GRCh38
rs745994048 Chr.17: 76155761-76155761 on Build GRCh38
rs754858020 Chr.17: 76155729-76155729 on Build GRCh38
rs767394977 Chr.17: 76155730-76155730 on Build GRCh38
rs1056420965 Chr.17: 76155548-76155549 on Build GRCh38
rs376187556 Chr.17: 76155579-76155579 on Build GRCh38
rs752503395 Chr.17: 76155746-76155746 on Build GRCh38
rs984841704 Chr.17: 76155846-76155846 on Build GRCh38
rs775841441 Chr.17: 76155588-76155588 on Build GRCh38
rs758260386 Chr.17: 76155541-76155541 on Build GRCh38
rs1019166181 Chr.17: 76155753-76155753 on Build GRCh38
rs79015157 Chr.17: 76155523-76155523 on Build GRCh38
rs143758012 Chr.17: 76155686-76155686 on Build GRCh38
rs774958836 Chr.17: 76155581-76155581 on Build GRCh38
rs376594896 Chr.17: 76155550-76155550 on Build GRCh38
rs779733866 Chr.17: 76155544-76155544 on Build GRCh38
rs1019225498 Chr.17: 76155719-76155719 on Build GRCh38
rs778978418 Chr.17: 76155533-76155533 on Build GRCh38
rs148853520 Chr.17: 76155914-76155914 on Build GRCh38
rs757149605 Chr.17: 76155862-76155862 on Build GRCh38
rs772594167 Chr.17: 76155583-76155583 on Build GRCh38
rs571209335 Chr.17: 76155908-76155908 on Build GRCh38
rs373845100 Chr.17: 76155747-76155747 on Build GRCh38
rs377719776 Chr.17: 76155618-76155618 on Build GRCh38
rs766478237 Chr.17: 76155713-76155713 on Build GRCh38
rs749897259 Chr.17: 76155615-76155615 on Build GRCh38
rs918369418 Chr.17: 76155557-76155557 on Build GRCh38
rs754802777 Chr.17: 76155646-76155646 on Build GRCh38
rs772601518 Chr.17: 76155696-76155696 on Build GRCh38
rs773199842 Chr.17: 76155561-76155561 on Build GRCh38
rs776916706 Chr.17: 76155622-76155622 on Build GRCh38
rs369511766 Chr.17: 76155547-76155547 on Build GRCh38
rs139884477 Chr.17: 76155666-76155666 on Build GRCh38
rs144982425 Chr.17: 76155919-76155919 on Build GRCh38
rs559937445 Chr.17: 76155655-76155655 on Build GRCh38
rs888292125 Chr.17: 76155600-76155600 on Build GRCh38
rs536793386 Chr.17: 76155946-76155946 on Build GRCh38
rs182175065 Chr.17: 76155513-76155513 on Build GRCh38
rs756746320 Chr.17: 76155528-76155528 on Build GRCh38
rs751534417 Chr.17: 76155715-76155715 on Build GRCh38
rs778204656 Chr.17: 76155647-76155647 on Build GRCh38
rs770908155 Chr.17: 76155580-76155580 on Build GRCh38
rs903663905 Chr.17: 76155598-76155598 on Build GRCh38
rs779143175 Chr.17: 76155750-76155750 on Build GRCh38
rs751041051 Chr.17: 76155836-76155836 on Build GRCh38
rs201646108 Chr.17: 76155639-76155639 on Build GRCh38
rs896385751 Chr.17: 76155566-76155566 on Build GRCh38
rs751409766 Chr.17: 76155640-76155640 on Build GRCh38
rs747501787 Chr.17: 76155595-76155595 on Build GRCh38
rs762466677 Chr.17: 76155712-76155712 on Build GRCh38
rs763795110 Chr.17: 76155597-76155597 on Build GRCh38
rs537944091 Chr.17: 76155837-76155837 on Build GRCh38
rs765637183 Chr.17: 76155657-76155657 on Build GRCh38
rs972319860 Chr.17: 76155774-76155774 on Build GRCh38
rs778079820 Chr.17: 76155562-76155562 on Build GRCh38
rs764629325 Chr.17: 76155612-76155612 on Build GRCh38
rs375336058 Chr.17: 76155540-76155540 on Build GRCh38
rs191102088 Chr.17: 76155923-76155923 on Build GRCh38
rs895701872 Chr.17: 76155631-76155631 on Build GRCh38
rs772165406 Chr.17: 76155555-76155555 on Build GRCh38
rs763606879 Chr.17: 76155763-76155763 on Build GRCh38

View Full Product Details