Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 35945614-35946128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGO3
Entrez Gene ID
192669
Gene Name
argonaute 3, RISC catalytic component
Gene Aliases
EIF2C3
UniGene
Hs.657659
Gene Chromosome Location
Chr.1: 35925198-36072037 on Build GRCh38
Transcripts
NM_024852.3 NM_177422.2 XM_005270575.3 XM_011540879.2 XM_017000523.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5196856 c.59G>T p.R20I Chr.1: 35945731-35945731 on Build GRCh38
7316593 c.144C>T p.L48L Chr.1: 35945816-35945816 on Build GRCh38
2080925 c.46A>G p.M16V Chr.1: 35945718-35945718 on Build GRCh38
7354803 c.46A>G p.M16V Chr.1: 35945718-35945718 on Build GRCh38
2080927 c.166G>C p.D56H Chr.1: 35945838-35945838 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745384722 Chr.1: 35945681-35945681 on Build GRCh38
rs746930765 Chr.1: 35945887-35945887 on Build GRCh38
rs747274055 Chr.1: 35945667-35945667 on Build GRCh38
rs1016191240 Chr.1: 35945952-35945952 on Build GRCh38
rs751730136 Chr.1: 35945644-35945644 on Build GRCh38
rs975248824 Chr.1: 35945665-35945668 on Build GRCh38
rs905319578 Chr.1: 35945814-35945814 on Build GRCh38
rs79054898 Chr.1: 35946093-35946093 on Build GRCh38
rs781069996 Chr.1: 35945899-35945899 on Build GRCh38
rs780074752 Chr.1: 35945683-35945683 on Build GRCh38
rs377527566 Chr.1: 35946060-35946060 on Build GRCh38
rs767870379 Chr.1: 35945740-35945740 on Build GRCh38
rs749863232 Chr.1: 35945857-35945857 on Build GRCh38
rs903354797 Chr.1: 35945898-35945898 on Build GRCh38
rs138986922 Chr.1: 35946096-35946096 on Build GRCh38
rs933438628 Chr.1: 35945906-35945906 on Build GRCh38
rs768780679 Chr.1: 35945896-35945896 on Build GRCh38
rs1009213582 Chr.1: 35945920-35945920 on Build GRCh38
rs755375459 Chr.1: 35945648-35945648 on Build GRCh38
rs562147491 Chr.1: 35945976-35945976 on Build GRCh38
rs756833758 Chr.1: 35945763-35945763 on Build GRCh38
rs1033403607 Chr.1: 35946085-35946085 on Build GRCh38
rs761219693 Chr.1: 35945747-35945747 on Build GRCh38
rs986753481 Chr.1: 35945966-35945966 on Build GRCh38
rs778149832 Chr.1: 35945668-35945668 on Build GRCh38
rs1056066740 Chr.1: 35945715-35945715 on Build GRCh38
rs781648693 Chr.1: 35945667-35945668 on Build GRCh38
rs747971290 Chr.1: 35945900-35945900 on Build GRCh38
rs770270625 Chr.1: 35945898-35945898 on Build GRCh38
rs899039691 Chr.1: 35945954-35945954 on Build GRCh38
rs1051780453 Chr.1: 35945909-35945909 on Build GRCh38
rs141269853 Chr.1: 35945701-35945701 on Build GRCh38
rs778279355 Chr.1: 35945678-35945678 on Build GRCh38
rs200388327 Chr.1: 35945666-35945667 on Build GRCh38
rs757898372 Chr.1: 35945881-35945881 on Build GRCh38
rs748537371 Chr.1: 35945773-35945776 on Build GRCh38
rs753217465 Chr.1: 35945744-35945744 on Build GRCh38
rs377039064 Chr.1: 35945753-35945753 on Build GRCh38
rs11807761 Chr.1: 35945937-35945937 on Build GRCh38
rs981880357 Chr.1: 35946079-35946079 on Build GRCh38
rs768553614 Chr.1: 35945697-35945697 on Build GRCh38
rs970535127 Chr.1: 35945979-35945979 on Build GRCh38
rs889141601 Chr.1: 35945922-35945922 on Build GRCh38
rs560343792 Chr.1: 35945916-35945916 on Build GRCh38
rs759834155 Chr.1: 35946012-35946012 on Build GRCh38
rs368054104 Chr.1: 35945664-35945664 on Build GRCh38
rs1003807450 Chr.1: 35945839-35945839 on Build GRCh38
rs147640766 Chr.1: 35945724-35945724 on Build GRCh38
rs756501447 Chr.1: 35945665-35945665 on Build GRCh38
rs768080657 Chr.1: 35945659-35945659 on Build GRCh38
rs139647049 Chr.1: 35945709-35945709 on Build GRCh38
rs372140569 Chr.1: 35945720-35945720 on Build GRCh38
rs1036399021 Chr.1: 35946058-35946058 on Build GRCh38
rs1021907303 Chr.1: 35945936-35945936 on Build GRCh38
rs748912666 Chr.1: 35945675-35945675 on Build GRCh38
rs939904261 Chr.1: 35946008-35946008 on Build GRCh38
rs754265595 Chr.1: 35945760-35945760 on Build GRCh38
rs771285091 Chr.1: 35945717-35945717 on Build GRCh38
rs373911284 Chr.1: 35945883-35945883 on Build GRCh38
rs975090745 Chr.1: 35945941-35945941 on Build GRCh38
rs372188316 Chr.1: 35945897-35945897 on Build GRCh38
rs112026957 Chr.1: 35945964-35945964 on Build GRCh38
rs746987961 Chr.1: 35945696-35945696 on Build GRCh38
rs759682013 Chr.1: 35945725-35945725 on Build GRCh38
rs369710801 Chr.1: 35945812-35945812 on Build GRCh38

View Full Product Details