Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 36004263-36004761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGO3
Entrez Gene ID
192669
Gene Name
argonaute 3, RISC catalytic component
Gene Aliases
EIF2C3
UniGene
Hs.657659
Gene Chromosome Location
Chr.1: 35925198-36072037 on Build GRCh38
Transcripts
NM_024852.3 NM_177422.2 XM_005270575.3 XM_005270576.3 XM_011540879.2 XM_011540882.2 XM_017000523.1 XM_017000524.1 XM_017000525.1 XM_017000526.1 XM_017000527.1 XM_017000528.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5380824 c.724C>T p.H242Y Chr.1: 36004406-36004406 on Build GRCh38
5380823 c.670G>T p.A224S Chr.1: 36004352-36004352 on Build GRCh38
6126437 c.705G>T p.M235I Chr.1: 36004387-36004387 on Build GRCh38
4834526 c.751C>G p.L251V Chr.1: 36004433-36004433 on Build GRCh38
5006796 c.699G>T p.Q233H Chr.1: 36004381-36004381 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs973274595 Chr.1: 36004667-36004667 on Build GRCh38
rs780857302 Chr.1: 36004516-36004516 on Build GRCh38
rs769273911 Chr.1: 36004520-36004520 on Build GRCh38
rs1031787461 Chr.1: 36004530-36004530 on Build GRCh38
rs368190615 Chr.1: 36004409-36004409 on Build GRCh38
rs954940400 Chr.1: 36004583-36004583 on Build GRCh38
rs766071931 Chr.1: 36004435-36004435 on Build GRCh38
rs867444535 Chr.1: 36004347-36004347 on Build GRCh38
rs369030612 Chr.1: 36004449-36004449 on Build GRCh38
rs775946785 Chr.1: 36004402-36004402 on Build GRCh38
rs773104942 Chr.1: 36004424-36004424 on Build GRCh38
rs964630457 Chr.1: 36004644-36004644 on Build GRCh38
rs1008943372 Chr.1: 36004617-36004617 on Build GRCh38
rs750942436 Chr.1: 36004506-36004506 on Build GRCh38
rs201650125 Chr.1: 36004574-36004574 on Build GRCh38
rs753920556 Chr.1: 36004480-36004480 on Build GRCh38
rs554231943 Chr.1: 36004510-36004510 on Build GRCh38
rs917902990 Chr.1: 36004563-36004563 on Build GRCh38
rs764385662 Chr.1: 36004478-36004478 on Build GRCh38
rs757443090 Chr.1: 36004491-36004491 on Build GRCh38
rs746638136 Chr.1: 36004316-36004316 on Build GRCh38
rs13374569 Chr.1: 36004677-36004677 on Build GRCh38
rs762928869 Chr.1: 36004431-36004431 on Build GRCh38
rs775486443 Chr.1: 36004505-36004505 on Build GRCh38
rs1005314472 Chr.1: 36004308-36004309 on Build GRCh38
rs376651394 Chr.1: 36004517-36004517 on Build GRCh38
rs1017607979 Chr.1: 36004618-36004618 on Build GRCh38
rs780527530 Chr.1: 36004308-36004308 on Build GRCh38
rs375455260 Chr.1: 36004291-36004291 on Build GRCh38
rs765445555 Chr.1: 36004498-36004498 on Build GRCh38
rs768360545 Chr.1: 36004380-36004380 on Build GRCh38
rs772764459 Chr.1: 36004521-36004521 on Build GRCh38
rs536359237 Chr.1: 36004733-36004733 on Build GRCh38
rs774266191 Chr.1: 36004436-36004436 on Build GRCh38
rs1034258905 Chr.1: 36004448-36004448 on Build GRCh38
rs769683388 Chr.1: 36004416-36004416 on Build GRCh38
rs372029316 Chr.1: 36004508-36004508 on Build GRCh38
rs1013090060 Chr.1: 36004381-36004381 on Build GRCh38
rs747683465 Chr.1: 36004407-36004407 on Build GRCh38
rs372792201 Chr.1: 36004495-36004495 on Build GRCh38
rs911498776 Chr.1: 36004679-36004679 on Build GRCh38
rs970718969 Chr.1: 36004610-36004610 on Build GRCh38
rs758802465 Chr.1: 36004294-36004294 on Build GRCh38
rs747414168 Chr.1: 36004515-36004515 on Build GRCh38
rs1025355649 Chr.1: 36004293-36004294 on Build GRCh38
rs924024739 Chr.1: 36004643-36004643 on Build GRCh38

View Full Product Details