Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 37537412-37537921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNIP1
Entrez Gene ID
79753
Gene Name
Smad nuclear interacting protein 1
Gene Aliases
PMRED
UniGene
Hs.47232
Gene Chromosome Location
Chr.1: 37534449-37554344 on Build GRCh38
Transcripts
NM_024700.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5780995 c.1074C>G p.L358L Chr.1: 37537865-37537865 on Build GRCh38
4007574 c.1147G>C p.D383H Chr.1: 37537792-37537792 on Build GRCh38
6482441 c.1112A>T p.H371L Chr.1: 37537827-37537827 on Build GRCh38
7268712 c.1101C>T p.Y367Y Chr.1: 37537838-37537838 on Build GRCh38
4007573 c.1154A>T p.E385V Chr.1: 37537785-37537785 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146974230 Chr.1: 37537783-37537783 on Build GRCh38
rs902827526 Chr.1: 37537638-37537638 on Build GRCh38
rs997539324 Chr.1: 37537729-37537729 on Build GRCh38
rs557256113 Chr.1: 37537520-37537520 on Build GRCh38
rs991241477 Chr.1: 37537543-37537543 on Build GRCh38
rs779068654 Chr.1: 37537827-37537827 on Build GRCh38
rs541651314 Chr.1: 37537512-37537512 on Build GRCh38
rs545664149 Chr.1: 37537628-37537628 on Build GRCh38
rs775819004 Chr.1: 37537756-37537759 on Build GRCh38
rs966018875 Chr.1: 37537472-37537472 on Build GRCh38
rs911196290 Chr.1: 37537538-37537538 on Build GRCh38
rs548475556 Chr.1: 37537817-37537817 on Build GRCh38
rs893322775 Chr.1: 37537667-37537667 on Build GRCh38
rs909729471 Chr.1: 37537510-37537510 on Build GRCh38
rs766161935 Chr.1: 37537796-37537796 on Build GRCh38
rs199853899 Chr.1: 37537611-37537611 on Build GRCh38
rs957964912 Chr.1: 37537761-37537761 on Build GRCh38
rs936700946 Chr.1: 37537508-37537509 on Build GRCh38
rs138931343 Chr.1: 37537475-37537475 on Build GRCh38
rs559016982 Chr.1: 37537452-37537452 on Build GRCh38
rs368571136 Chr.1: 37537894-37537894 on Build GRCh38
rs775783956 Chr.1: 37537652-37537671 on Build GRCh38
rs111594144 Chr.1: 37537717-37537717 on Build GRCh38
rs768997906 Chr.1: 37537721-37537721 on Build GRCh38
rs553457093 Chr.1: 37537488-37537488 on Build GRCh38
rs764552168 Chr.1: 37537767-37537769 on Build GRCh38
rs917761478 Chr.1: 37537450-37537450 on Build GRCh38
rs754296336 Chr.1: 37537766-37537767 on Build GRCh38
rs375630499 Chr.1: 37537718-37537718 on Build GRCh38
rs949775423 Chr.1: 37537612-37537612 on Build GRCh38
rs958988187 Chr.1: 37537675-37537675 on Build GRCh38
rs745872132 Chr.1: 37537887-37537887 on Build GRCh38
rs574622753 Chr.1: 37537501-37537501 on Build GRCh38
rs781109084 Chr.1: 37537819-37537819 on Build GRCh38
rs751508731 Chr.1: 37537799-37537799 on Build GRCh38
rs561622791 Chr.1: 37537677-37537677 on Build GRCh38
rs387906986 Chr.1: 37537842-37537842 on Build GRCh38
rs1041174049 Chr.1: 37537818-37537818 on Build GRCh38
rs143680314 Chr.1: 37537791-37537791 on Build GRCh38
rs575425926 Chr.1: 37537536-37537536 on Build GRCh38
rs901969191 Chr.1: 37537726-37537726 on Build GRCh38
rs535133251 Chr.1: 37537476-37537476 on Build GRCh38
rs897354658 Chr.1: 37537632-37537633 on Build GRCh38
rs772823656 Chr.1: 37537693-37537693 on Build GRCh38
rs368368736 Chr.1: 37537781-37537781 on Build GRCh38
rs866874156 Chr.1: 37537804-37537804 on Build GRCh38
rs772192233 Chr.1: 37537891-37537891 on Build GRCh38
rs539940441 Chr.1: 37537657-37537657 on Build GRCh38
rs774360369 Chr.1: 37537838-37537838 on Build GRCh38
rs570205503 Chr.1: 37537820-37537820 on Build GRCh38
rs769906071 Chr.1: 37537719-37537719 on Build GRCh38
rs528034219 Chr.1: 37537654-37537654 on Build GRCh38
rs944127078 Chr.1: 37537579-37537579 on Build GRCh38
rs761093271 Chr.1: 37537762-37537764 on Build GRCh38
rs776315069 Chr.1: 37537730-37537730 on Build GRCh38
rs758692166 Chr.1: 37537776-37537781 on Build GRCh38
rs142861495 Chr.1: 37537647-37537647 on Build GRCh38
rs992899386 Chr.1: 37537651-37537652 on Build GRCh38
rs16824209 Chr.1: 37537592-37537592 on Build GRCh38
rs746414136 Chr.1: 37537711-37537711 on Build GRCh38
rs915632305 Chr.1: 37537585-37537585 on Build GRCh38
rs780076866 Chr.1: 37537797-37537800 on Build GRCh38
rs1013345412 Chr.1: 37537691-37537694 on Build GRCh38
rs61755314 Chr.1: 37537821-37537821 on Build GRCh38
rs950749390 Chr.1: 37537778-37537778 on Build GRCh38
rs775496140 Chr.1: 37537766-37537766 on Build GRCh38
rs372672826 Chr.1: 37537794-37537794 on Build GRCh38
rs1026732008 Chr.1: 37537486-37537486 on Build GRCh38
rs1033364266 Chr.1: 37537668-37537668 on Build GRCh38
rs966609876 Chr.1: 37537714-37537714 on Build GRCh38

View Full Product Details